Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 8911047-8911644 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKIP1
Entrez Gene ID
56672
Gene Name
A-kinase interacting protein 1
Gene Aliases
BCA3, C11orf17
UniGene
Hs.131180
Gene Chromosome Location
Chr.11: 8911117-8920079 on Build GRCh38
Transcripts
NM_001206646.1 NM_001206647.1 NM_001206648.1 NM_020642.3 NR_045417.1 NR_045418.1 XM_017018011.1 XM_017018012.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1703579 c.55C>T p.Q19* Chr.11: 8911504-8911504 on Build GRCh38
4973185 c.173A>G p.H58R Chr.11: 8911622-8911622 on Build GRCh38
6416587 c.194C>G p.A65G Chr.11: 8911643-8911643 on Build GRCh38
6429212 c.131G>A p.R44H Chr.11: 8911580-8911580 on Build GRCh38
3935662 c.180G>C p.E60D Chr.11: 8911629-8911629 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780983316 Chr.11: 8911481-8911481 on Build GRCh38
rs375798637 Chr.11: 8911429-8911429 on Build GRCh38
rs916450068 Chr.11: 8911292-8911292 on Build GRCh38
rs114590997 Chr.11: 8911150-8911150 on Build GRCh38
rs771060497 Chr.11: 8911504-8911504 on Build GRCh38
rs749921346 Chr.11: 8911467-8911467 on Build GRCh38
rs7108205 Chr.11: 8911466-8911466 on Build GRCh38
rs973947147 Chr.11: 8911622-8911622 on Build GRCh38
rs990700013 Chr.11: 8911598-8911598 on Build GRCh38
rs1034722082 Chr.11: 8911519-8911519 on Build GRCh38
rs574547140 Chr.11: 8911274-8911274 on Build GRCh38
rs79723679 Chr.11: 8911415-8911415 on Build GRCh38
rs939136833 Chr.11: 8911174-8911174 on Build GRCh38
rs1051263698 Chr.11: 8911224-8911224 on Build GRCh38
rs185296496 Chr.11: 8911297-8911297 on Build GRCh38
rs769923554 Chr.11: 8911421-8911421 on Build GRCh38
rs1029857104 Chr.11: 8911175-8911175 on Build GRCh38
rs200231500 Chr.11: 8911463-8911463 on Build GRCh38
rs7927697 Chr.11: 8911207-8911207 on Build GRCh38
rs2568026 Chr.11: 8911186-8911186 on Build GRCh38
rs960688087 Chr.11: 8911231-8911231 on Build GRCh38
rs978381810 Chr.11: 8911153-8911153 on Build GRCh38
rs1011108569 Chr.11: 8911252-8911252 on Build GRCh38
rs893261543 Chr.11: 8911444-8911444 on Build GRCh38
rs943237416 Chr.11: 8911291-8911291 on Build GRCh38
rs369093977 Chr.11: 8911472-8911472 on Build GRCh38
rs749350396 Chr.11: 8911503-8911503 on Build GRCh38
rs886121597 Chr.11: 8911339-8911339 on Build GRCh38
rs116521366 Chr.11: 8911457-8911457 on Build GRCh38
rs994183739 Chr.11: 8911223-8911223 on Build GRCh38
rs887383777 Chr.11: 8911177-8911177 on Build GRCh38
rs11042121 Chr.11: 8911093-8911093 on Build GRCh38
rs943104356 Chr.11: 8911118-8911118 on Build GRCh38
rs917926510 Chr.11: 8911373-8911373 on Build GRCh38
rs903703749 Chr.11: 8911113-8911113 on Build GRCh38
rs779549139 Chr.11: 8911523-8911524 on Build GRCh38
rs768073181 Chr.11: 8911225-8911225 on Build GRCh38
rs891360433 Chr.11: 8911413-8911413 on Build GRCh38
rs545970574 Chr.11: 8911354-8911354 on Build GRCh38
rs539571491 Chr.11: 8911155-8911155 on Build GRCh38
rs533464078 Chr.11: 8911595-8911595 on Build GRCh38
rs1022054766 Chr.11: 8911255-8911255 on Build GRCh38
rs114591976 Chr.11: 8911273-8911273 on Build GRCh38
rs566810408 Chr.11: 8911609-8911609 on Build GRCh38
rs367819323 Chr.11: 8911523-8911523 on Build GRCh38
rs752010298 Chr.11: 8911447-8911447 on Build GRCh38
rs142557297 Chr.11: 8911525-8911525 on Build GRCh38
rs562271182 Chr.11: 8911081-8911081 on Build GRCh38
rs962839013 Chr.11: 8911607-8911607 on Build GRCh38
rs144370960 Chr.11: 8911604-8911604 on Build GRCh38
rs762546108 Chr.11: 8911561-8911561 on Build GRCh38
rs766405370 Chr.11: 8911474-8911474 on Build GRCh38
rs756474559 Chr.11: 8911366-8911366 on Build GRCh38
rs932970722 Chr.11: 8911311-8911311 on Build GRCh38
rs572199842 Chr.11: 8911368-8911368 on Build GRCh38
rs1133833 Chr.11: 8911517-8911517 on Build GRCh38
rs141902783 Chr.11: 8911237-8911237 on Build GRCh38
rs534878273 Chr.11: 8911240-8911240 on Build GRCh38
rs748915072 Chr.11: 8911476-8911476 on Build GRCh38
rs891391527 Chr.11: 8911227-8911227 on Build GRCh38
rs778965463 Chr.11: 8911505-8911505 on Build GRCh38
rs777961183 Chr.11: 8911590-8911590 on Build GRCh38
rs754877978 Chr.11: 8911479-8911479 on Build GRCh38
rs977795725 Chr.11: 8911267-8911267 on Build GRCh38
rs933044487 Chr.11: 8911617-8911618 on Build GRCh38
rs958662873 Chr.11: 8911277-8911277 on Build GRCh38
rs754876313 Chr.11: 8911408-8911408 on Build GRCh38
rs780337952 Chr.11: 8911389-8911389 on Build GRCh38
rs979770501 Chr.11: 8911280-8911280 on Build GRCh38
rs752458696 Chr.11: 8911484-8911484 on Build GRCh38
rs202156920 Chr.11: 8911600-8911600 on Build GRCh38
rs370192864 Chr.11: 8911327-8911327 on Build GRCh38
rs199973159 Chr.11: 8911515-8911515 on Build GRCh38
rs897113332 Chr.11: 8911213-8911213 on Build GRCh38
rs140538679 Chr.11: 8911566-8911566 on Build GRCh38
rs774890785 Chr.11: 8911432-8911432 on Build GRCh38
rs376220668 Chr.11: 8911279-8911279 on Build GRCh38
rs990647720 Chr.11: 8911283-8911283 on Build GRCh38
rs748337428 Chr.11: 8911416-8911416 on Build GRCh38
rs112956363 Chr.11: 8911623-8911623 on Build GRCh38
rs767387556 Chr.11: 8911578-8911578 on Build GRCh38
rs148414499 Chr.11: 8911518-8911518 on Build GRCh38
rs916647205 Chr.11: 8911191-8911191 on Build GRCh38
rs572763295 Chr.11: 8911414-8911414 on Build GRCh38
rs1051074787 Chr.11: 8911349-8911349 on Build GRCh38
rs964460527 Chr.11: 8911092-8911092 on Build GRCh38
rs11550425 Chr.11: 8911581-8911581 on Build GRCh38
rs997174208 Chr.11: 8911164-8911164 on Build GRCh38
rs557228700 Chr.11: 8911302-8911302 on Build GRCh38
rs769464587 Chr.11: 8911553-8911553 on Build GRCh38
rs960634457 Chr.11: 8911278-8911278 on Build GRCh38
rs759883274 Chr.11: 8911453-8911453 on Build GRCh38
rs1038776267 Chr.11: 8911294-8911294 on Build GRCh38
rs765921319 Chr.11: 8911563-8911563 on Build GRCh38
rs971171986 Chr.11: 8911392-8911392 on Build GRCh38
rs754128869 Chr.11: 8911597-8911597 on Build GRCh38
rs1038821429 Chr.11: 8911320-8911320 on Build GRCh38
rs561103887 Chr.11: 8911557-8911557 on Build GRCh38
rs980842450 Chr.11: 8911188-8911188 on Build GRCh38
rs1039200926 Chr.11: 8911146-8911146 on Build GRCh38
rs773054370 Chr.11: 8911559-8911559 on Build GRCh38
rs755915688 Chr.11: 8911583-8911583 on Build GRCh38
rs150966123 Chr.11: 8911541-8911541 on Build GRCh38
rs918697796 Chr.11: 8911324-8911324 on Build GRCh38
rs925226655 Chr.11: 8911586-8911586 on Build GRCh38
rs111610807 Chr.11: 8911442-8911442 on Build GRCh38
rs748815079 Chr.11: 8911595-8911596 on Build GRCh38
rs914932885 Chr.11: 8911131-8911131 on Build GRCh38
rs756322711 Chr.11: 8911485-8911485 on Build GRCh38
rs966742248 Chr.11: 8911266-8911266 on Build GRCh38
rs913852011 Chr.11: 8911093-8911093 on Build GRCh38
rs771400184 Chr.11: 8911431-8911431 on Build GRCh38
rs927870974 Chr.11: 8911157-8911157 on Build GRCh38
rs941480433 Chr.11: 8911196-8911196 on Build GRCh38
rs1043589097 Chr.11: 8911108-8911108 on Build GRCh38
rs1017076576 Chr.11: 8911183-8911183 on Build GRCh38
rs767853742 Chr.11: 8911455-8911455 on Build GRCh38
rs759469180 Chr.11: 8911568-8911568 on Build GRCh38
rs745961097 Chr.11: 8911624-8911624 on Build GRCh38
rs201278953 Chr.11: 8911411-8911411 on Build GRCh38
rs755735471 Chr.11: 8911442-8911443 on Build GRCh38
rs777902254 Chr.11: 8911500-8911500 on Build GRCh38
rs964492037 Chr.11: 8911251-8911251 on Build GRCh38
rs773295105 Chr.11: 8911426-8911426 on Build GRCh38

View Full Product Details