Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 58709930-58710388 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLYAT
Entrez Gene ID
10249
Gene Name
glycine-N-acyltransferase
Gene Aliases
ACGNAT, CAT, GAT
UniGene
Hs.145384
Gene Chromosome Location
Chr.11: 58708757-58731974 on Build GRCh38
Transcripts
NM_005838.3 NM_201648.2 XM_017017087.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1704047 c.614C>T p.P205L Chr.11: 58710043-58710043 on Build GRCh38
5773446 c.717C>T p.L239L Chr.11: 58709940-58709940 on Build GRCh38
928907 c.701T>C p.L234S Chr.11: 58709956-58709956 on Build GRCh38
7416320 c.680A>T p.E227V Chr.11: 58709977-58709977 on Build GRCh38
5932770 c.631G>A p.G211R Chr.11: 58710026-58710026 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376146569 Chr.11: 58709992-58709992 on Build GRCh38
rs1012981216 Chr.11: 58710232-58710232 on Build GRCh38
rs745647355 Chr.11: 58710200-58710200 on Build GRCh38
rs577561815 Chr.11: 58710091-58710091 on Build GRCh38
rs368483264 Chr.11: 58710149-58710149 on Build GRCh38
rs999097247 Chr.11: 58710313-58710313 on Build GRCh38
rs376369879 Chr.11: 58710204-58710204 on Build GRCh38
rs1048243039 Chr.11: 58710252-58710252 on Build GRCh38
rs750126135 Chr.11: 58710167-58710167 on Build GRCh38
rs763046132 Chr.11: 58710004-58710004 on Build GRCh38
rs958924955 Chr.11: 58710013-58710013 on Build GRCh38
rs553813078 Chr.11: 58710201-58710201 on Build GRCh38
rs868500192 Chr.11: 58709956-58709956 on Build GRCh38
rs147691659 Chr.11: 58710094-58710094 on Build GRCh38
rs962074796 Chr.11: 58710228-58710228 on Build GRCh38
rs956397604 Chr.11: 58710346-58710346 on Build GRCh38
rs1045360 Chr.11: 58710145-58710145 on Build GRCh38
rs2509910 Chr.11: 58710212-58710212 on Build GRCh38
rs942436085 Chr.11: 58710090-58710090 on Build GRCh38
rs887143567 Chr.11: 58710156-58710156 on Build GRCh38
rs763642751 Chr.11: 58710055-58710055 on Build GRCh38
rs199765405 Chr.11: 58710032-58710032 on Build GRCh38
rs751031634 Chr.11: 58710035-58710035 on Build GRCh38
rs866007858 Chr.11: 58710199-58710199 on Build GRCh38
rs751843213 Chr.11: 58710116-58710116 on Build GRCh38
rs760274079 Chr.11: 58710028-58710028 on Build GRCh38
rs930735997 Chr.11: 58710243-58710243 on Build GRCh38
rs201464772 Chr.11: 58709953-58709953 on Build GRCh38
rs578126220 Chr.11: 58710237-58710237 on Build GRCh38
rs866591913 Chr.11: 58710073-58710073 on Build GRCh38
rs757165153 Chr.11: 58710164-58710164 on Build GRCh38
rs1029141555 Chr.11: 58710095-58710095 on Build GRCh38
rs760252024 Chr.11: 58710085-58710085 on Build GRCh38
rs762730101 Chr.11: 58710002-58710002 on Build GRCh38
rs755407444 Chr.11: 58710129-58710129 on Build GRCh38
rs920706823 Chr.11: 58710234-58710234 on Build GRCh38
rs866073131 Chr.11: 58710300-58710300 on Build GRCh38
rs573953220 Chr.11: 58710342-58710342 on Build GRCh38
rs761807213 Chr.11: 58710034-58710036 on Build GRCh38
rs754284049 Chr.11: 58710316-58710316 on Build GRCh38
rs145499766 Chr.11: 58710061-58710061 on Build GRCh38
rs766712827 Chr.11: 58710111-58710111 on Build GRCh38
rs768718220 Chr.11: 58709966-58709966 on Build GRCh38
rs138125182 Chr.11: 58710062-58710062 on Build GRCh38
rs767626591 Chr.11: 58710130-58710130 on Build GRCh38
rs371787235 Chr.11: 58710071-58710071 on Build GRCh38
rs373776314 Chr.11: 58709973-58709973 on Build GRCh38
rs752125820 Chr.11: 58710023-58710023 on Build GRCh38
rs192235966 Chr.11: 58709952-58709952 on Build GRCh38
rs778950033 Chr.11: 58710166-58710166 on Build GRCh38
rs140835375 Chr.11: 58710133-58710133 on Build GRCh38
rs906158005 Chr.11: 58710318-58710318 on Build GRCh38
rs775281871 Chr.11: 58710083-58710083 on Build GRCh38
rs535306848 Chr.11: 58710036-58710036 on Build GRCh38
rs780414226 Chr.11: 58709964-58709964 on Build GRCh38
rs755173392 Chr.11: 58710056-58710056 on Build GRCh38
rs1045357 Chr.11: 58710194-58710194 on Build GRCh38
rs749236606 Chr.11: 58710063-58710063 on Build GRCh38
rs147885245 Chr.11: 58710008-58710008 on Build GRCh38
rs771594775 Chr.11: 58710076-58710076 on Build GRCh38
rs894566495 Chr.11: 58710213-58710213 on Build GRCh38
rs372428189 Chr.11: 58710176-58710176 on Build GRCh38
rs538149430 Chr.11: 58710039-58710039 on Build GRCh38
rs1045359 Chr.11: 58710153-58710153 on Build GRCh38
rs1045358 Chr.11: 58710193-58710193 on Build GRCh38
rs753019490 Chr.11: 58710134-58710134 on Build GRCh38
rs370913726 Chr.11: 58709958-58709958 on Build GRCh38
rs770645645 Chr.11: 58710067-58710067 on Build GRCh38
rs376065098 Chr.11: 58710137-58710137 on Build GRCh38
rs774950986 Chr.11: 58710206-58710206 on Build GRCh38
rs947948054 Chr.11: 58710297-58710297 on Build GRCh38
rs533940812 Chr.11: 58709965-58709965 on Build GRCh38
rs977606176 Chr.11: 58710038-58710038 on Build GRCh38
rs767050602 Chr.11: 58710146-58710146 on Build GRCh38
rs537474337 Chr.11: 58710274-58710274 on Build GRCh38
rs538282101 Chr.11: 58710177-58710177 on Build GRCh38
rs561947788 Chr.11: 58710295-58710295 on Build GRCh38
rs763589399 Chr.11: 58710102-58710102 on Build GRCh38
rs933902924 Chr.11: 58710141-58710141 on Build GRCh38
rs935025542 Chr.11: 58710325-58710325 on Build GRCh38
rs866202019 Chr.11: 58710016-58710016 on Build GRCh38
rs777834138 Chr.11: 58710148-58710148 on Build GRCh38
rs1046312280 Chr.11: 58710298-58710298 on Build GRCh38
rs769693616 Chr.11: 58709982-58709982 on Build GRCh38
rs141524728 Chr.11: 58710011-58710011 on Build GRCh38

View Full Product Details