Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 47247404-47247897 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACP2
Entrez Gene ID
53
Gene Name
acid phosphatase 2, lysosomal
Gene Aliases
LAP
UniGene
Hs.532492
Gene Chromosome Location
Chr.11: 47239302-47248847 on Build GRCh38
Transcripts
NM_001302489.1 NM_001302490.1 NM_001302491.1 NM_001302492.1 NM_001610.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3447757 c.262C>T p.H88Y Chr.11: 47247676-47247676 on Build GRCh38
1727111 c.268T>G p.F90V Chr.11: 47247670-47247670 on Build GRCh38
5838641 c.251G>A p.R84Q Chr.11: 47247687-47247687 on Build GRCh38
1181638 c.256C>T p.R86C Chr.11: 47247682-47247682 on Build GRCh38
6132708 c.247C>G p.L83V Chr.11: 47247691-47247691 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1054746886 Chr.11: 47247497-47247497 on Build GRCh38
rs778061039 Chr.11: 47247661-47247661 on Build GRCh38
rs376581324 Chr.11: 47247757-47247757 on Build GRCh38
rs372934577 Chr.11: 47247770-47247770 on Build GRCh38
rs766434078 Chr.11: 47247746-47247746 on Build GRCh38
rs765216967 Chr.11: 47247613-47247613 on Build GRCh38
rs371643693 Chr.11: 47247725-47247725 on Build GRCh38
rs367900188 Chr.11: 47247681-47247681 on Build GRCh38
rs758668203 Chr.11: 47247680-47247681 on Build GRCh38
rs4752973 Chr.11: 47247608-47247608 on Build GRCh38
rs58528246 Chr.11: 47247818-47247818 on Build GRCh38
rs767763638 Chr.11: 47247583-47247583 on Build GRCh38
rs267602900 Chr.11: 47247692-47247692 on Build GRCh38
rs771275960 Chr.11: 47247520-47247520 on Build GRCh38
rs947551990 Chr.11: 47247833-47247833 on Build GRCh38
rs759407660 Chr.11: 47247755-47247755 on Build GRCh38
rs115597089 Chr.11: 47247517-47247517 on Build GRCh38
rs371472212 Chr.11: 47247637-47247637 on Build GRCh38
rs939131323 Chr.11: 47247863-47247863 on Build GRCh38
rs775932737 Chr.11: 47247607-47247607 on Build GRCh38
rs763044082 Chr.11: 47247739-47247739 on Build GRCh38
rs775095071 Chr.11: 47247574-47247574 on Build GRCh38
rs547906234 Chr.11: 47247436-47247436 on Build GRCh38
rs151129607 Chr.11: 47247647-47247647 on Build GRCh38
rs372895853 Chr.11: 47247656-47247656 on Build GRCh38
rs763818091 Chr.11: 47247603-47247603 on Build GRCh38
rs749291287 Chr.11: 47247670-47247670 on Build GRCh38
rs921187565 Chr.11: 47247718-47247718 on Build GRCh38
rs751702166 Chr.11: 47247557-47247557 on Build GRCh38
rs375308541 Chr.11: 47247673-47247673 on Build GRCh38
rs201262321 Chr.11: 47247684-47247684 on Build GRCh38
rs779374515 Chr.11: 47247674-47247674 on Build GRCh38
rs1054715817 Chr.11: 47247866-47247866 on Build GRCh38
rs774874281 Chr.11: 47247521-47247521 on Build GRCh38
rs773275558 Chr.11: 47247735-47247735 on Build GRCh38
rs756234829 Chr.11: 47247658-47247658 on Build GRCh38
rs45603841 Chr.11: 47247578-47247578 on Build GRCh38
rs772476447 Chr.11: 47247556-47247556 on Build GRCh38
rs556408366 Chr.11: 47247713-47247713 on Build GRCh38
rs746185729 Chr.11: 47247526-47247526 on Build GRCh38
rs768150336 Chr.11: 47247716-47247716 on Build GRCh38
rs985472476 Chr.11: 47247648-47247648 on Build GRCh38
rs777799689 Chr.11: 47247722-47247722 on Build GRCh38
rs747401772 Chr.11: 47247688-47247688 on Build GRCh38
rs12285351 Chr.11: 47247798-47247798 on Build GRCh38
rs751247528 Chr.11: 47247751-47247751 on Build GRCh38
rs761206449 Chr.11: 47247726-47247726 on Build GRCh38
rs1015461061 Chr.11: 47247687-47247687 on Build GRCh38
rs769467420 Chr.11: 47247733-47247733 on Build GRCh38
rs999996277 Chr.11: 47247611-47247611 on Build GRCh38
rs371223401 Chr.11: 47247477-47247477 on Build GRCh38
rs758234642 Chr.11: 47247645-47247645 on Build GRCh38
rs748420203 Chr.11: 47247538-47247540 on Build GRCh38
rs934446911 Chr.11: 47247790-47247790 on Build GRCh38
rs929172589 Chr.11: 47247780-47247780 on Build GRCh38
rs564413683 Chr.11: 47247773-47247773 on Build GRCh38
rs199935537 Chr.11: 47247682-47247682 on Build GRCh38

View Full Product Details