Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 63974375-63974933 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COX8A
Entrez Gene ID
1351
Gene Name
cytochrome c oxidase subunit 8A
Gene Aliases
COX, COX8, COX8-2, COX8L, VIII, VIII-L
UniGene
Hs.743989
Gene Chromosome Location
Chr.11: 63974607-63976543 on Build GRCh38
Transcripts
NM_004074.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4837894 c.36G>C p.L12F Chr.11: 63974716-63974716 on Build GRCh38
5360819 c.50G>C p.R17P Chr.11: 63974730-63974730 on Build GRCh38
1202112 c.46G>A p.A16T Chr.11: 63974726-63974726 on Build GRCh38
1298356 c.83C>G p.S28W Chr.11: 63974763-63974763 on Build GRCh38
6246017 c.27G>T p.L9L Chr.11: 63974707-63974707 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs916191228 Chr.11: 63974581-63974581 on Build GRCh38
rs756789173 Chr.11: 63974653-63974653 on Build GRCh38
rs1043197197 Chr.11: 63974804-63974804 on Build GRCh38
rs144339685 Chr.11: 63974747-63974747 on Build GRCh38
rs745767608 Chr.11: 63974654-63974654 on Build GRCh38
rs773161061 Chr.11: 63974783-63974783 on Build GRCh38
rs772015686 Chr.11: 63974699-63974699 on Build GRCh38
rs749102794 Chr.11: 63974678-63974678 on Build GRCh38
rs76083469 Chr.11: 63974419-63974419 on Build GRCh38
rs570320155 Chr.11: 63974881-63974881 on Build GRCh38
rs902937414 Chr.11: 63974560-63974560 on Build GRCh38
rs1039889774 Chr.11: 63974499-63974499 on Build GRCh38
rs974125864 Chr.11: 63974400-63974400 on Build GRCh38
rs751020656 Chr.11: 63974652-63974652 on Build GRCh38
rs570454012 Chr.11: 63974409-63974409 on Build GRCh38
rs991792225 Chr.11: 63974615-63974615 on Build GRCh38
rs755942347 Chr.11: 63974755-63974755 on Build GRCh38
rs138925707 Chr.11: 63974613-63974613 on Build GRCh38
rs1047505724 Chr.11: 63974429-63974429 on Build GRCh38
rs1023698461 Chr.11: 63974880-63974880 on Build GRCh38
rs769493036 Chr.11: 63974687-63974687 on Build GRCh38
rs558637998 Chr.11: 63974416-63974416 on Build GRCh38
rs768605109 Chr.11: 63974769-63974769 on Build GRCh38
rs769823264 Chr.11: 63974656-63974656 on Build GRCh38
rs372550956 Chr.11: 63974803-63974803 on Build GRCh38
rs779713502 Chr.11: 63974661-63974661 on Build GRCh38
rs760414755 Chr.11: 63974879-63974879 on Build GRCh38
rs1026767360 Chr.11: 63974855-63974855 on Build GRCh38
rs891413262 Chr.11: 63974448-63974448 on Build GRCh38
rs750392908 Chr.11: 63974817-63974817 on Build GRCh38
rs199919249 Chr.11: 63974711-63974711 on Build GRCh38
rs148986341 Chr.11: 63974685-63974685 on Build GRCh38
rs774738979 Chr.11: 63974686-63974686 on Build GRCh38
rs528113495 Chr.11: 63974773-63974773 on Build GRCh38
rs947593863 Chr.11: 63974801-63974801 on Build GRCh38
rs11550241 Chr.11: 63974733-63974733 on Build GRCh38
rs868191298 Chr.11: 63974573-63974573 on Build GRCh38
rs961136220 Chr.11: 63974538-63974538 on Build GRCh38
rs766913092 Chr.11: 63974713-63974713 on Build GRCh38
rs368336163 Chr.11: 63974749-63974749 on Build GRCh38
rs753797950 Chr.11: 63974819-63974819 on Build GRCh38
rs749849047 Chr.11: 63974812-63974812 on Build GRCh38
rs1011686373 Chr.11: 63974506-63974506 on Build GRCh38
rs555126304 Chr.11: 63974572-63974572 on Build GRCh38
rs759915558 Chr.11: 63974729-63974729 on Build GRCh38
rs954091915 Chr.11: 63974533-63974533 on Build GRCh38
rs755675454 Chr.11: 63974815-63974815 on Build GRCh38
rs148768920 Chr.11: 63974787-63974787 on Build GRCh38
rs754143036 Chr.11: 63974717-63974717 on Build GRCh38
rs758931792 Chr.11: 63974559-63974559 on Build GRCh38
rs755016237 Chr.11: 63974826-63974826 on Build GRCh38
rs1019443447 Chr.11: 63974537-63974537 on Build GRCh38
rs777415880 Chr.11: 63974649-63974649 on Build GRCh38
rs7130187 Chr.11: 63974702-63974702 on Build GRCh38
rs973216580 Chr.11: 63974709-63974709 on Build GRCh38
rs778838324 Chr.11: 63974841-63974841 on Build GRCh38
rs920048436 Chr.11: 63974734-63974734 on Build GRCh38
rs200786823 Chr.11: 63974810-63974810 on Build GRCh38
rs111725597 Chr.11: 63974892-63974892 on Build GRCh38
rs1008095683 Chr.11: 63974534-63974534 on Build GRCh38
rs761946856 Chr.11: 63974694-63974694 on Build GRCh38
rs561481960 Chr.11: 63974806-63974806 on Build GRCh38
rs373910843 Chr.11: 63974811-63974811 on Build GRCh38
rs534780479 Chr.11: 63974505-63974505 on Build GRCh38
rs772396588 Chr.11: 63974776-63974776 on Build GRCh38
rs1022817952 Chr.11: 63974645-63974645 on Build GRCh38
rs952554190 Chr.11: 63974858-63974858 on Build GRCh38
rs765561443 Chr.11: 63974802-63974802 on Build GRCh38
rs757978708 Chr.11: 63974648-63974648 on Build GRCh38
rs768033056 Chr.11: 63974816-63974816 on Build GRCh38
rs766632161 Chr.11: 63974635-63974635 on Build GRCh38
rs539022073 Chr.11: 63974410-63974410 on Build GRCh38
rs749046278 Chr.11: 63974764-63974764 on Build GRCh38
rs773908478 Chr.11: 63974672-63974672 on Build GRCh38
rs61759492 Chr.11: 63974791-63974791 on Build GRCh38
rs972449609 Chr.11: 63974555-63974555 on Build GRCh38
rs944255928 Chr.11: 63974469-63974469 on Build GRCh38
rs919996247 Chr.11: 63974404-63974404 on Build GRCh38
rs760075029 Chr.11: 63974797-63974797 on Build GRCh38
rs951437054 Chr.11: 63974741-63974741 on Build GRCh38
rs762703645 Chr.11: 63974474-63974474 on Build GRCh38
rs893208869 Chr.11: 63974662-63974668 on Build GRCh38
rs754259525 Chr.11: 63974636-63974636 on Build GRCh38
rs780158354 Chr.11: 63974763-63974763 on Build GRCh38
rs963289242 Chr.11: 63974884-63974884 on Build GRCh38
rs893103428 Chr.11: 63974458-63974458 on Build GRCh38
rs948955208 Chr.11: 63974610-63974610 on Build GRCh38

View Full Product Details