Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 108691683-108692281 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX10
Entrez Gene ID
1662
Gene Name
DEAD-box helicase 10
Gene Aliases
HRH-J8
UniGene
Hs.591931
Gene Chromosome Location
Chr.11: 108665025-108940930 on Build GRCh38
Transcripts
NM_004398.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2111028 c.992G>A p.R331Q Chr.11: 108691892-108691892 on Build GRCh38
5440192 c.1080G>C p.E360D Chr.11: 108691980-108691980 on Build GRCh38
4433390 c.1096G>A p.A366T Chr.11: 108691996-108691996 on Build GRCh38
5983883 c.1105C>T p.L369F Chr.11: 108692005-108692005 on Build GRCh38
7303171 c.1043G>A p.R348Q Chr.11: 108691943-108691943 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201497533 Chr.11: 108692074-108692074 on Build GRCh38
rs767606534 Chr.11: 108691932-108691932 on Build GRCh38
rs531615501 Chr.11: 108692107-108692107 on Build GRCh38
rs765260083 Chr.11: 108691833-108691833 on Build GRCh38
rs776492184 Chr.11: 108691904-108691904 on Build GRCh38
rs748741089 Chr.11: 108692083-108692083 on Build GRCh38
rs978895027 Chr.11: 108691817-108691817 on Build GRCh38
rs780967981 Chr.11: 108691987-108691987 on Build GRCh38
rs766342554 Chr.11: 108691924-108691924 on Build GRCh38
rs550134198 Chr.11: 108692242-108692242 on Build GRCh38
rs777883217 Chr.11: 108691802-108691802 on Build GRCh38
rs756229681 Chr.11: 108691890-108691890 on Build GRCh38
rs752523415 Chr.11: 108692028-108692028 on Build GRCh38
rs960096036 Chr.11: 108691798-108691798 on Build GRCh38
rs995424642 Chr.11: 108691723-108691723 on Build GRCh38
rs780526276 Chr.11: 108692061-108692061 on Build GRCh38
rs987705997 Chr.11: 108692186-108692186 on Build GRCh38
rs202183817 Chr.11: 108691909-108691909 on Build GRCh38
rs35032499 Chr.11: 108691966-108691967 on Build GRCh38
rs757648306 Chr.11: 108691957-108691957 on Build GRCh38
rs759430194 Chr.11: 108691931-108691931 on Build GRCh38
rs1035558246 Chr.11: 108691787-108691787 on Build GRCh38
rs148757704 Chr.11: 108691939-108691939 on Build GRCh38
rs368030768 Chr.11: 108691998-108691998 on Build GRCh38
rs747487799 Chr.11: 108691891-108691891 on Build GRCh38
rs370170694 Chr.11: 108692085-108692085 on Build GRCh38
rs922227272 Chr.11: 108692125-108692128 on Build GRCh38
rs777624816 Chr.11: 108692080-108692080 on Build GRCh38
rs371381151 Chr.11: 108692058-108692058 on Build GRCh38
rs781665971 Chr.11: 108692096-108692096 on Build GRCh38
rs558301275 Chr.11: 108691715-108691715 on Build GRCh38
rs913023739 Chr.11: 108691813-108691813 on Build GRCh38
rs1047735860 Chr.11: 108692156-108692156 on Build GRCh38
rs560196334 Chr.11: 108692020-108692020 on Build GRCh38
rs572103446 Chr.11: 108692048-108692048 on Build GRCh38
rs376588411 Chr.11: 108692049-108692049 on Build GRCh38
rs762534054 Chr.11: 108692000-108692000 on Build GRCh38
rs531114848 Chr.11: 108692155-108692155 on Build GRCh38
rs753919480 Chr.11: 108691949-108691949 on Build GRCh38
rs750986592 Chr.11: 108692056-108692056 on Build GRCh38
rs910957783 Chr.11: 108692072-108692072 on Build GRCh38
rs564938347 Chr.11: 108691883-108691883 on Build GRCh38
rs759232661 Chr.11: 108692021-108692021 on Build GRCh38
rs190268566 Chr.11: 108691880-108691880 on Build GRCh38
rs767253327 Chr.11: 108692022-108692022 on Build GRCh38
rs762755684 Chr.11: 108691910-108691910 on Build GRCh38
rs770527153 Chr.11: 108691903-108691903 on Build GRCh38
rs1023646572 Chr.11: 108691747-108691747 on Build GRCh38
rs910914445 Chr.11: 108691820-108691820 on Build GRCh38
rs369000311 Chr.11: 108692053-108692053 on Build GRCh38
rs560321344 Chr.11: 108692149-108692149 on Build GRCh38
rs779082320 Chr.11: 108691900-108691900 on Build GRCh38
rs764314900 Chr.11: 108692038-108692038 on Build GRCh38
rs910073539 Chr.11: 108692213-108692213 on Build GRCh38
rs765383608 Chr.11: 108691782-108691782 on Build GRCh38
rs200784856 Chr.11: 108691855-108691855 on Build GRCh38
rs760823392 Chr.11: 108692034-108692034 on Build GRCh38
rs758850593 Chr.11: 108692060-108692060 on Build GRCh38
rs199899756 Chr.11: 108691988-108691988 on Build GRCh38
rs771431694 Chr.11: 108691902-108691902 on Build GRCh38
rs932235371 Chr.11: 108691841-108691841 on Build GRCh38
rs759871345 Chr.11: 108691763-108691763 on Build GRCh38
rs988660359 Chr.11: 108692007-108692007 on Build GRCh38
rs779534421 Chr.11: 108691972-108691972 on Build GRCh38
rs749749021 Chr.11: 108691892-108691892 on Build GRCh38
rs746220787 Chr.11: 108692193-108692193 on Build GRCh38
rs746215092 Chr.11: 108691973-108691973 on Build GRCh38
rs888539853 Chr.11: 108691735-108691735 on Build GRCh38
rs899129030 Chr.11: 108691719-108691719 on Build GRCh38
rs1035941935 Chr.11: 108692054-108692054 on Build GRCh38
rs375597924 Chr.11: 108691843-108691843 on Build GRCh38
rs144415130 Chr.11: 108691905-108691905 on Build GRCh38
rs115677566 Chr.11: 108691814-108691814 on Build GRCh38
rs866988761 Chr.11: 108692221-108692221 on Build GRCh38
rs772127493 Chr.11: 108691732-108691732 on Build GRCh38
rs772524933 Chr.11: 108691977-108691977 on Build GRCh38
rs978423234 Chr.11: 108692122-108692122 on Build GRCh38
rs751398563 Chr.11: 108691872-108691872 on Build GRCh38
rs1045073013 Chr.11: 108692004-108692004 on Build GRCh38
rs184682388 Chr.11: 108691832-108691832 on Build GRCh38
rs770758577 Chr.11: 108692001-108692001 on Build GRCh38
rs755087241 Chr.11: 108691881-108691881 on Build GRCh38
rs905635810 Chr.11: 108691880-108691892 on Build GRCh38
rs756282325 Chr.11: 108691856-108691858 on Build GRCh38
rs905796925 Chr.11: 108692011-108692011 on Build GRCh38
rs113143585 Chr.11: 108691779-108691779 on Build GRCh38
rs548283063 Chr.11: 108691806-108691806 on Build GRCh38
rs752862350 Chr.11: 108691786-108691786 on Build GRCh38
rs748209452 Chr.11: 108691885-108691885 on Build GRCh38
rs777793234 Chr.11: 108691943-108691943 on Build GRCh38
rs769475642 Chr.11: 108691908-108691908 on Build GRCh38
rs534409449 Chr.11: 108691807-108691807 on Build GRCh38
rs374973727 Chr.11: 108691938-108691938 on Build GRCh38
rs1006014126 Chr.11: 108691745-108691746 on Build GRCh38
rs151120404 Chr.11: 108692075-108692075 on Build GRCh38
rs752970592 Chr.11: 108691937-108691937 on Build GRCh38

View Full Product Details