Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 102324115-102324706 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BIRC3
Entrez Gene ID
330
Gene Name
baculoviral IAP repeat containing 3
Gene Aliases
AIP1, API2, CIAP2, HAIP1, HIAP1, MALT2, MIHC, RNF49, c-IAP2
UniGene
Hs.127799
Gene Chromosome Location
Chr.11: 102317450-102339403 on Build GRCh38
Transcripts
NM_001165.4 NM_182962.2 XM_017017643.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2108117 c.49G>A p.A17T Chr.11: 102324558-102324558 on Build GRCh38
4990037 c.71A>T p.Y24F Chr.11: 102324580-102324580 on Build GRCh38
1704480 c.118C>T p.P40S Chr.11: 102324627-102324627 on Build GRCh38
1746003 c.73G>T p.D25Y Chr.11: 102324582-102324582 on Build GRCh38
4990038 c.119C>T p.P40L Chr.11: 102324628-102324628 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1050770403 Chr.11: 102324520-102324520 on Build GRCh38
rs1055078 Chr.11: 102324638-102324638 on Build GRCh38
rs772261113 Chr.11: 102324459-102324459 on Build GRCh38
rs751116953 Chr.11: 102324557-102324557 on Build GRCh38
rs373107755 Chr.11: 102324506-102324506 on Build GRCh38
rs1026259447 Chr.11: 102324507-102324507 on Build GRCh38
rs186307109 Chr.11: 102324558-102324558 on Build GRCh38
rs544577616 Chr.11: 102324182-102324182 on Build GRCh38
rs1041070369 Chr.11: 102324494-102324494 on Build GRCh38
rs370738985 Chr.11: 102324582-102324582 on Build GRCh38
rs566629468 Chr.11: 102324149-102324149 on Build GRCh38
rs1006578210 Chr.11: 102324426-102324426 on Build GRCh38
rs978016131 Chr.11: 102324657-102324657 on Build GRCh38
rs961740657 Chr.11: 102324302-102324302 on Build GRCh38
rs570461838 Chr.11: 102324253-102324254 on Build GRCh38
rs902341681 Chr.11: 102324366-102324366 on Build GRCh38
rs898754712 Chr.11: 102324451-102324451 on Build GRCh38
rs971722216 Chr.11: 102324307-102324307 on Build GRCh38
rs376445512 Chr.11: 102324519-102324519 on Build GRCh38
rs762438913 Chr.11: 102324546-102324546 on Build GRCh38
rs375640711 Chr.11: 102324587-102324587 on Build GRCh38
rs879232042 Chr.11: 102324486-102324486 on Build GRCh38
rs766043554 Chr.11: 102324550-102324550 on Build GRCh38
rs927316531 Chr.11: 102324247-102324247 on Build GRCh38
rs761068492 Chr.11: 102324591-102324591 on Build GRCh38
rs1055079 Chr.11: 102324665-102324665 on Build GRCh38
rs764638130 Chr.11: 102324516-102324516 on Build GRCh38
rs762397304 Chr.11: 102324664-102324664 on Build GRCh38
rs181803711 Chr.11: 102324399-102324399 on Build GRCh38
rs144852435 Chr.11: 102324563-102324563 on Build GRCh38
rs775211808 Chr.11: 102324336-102324336 on Build GRCh38
rs1035350946 Chr.11: 102324612-102324612 on Build GRCh38
rs994411767 Chr.11: 102324452-102324452 on Build GRCh38
rs992142363 Chr.11: 102324388-102324388 on Build GRCh38
rs17882391 Chr.11: 102324393-102324393 on Build GRCh38
rs759088779 Chr.11: 102324681-102324681 on Build GRCh38
rs765765029 Chr.11: 102324676-102324676 on Build GRCh38
rs376038375 Chr.11: 102324202-102324202 on Build GRCh38
rs770856613 Chr.11: 102324581-102324581 on Build GRCh38
rs141553919 Chr.11: 102324417-102324417 on Build GRCh38
rs190140643 Chr.11: 102324614-102324614 on Build GRCh38
rs1042648714 Chr.11: 102324358-102324358 on Build GRCh38
rs377518449 Chr.11: 102324482-102324482 on Build GRCh38
rs574202531 Chr.11: 102324435-102324435 on Build GRCh38
rs775814824 Chr.11: 102324588-102324588 on Build GRCh38
rs558145768 Chr.11: 102324277-102324277 on Build GRCh38
rs990087823 Chr.11: 102324285-102324285 on Build GRCh38
rs937357964 Chr.11: 102324276-102324276 on Build GRCh38
rs373862803 Chr.11: 102324255-102324256 on Build GRCh38
rs775866384 Chr.11: 102324466-102324466 on Build GRCh38
rs142523935 Chr.11: 102324540-102324540 on Build GRCh38
rs1028834336 Chr.11: 102324171-102324171 on Build GRCh38
rs1016568838 Chr.11: 102324441-102324441 on Build GRCh38
rs953326771 Chr.11: 102324174-102324174 on Build GRCh38
rs151238797 Chr.11: 102324308-102324308 on Build GRCh38
rs1802410 Chr.11: 102324353-102324353 on Build GRCh38
rs947520471 Chr.11: 102324375-102324375 on Build GRCh38
rs749371276 Chr.11: 102324575-102324575 on Build GRCh38
rs765060822 Chr.11: 102324425-102324425 on Build GRCh38
rs915162818 Chr.11: 102324297-102324297 on Build GRCh38
rs751101959 Chr.11: 102324677-102324677 on Build GRCh38
rs371507673 Chr.11: 102324208-102324208 on Build GRCh38
rs777182261 Chr.11: 102324630-102324630 on Build GRCh38

View Full Product Details