Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 108688632-108689212 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX10
Entrez Gene ID
1662
Gene Name
DEAD-box helicase 10
Gene Aliases
HRH-J8
UniGene
Hs.591931
Gene Chromosome Location
Chr.11: 108665025-108940930 on Build GRCh38
Transcripts
NM_004398.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4145218 c.850A>C p.T284P Chr.11: 108688937-108688937 on Build GRCh38
1351106 c.915T>C p.Y305Y Chr.11: 108689002-108689002 on Build GRCh38
4676732 c.965G>A p.S322N Chr.11: 108689052-108689052 on Build GRCh38
1351104 c.857C>T p.A286V Chr.11: 108688944-108688944 on Build GRCh38
4676731 c.955delT p.S321fs*32 Chr.11: 108689042-108689042 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770224673 Chr.11: 108689091-108689091 on Build GRCh38
rs1034879519 Chr.11: 108688738-108688738 on Build GRCh38
rs764868857 Chr.11: 108689149-108689149 on Build GRCh38
rs751497585 Chr.11: 108688919-108688926 on Build GRCh38
rs995002999 Chr.11: 108688733-108688733 on Build GRCh38
rs544537296 Chr.11: 108689119-108689119 on Build GRCh38
rs754978266 Chr.11: 108689023-108689025 on Build GRCh38
rs910885703 Chr.11: 108688880-108688880 on Build GRCh38
rs867559573 Chr.11: 108689129-108689129 on Build GRCh38
rs370942127 Chr.11: 108688932-108688932 on Build GRCh38
rs772466431 Chr.11: 108688975-108688975 on Build GRCh38
rs369002763 Chr.11: 108688886-108688886 on Build GRCh38
rs780197893 Chr.11: 108688951-108688951 on Build GRCh38
rs568660099 Chr.11: 108688853-108688853 on Build GRCh38
rs899354281 Chr.11: 108688725-108688725 on Build GRCh38
rs1055841681 Chr.11: 108689001-108689001 on Build GRCh38
rs753770763 Chr.11: 108688922-108688922 on Build GRCh38
rs557182697 Chr.11: 108689181-108689181 on Build GRCh38
rs770314563 Chr.11: 108688970-108688970 on Build GRCh38
rs191049993 Chr.11: 108688906-108688906 on Build GRCh38
rs940319967 Chr.11: 108689116-108689116 on Build GRCh38
rs773777930 Chr.11: 108689092-108689092 on Build GRCh38
rs777737074 Chr.11: 108688921-108688921 on Build GRCh38
rs936425413 Chr.11: 108688747-108688747 on Build GRCh38
rs750229642 Chr.11: 108689036-108689036 on Build GRCh38
rs759245506 Chr.11: 108689041-108689041 on Build GRCh38
rs548708347 Chr.11: 108688824-108688824 on Build GRCh38
rs199608454 Chr.11: 108688968-108688968 on Build GRCh38
rs12283078 Chr.11: 108689185-108689185 on Build GRCh38
rs774940938 Chr.11: 108689150-108689150 on Build GRCh38
rs776223851 Chr.11: 108689107-108689107 on Build GRCh38
rs748615841 Chr.11: 108688775-108688775 on Build GRCh38
rs535295905 Chr.11: 108689121-108689121 on Build GRCh38
rs780290286 Chr.11: 108689070-108689070 on Build GRCh38
rs760207402 Chr.11: 108689103-108689103 on Build GRCh38
rs760212873 Chr.11: 108689002-108689002 on Build GRCh38
rs777171897 Chr.11: 108688962-108688962 on Build GRCh38
rs763464362 Chr.11: 108689021-108689021 on Build GRCh38
rs752839355 Chr.11: 108689077-108689079 on Build GRCh38
rs775049164 Chr.11: 108688988-108688988 on Build GRCh38
rs957902525 Chr.11: 108688978-108688978 on Build GRCh38
rs774805075 Chr.11: 108689096-108689096 on Build GRCh38
rs71489912 Chr.11: 108688826-108688826 on Build GRCh38
rs755247734 Chr.11: 108689084-108689084 on Build GRCh38
rs768655680 Chr.11: 108688958-108688958 on Build GRCh38
rs749447289 Chr.11: 108688661-108688661 on Build GRCh38
rs923649980 Chr.11: 108688694-108688694 on Build GRCh38
rs747107531 Chr.11: 108688955-108688955 on Build GRCh38
rs553516531 Chr.11: 108689090-108689090 on Build GRCh38
rs911577323 Chr.11: 108689110-108689110 on Build GRCh38
rs763814164 Chr.11: 108689106-108689106 on Build GRCh38
rs757194306 Chr.11: 108689022-108689022 on Build GRCh38
rs758334785 Chr.11: 108689046-108689046 on Build GRCh38
rs34507955 Chr.11: 108689184-108689185 on Build GRCh38
rs200433596 Chr.11: 108688923-108688923 on Build GRCh38
rs763317883 Chr.11: 108688979-108688979 on Build GRCh38
rs545739696 Chr.11: 108689186-108689186 on Build GRCh38
rs773546509 Chr.11: 108688983-108688983 on Build GRCh38
rs767432517 Chr.11: 108689073-108689073 on Build GRCh38
rs755651414 Chr.11: 108688907-108688907 on Build GRCh38
rs752242526 Chr.11: 108688903-108688903 on Build GRCh38
rs759145134 Chr.11: 108688895-108688895 on Build GRCh38
rs143056342 Chr.11: 108689072-108689073 on Build GRCh38
rs570261370 Chr.11: 108688790-108688790 on Build GRCh38
rs771251442 Chr.11: 108689094-108689094 on Build GRCh38
rs772362480 Chr.11: 108688940-108688940 on Build GRCh38
rs74689490 Chr.11: 108688937-108688937 on Build GRCh38
rs970941833 Chr.11: 108688660-108688660 on Build GRCh38
rs548816030 Chr.11: 108688744-108688744 on Build GRCh38
rs138543405 Chr.11: 108688964-108688964 on Build GRCh38
rs1012176409 Chr.11: 108689051-108689051 on Build GRCh38
rs747242977 Chr.11: 108689083-108689083 on Build GRCh38
rs185799121 Chr.11: 108688900-108688900 on Build GRCh38
rs143049163 Chr.11: 108689034-108689034 on Build GRCh38

View Full Product Details