Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 106097048-106097599 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AASDHPPT
Entrez Gene ID
60496
Gene Name
aminoadipate-semialdehyde dehydrogenase-phosphopantetheinyl transferase
Gene Aliases
AASD-PPT, ACPS, CGI-80, LYS2, LYS5
UniGene
Hs.524009
Gene Chromosome Location
Chr.11: 106077565-106098692 on Build GRCh38
Transcripts
NM_015423.2

SNP Detail

SNP ID Chromosome Location
rs551363423 Chr.11: 106097258-106097258 on Build GRCh38
rs147946690 Chr.11: 106097190-106097190 on Build GRCh38
rs960579794 Chr.11: 106097289-106097290 on Build GRCh38
rs866061106 Chr.11: 106097135-106097135 on Build GRCh38
rs923381072 Chr.11: 106097173-106097173 on Build GRCh38
rs554205961 Chr.11: 106097323-106097323 on Build GRCh38
rs909209454 Chr.11: 106097464-106097464 on Build GRCh38
rs567435411 Chr.11: 106097162-106097162 on Build GRCh38
rs773121274 Chr.11: 106097150-106097151 on Build GRCh38
rs1053374571 Chr.11: 106097395-106097395 on Build GRCh38
rs957938545 Chr.11: 106097086-106097086 on Build GRCh38
rs1046548540 Chr.11: 106097275-106097275 on Build GRCh38
rs185327669 Chr.11: 106097149-106097149 on Build GRCh38
rs189738400 Chr.11: 106097221-106097221 on Build GRCh38
rs976629826 Chr.11: 106097420-106097420 on Build GRCh38
rs963405872 Chr.11: 106097468-106097468 on Build GRCh38
rs145522053 Chr.11: 106097428-106097428 on Build GRCh38
rs73537649 Chr.11: 106097560-106097560 on Build GRCh38
rs909240748 Chr.11: 106097212-106097212 on Build GRCh38
rs1046546156 Chr.11: 106097572-106097572 on Build GRCh38
rs900916904 Chr.11: 106097105-106097105 on Build GRCh38
rs999017419 Chr.11: 106097078-106097078 on Build GRCh38
rs916015693 Chr.11: 106097317-106097317 on Build GRCh38
rs181132335 Chr.11: 106097134-106097134 on Build GRCh38
rs1005208729 Chr.11: 106097552-106097552 on Build GRCh38
rs147804891 Chr.11: 106097448-106097448 on Build GRCh38
rs141764480 Chr.11: 106097379-106097379 on Build GRCh38
rs775187307 Chr.11: 106097318-106097318 on Build GRCh38
rs545706895 Chr.11: 106097384-106097385 on Build GRCh38
rs1031951797 Chr.11: 106097310-106097310 on Build GRCh38
rs539310369 Chr.11: 106097303-106097303 on Build GRCh38
rs148845850 Chr.11: 106097567-106097567 on Build GRCh38
rs901301559 Chr.11: 106097412-106097412 on Build GRCh38
rs1036654591 Chr.11: 106097222-106097222 on Build GRCh38
rs889563504 Chr.11: 106097528-106097529 on Build GRCh38
rs986238064 Chr.11: 106097459-106097459 on Build GRCh38
rs931950553 Chr.11: 106097189-106097189 on Build GRCh38
rs978763446 Chr.11: 106097346-106097346 on Build GRCh38
rs1020836417 Chr.11: 106097355-106097355 on Build GRCh38
rs113901481 Chr.11: 106097103-106097103 on Build GRCh38
rs972271643 Chr.11: 106097548-106097548 on Build GRCh38
rs11226919 Chr.11: 106097484-106097484 on Build GRCh38
rs183456565 Chr.11: 106097277-106097277 on Build GRCh38
rs1009433022 Chr.11: 106097335-106097335 on Build GRCh38
rs533679053 Chr.11: 106097233-106097233 on Build GRCh38
rs796295245 Chr.11: 106097496-106097496 on Build GRCh38
rs868621769 Chr.11: 106097343-106097343 on Build GRCh38
rs1016842745 Chr.11: 106097201-106097201 on Build GRCh38
rs762473708 Chr.11: 106097396-106097396 on Build GRCh38
rs576712227 Chr.11: 106097347-106097347 on Build GRCh38
rs953472807 Chr.11: 106097458-106097458 on Build GRCh38
rs72977072 Chr.11: 106097074-106097074 on Build GRCh38

View Full Product Details