Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 4368351-4368894 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGF23
Entrez Gene ID
8074
Gene Name
fibroblast growth factor 23
Gene Aliases
ADHR, FGFN, HPDR2, HYPF, PHPTC
UniGene
Hs.287370
Gene Chromosome Location
Chr.12: 4368227-4379728 on Build GRCh38
Transcripts
NM_020638.2

SNP Detail

SNP ID Chromosome Location
rs1006829668 Chr.12: 4368673-4368673 on Build GRCh38
rs954989010 Chr.12: 4368553-4368553 on Build GRCh38
rs767822391 Chr.12: 4368867-4368867 on Build GRCh38
rs879859887 Chr.12: 4368537-4368537 on Build GRCh38
rs1056424983 Chr.12: 4368394-4368394 on Build GRCh38
rs886049397 Chr.12: 4368418-4368418 on Build GRCh38
rs1038470722 Chr.12: 4368853-4368853 on Build GRCh38
rs945905010 Chr.12: 4368704-4368704 on Build GRCh38
rs942543525 Chr.12: 4368410-4368410 on Build GRCh38
rs13312801 Chr.12: 4368540-4368540 on Build GRCh38
rs1030339194 Chr.12: 4368841-4368841 on Build GRCh38
rs914367894 Chr.12: 4368601-4368601 on Build GRCh38
rs762018989 Chr.12: 4368795-4368795 on Build GRCh38
rs13312799 Chr.12: 4368808-4368808 on Build GRCh38
rs896710141 Chr.12: 4368499-4368499 on Build GRCh38
rs986708682 Chr.12: 4368851-4368851 on Build GRCh38
rs562433374 Chr.12: 4368768-4368768 on Build GRCh38
rs183802802 Chr.12: 4368457-4368457 on Build GRCh38
rs911054802 Chr.12: 4368388-4368388 on Build GRCh38
rs571095026 Chr.12: 4368732-4368732 on Build GRCh38
rs954732003 Chr.12: 4368847-4368847 on Build GRCh38
rs536751063 Chr.12: 4368403-4368403 on Build GRCh38
rs903622041 Chr.12: 4368455-4368455 on Build GRCh38
rs13312800 Chr.12: 4368571-4368571 on Build GRCh38
rs989930690 Chr.12: 4368594-4368594 on Build GRCh38
rs1052542528 Chr.12: 4368496-4368496 on Build GRCh38
rs939363096 Chr.12: 4368380-4368380 on Build GRCh38
rs780145979 Chr.12: 4368579-4368579 on Build GRCh38
rs963778003 Chr.12: 4368585-4368585 on Build GRCh38
rs1013795298 Chr.12: 4368530-4368530 on Build GRCh38
rs911169995 Chr.12: 4368854-4368854 on Build GRCh38
rs1056372447 Chr.12: 4368809-4368809 on Build GRCh38
rs191299667 Chr.12: 4368433-4368433 on Build GRCh38
rs1025895715 Chr.12: 4368539-4368539 on Build GRCh38

View Full Product Details