Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 118750208-118750732 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX6
Entrez Gene ID
1656
Gene Name
DEAD-box helicase 6
Gene Aliases
HLR2, P54, RCK
UniGene
Hs.408461
Gene Chromosome Location
Chr.11: 118747763-118791263 on Build GRCh38
Transcripts
NM_001257191.2 NM_004397.5 XM_005271417.3 XM_011542644.1 XM_011542645.1 XM_017017251.1

SNP Detail

SNP ID Chromosome Location
rs782571068 Chr.11: 118750551-118750551 on Build GRCh38
rs138558622 Chr.11: 118750457-118750457 on Build GRCh38
rs543774123 Chr.11: 118750263-118750264 on Build GRCh38
rs573771337 Chr.11: 118750368-118750368 on Build GRCh38
rs560278277 Chr.11: 118750517-118750517 on Build GRCh38
rs527413509 Chr.11: 118750545-118750545 on Build GRCh38
rs117409833 Chr.11: 118750567-118750567 on Build GRCh38
rs782473983 Chr.11: 118750598-118750598 on Build GRCh38
rs141755095 Chr.11: 118750700-118750700 on Build GRCh38
rs191280295 Chr.11: 118750372-118750372 on Build GRCh38
rs375108343 Chr.11: 118750675-118750675 on Build GRCh38
rs34760273 Chr.11: 118750576-118750577 on Build GRCh38
rs533406033 Chr.11: 118750437-118750437 on Build GRCh38
rs112705333 Chr.11: 118750523-118750523 on Build GRCh38
rs529466501 Chr.11: 118750376-118750376 on Build GRCh38
rs547217508 Chr.11: 118750629-118750629 on Build GRCh38
rs539285205 Chr.11: 118750692-118750692 on Build GRCh38
rs544888273 Chr.11: 118750660-118750660 on Build GRCh38
rs552178782 Chr.11: 118750547-118750547 on Build GRCh38
rs61901394 Chr.11: 118750276-118750276 on Build GRCh38
rs181664327 Chr.11: 118750465-118750465 on Build GRCh38
rs562267182 Chr.11: 118750299-118750299 on Build GRCh38
rs200171136 Chr.11: 118750516-118750516 on Build GRCh38
rs570487334 Chr.11: 118750561-118750562 on Build GRCh38
rs540468560 Chr.11: 118750239-118750239 on Build GRCh38
rs561765596 Chr.11: 118750264-118750264 on Build GRCh38
rs563860697 Chr.11: 118750492-118750492 on Build GRCh38
rs782818652 Chr.11: 118750462-118750462 on Build GRCh38
rs535503328 Chr.11: 118750613-118750613 on Build GRCh38
rs782756667 Chr.11: 118750313-118750313 on Build GRCh38
rs543448375 Chr.11: 118750235-118750235 on Build GRCh38

View Full Product Details