Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 57606226-57606747 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DTX3
Entrez Gene ID
196403
Gene Name
deltex E3 ubiquitin ligase 3
Gene Aliases
RNF154, deltex3
UniGene
Hs.32374
Gene Chromosome Location
Chr.12: 57604327-57609804 on Build GRCh38
Transcripts
NM_001286245.1 NM_001286246.1 NM_178502.3 XM_005268697.1 XM_005268698.1 XM_005268700.1 XM_005268703.1 XM_011538022.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7422041 c.12A>T p.L4L Chr.12: 57606501-57606501 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs764296814 Chr.12: 57606681-57606681 on Build GRCh38
rs778797097 Chr.12: 57606503-57606503 on Build GRCh38
rs894096150 Chr.12: 57606379-57606379 on Build GRCh38
rs755289 Chr.12: 57606433-57606433 on Build GRCh38
rs780015718 Chr.12: 57606548-57606548 on Build GRCh38
rs549134767 Chr.12: 57606470-57606470 on Build GRCh38
rs755288 Chr.12: 57606413-57606413 on Build GRCh38
rs1039247963 Chr.12: 57606372-57606372 on Build GRCh38
rs774202802 Chr.12: 57606347-57606347 on Build GRCh38
rs578177641 Chr.12: 57606323-57606323 on Build GRCh38
rs895226455 Chr.12: 57606705-57606705 on Build GRCh38
rs910975965 Chr.12: 57606511-57606511 on Build GRCh38
rs377124900 Chr.12: 57606461-57606461 on Build GRCh38
rs549339784 Chr.12: 57606394-57606394 on Build GRCh38
rs1008165223 Chr.12: 57606369-57606369 on Build GRCh38
rs887364573 Chr.12: 57606460-57606460 on Build GRCh38
rs959220304 Chr.12: 57606429-57606429 on Build GRCh38
rs703834 Chr.12: 57606595-57606595 on Build GRCh38
rs151324459 Chr.12: 57606264-57606264 on Build GRCh38
rs1035079935 Chr.12: 57606688-57606688 on Build GRCh38
rs1035244482 Chr.12: 57606425-57606426 on Build GRCh38
rs781325167 Chr.12: 57606567-57606567 on Build GRCh38
rs959524992 Chr.12: 57606273-57606273 on Build GRCh38
rs545470284 Chr.12: 57606342-57606342 on Build GRCh38
rs758357425 Chr.12: 57606545-57606545 on Build GRCh38
rs918298887 Chr.12: 57606338-57606338 on Build GRCh38
rs967164492 Chr.12: 57606399-57606399 on Build GRCh38
rs767046678 Chr.12: 57606475-57606475 on Build GRCh38
rs768775507 Chr.12: 57606552-57606552 on Build GRCh38
rs377283663 Chr.12: 57606481-57606481 on Build GRCh38
rs1052334252 Chr.12: 57606343-57606343 on Build GRCh38
rs1003970140 Chr.12: 57606687-57606687 on Build GRCh38
rs79329099 Chr.12: 57606642-57606642 on Build GRCh38
rs894058351 Chr.12: 57606627-57606627 on Build GRCh38
rs1033751888 Chr.12: 57606272-57606272 on Build GRCh38
rs773722332 Chr.12: 57606469-57606469 on Build GRCh38
rs1042355366 Chr.12: 57606643-57606643 on Build GRCh38
rs762304748 Chr.12: 57606529-57606531 on Build GRCh38
rs186859788 Chr.12: 57606431-57606431 on Build GRCh38
rs779957983 Chr.12: 57606262-57606262 on Build GRCh38
rs770128654 Chr.12: 57606467-57606467 on Build GRCh38
rs77762493 Chr.12: 57606361-57606361 on Build GRCh38
rs554539439 Chr.12: 57606263-57606263 on Build GRCh38
rs747095815 Chr.12: 57606549-57606549 on Build GRCh38
rs750413812 Chr.12: 57606529-57606529 on Build GRCh38
rs1013426483 Chr.12: 57606605-57606605 on Build GRCh38
rs182169563 Chr.12: 57606380-57606380 on Build GRCh38
rs1002192094 Chr.12: 57606510-57606510 on Build GRCh38
rs768806767 Chr.12: 57606449-57606449 on Build GRCh38
rs757180119 Chr.12: 57606489-57606489 on Build GRCh38
rs777091755 Chr.12: 57606445-57606446 on Build GRCh38
rs772944627 Chr.12: 57606659-57606659 on Build GRCh38
rs117444441 Chr.12: 57606432-57606432 on Build GRCh38
rs1036736122 Chr.12: 57606331-57606331 on Build GRCh38
rs753526433 Chr.12: 57606488-57606488 on Build GRCh38
rs192781578 Chr.12: 57606485-57606485 on Build GRCh38
rs576444586 Chr.12: 57606664-57606664 on Build GRCh38
rs896537627 Chr.12: 57606601-57606601 on Build GRCh38
rs752377079 Chr.12: 57606697-57606697 on Build GRCh38
rs765693743 Chr.12: 57606565-57606565 on Build GRCh38
rs960569186 Chr.12: 57606274-57606274 on Build GRCh38
rs75051978 Chr.12: 57606289-57606289 on Build GRCh38
rs73346089 Chr.12: 57606450-57606450 on Build GRCh38
rs940995462 Chr.12: 57606296-57606296 on Build GRCh38
rs756792875 Chr.12: 57606462-57606462 on Build GRCh38
rs966989387 Chr.12: 57606340-57606340 on Build GRCh38
rs537338507 Chr.12: 57606686-57606686 on Build GRCh38
rs543676355 Chr.12: 57606271-57606271 on Build GRCh38
rs1020890493 Chr.12: 57606321-57606321 on Build GRCh38
rs962621473 Chr.12: 57606436-57606436 on Build GRCh38

View Full Product Details