Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 108516775-108517352 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FICD
Entrez Gene ID
11153
Gene Name
FIC domain containing
Gene Aliases
HIP13, HYPE, UNQ3041
UniGene
Hs.661891
Gene Chromosome Location
Chr.12: 108515274-108519603 on Build GRCh38
Transcripts
NM_007076.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
334997 c.115G>T p.A39S Chr.12: 108517087-108517087 on Build GRCh38
7224922 c.213C>T p.A71A Chr.12: 108517185-108517185 on Build GRCh38
311195 c.264C>T p.G88G Chr.12: 108517236-108517236 on Build GRCh38
6664019 c.247A>G p.S83G Chr.12: 108517219-108517219 on Build GRCh38
934564 c.130C>T p.L44L Chr.12: 108517102-108517102 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374636127 Chr.12: 108517206-108517206 on Build GRCh38
rs760352330 Chr.12: 108517122-108517123 on Build GRCh38
rs753928259 Chr.12: 108517199-108517199 on Build GRCh38
rs201987704 Chr.12: 108517238-108517238 on Build GRCh38
rs771785980 Chr.12: 108517166-108517166 on Build GRCh38
rs373520821 Chr.12: 108516987-108516987 on Build GRCh38
rs750303741 Chr.12: 108517303-108517304 on Build GRCh38
rs756246676 Chr.12: 108517125-108517125 on Build GRCh38
rs913831960 Chr.12: 108516981-108516981 on Build GRCh38
rs375796986 Chr.12: 108516991-108516991 on Build GRCh38
rs149123644 Chr.12: 108516933-108516933 on Build GRCh38
rs550773541 Chr.12: 108516948-108516948 on Build GRCh38
rs370547934 Chr.12: 108517154-108517154 on Build GRCh38
rs767737843 Chr.12: 108517115-108517115 on Build GRCh38
rs140839889 Chr.12: 108516982-108516982 on Build GRCh38
rs770452327 Chr.12: 108516986-108516986 on Build GRCh38
rs148872410 Chr.12: 108517069-108517069 on Build GRCh38
rs555828094 Chr.12: 108517326-108517326 on Build GRCh38
rs754343901 Chr.12: 108517137-108517137 on Build GRCh38
rs778377849 Chr.12: 108517039-108517039 on Build GRCh38
rs746608375 Chr.12: 108516847-108516847 on Build GRCh38
rs372991038 Chr.12: 108517175-108517175 on Build GRCh38
rs143692085 Chr.12: 108517236-108517236 on Build GRCh38
rs762018453 Chr.12: 108517282-108517282 on Build GRCh38
rs770734583 Chr.12: 108517268-108517268 on Build GRCh38
rs779410622 Chr.12: 108517156-108517156 on Build GRCh38
rs938663797 Chr.12: 108516968-108516968 on Build GRCh38
rs2303634 Chr.12: 108517185-108517185 on Build GRCh38
rs201767952 Chr.12: 108517101-108517101 on Build GRCh38
rs746251994 Chr.12: 108517165-108517165 on Build GRCh38
rs150112694 Chr.12: 108517014-108517014 on Build GRCh38
rs749607127 Chr.12: 108516991-108516992 on Build GRCh38
rs757784877 Chr.12: 108517205-108517205 on Build GRCh38
rs200769403 Chr.12: 108517015-108517015 on Build GRCh38
rs1038762429 Chr.12: 108517037-108517037 on Build GRCh38
rs775258174 Chr.12: 108517113-108517115 on Build GRCh38
rs758857687 Chr.12: 108517235-108517235 on Build GRCh38
rs143126238 Chr.12: 108517060-108517060 on Build GRCh38
rs759014098 Chr.12: 108517320-108517320 on Build GRCh38
rs199500206 Chr.12: 108516923-108516923 on Build GRCh38
rs776045064 Chr.12: 108517176-108517176 on Build GRCh38
rs769650858 Chr.12: 108517257-108517257 on Build GRCh38
rs566200700 Chr.12: 108517168-108517168 on Build GRCh38
rs369729485 Chr.12: 108517100-108517100 on Build GRCh38
rs778089175 Chr.12: 108517131-108517131 on Build GRCh38
rs970197718 Chr.12: 108516890-108516890 on Build GRCh38
rs995219692 Chr.12: 108516879-108516879 on Build GRCh38
rs763761762 Chr.12: 108516922-108516922 on Build GRCh38
rs980140473 Chr.12: 108517173-108517173 on Build GRCh38
rs771440208 Chr.12: 108517051-108517051 on Build GRCh38
rs745482581 Chr.12: 108517062-108517062 on Build GRCh38
rs939045447 Chr.12: 108517249-108517249 on Build GRCh38
rs375782325 Chr.12: 108517181-108517181 on Build GRCh38
rs138159681 Chr.12: 108516927-108516927 on Build GRCh38
rs746642946 Chr.12: 108517237-108517237 on Build GRCh38
rs781219255 Chr.12: 108517022-108517022 on Build GRCh38
rs200561055 Chr.12: 108516988-108516988 on Build GRCh38
rs748310807 Chr.12: 108517029-108517029 on Build GRCh38
rs773161988 Chr.12: 108517262-108517262 on Build GRCh38
rs760504118 Chr.12: 108517004-108517004 on Build GRCh38
rs755064158 Chr.12: 108516941-108516941 on Build GRCh38
rs1044922453 Chr.12: 108517312-108517312 on Build GRCh38
rs781308114 Chr.12: 108516947-108516947 on Build GRCh38
rs765792790 Chr.12: 108517289-108517289 on Build GRCh38
rs757909802 Chr.12: 108517141-108517141 on Build GRCh38
rs751766622 Chr.12: 108517016-108517016 on Build GRCh38
rs963399249 Chr.12: 108516878-108516878 on Build GRCh38
rs1042012457 Chr.12: 108516992-108516992 on Build GRCh38
rs377109715 Chr.12: 108517119-108517119 on Build GRCh38
rs774918691 Chr.12: 108517055-108517055 on Build GRCh38
rs527595045 Chr.12: 108516797-108516797 on Build GRCh38
rs368645109 Chr.12: 108517231-108517231 on Build GRCh38
rs201497382 Chr.12: 108517290-108517290 on Build GRCh38
rs76973043 Chr.12: 108516979-108516979 on Build GRCh38
rs1026338920 Chr.12: 108516894-108516894 on Build GRCh38
rs758100323 Chr.12: 108517317-108517318 on Build GRCh38
rs747023454 Chr.12: 108516934-108516934 on Build GRCh38
rs763870221 Chr.12: 108517080-108517080 on Build GRCh38
rs765076922 Chr.12: 108517299-108517299 on Build GRCh38
rs868228913 Chr.12: 108517136-108517136 on Build GRCh38
rs777307945 Chr.12: 108516838-108516838 on Build GRCh38
rs760261656 Chr.12: 108517078-108517078 on Build GRCh38
rs776217731 Chr.12: 108517172-108517172 on Build GRCh38
rs567586731 Chr.12: 108517167-108517167 on Build GRCh38
rs750284577 Chr.12: 108517104-108517104 on Build GRCh38
rs775872757 Chr.12: 108516953-108516953 on Build GRCh38
rs771793491 Chr.12: 108517082-108517082 on Build GRCh38
rs749551801 Chr.12: 108517040-108517040 on Build GRCh38
rs763730610 Chr.12: 108517162-108517162 on Build GRCh38
rs755260542 Chr.12: 108517017-108517017 on Build GRCh38
rs77006290 Chr.12: 108517227-108517227 on Build GRCh38
rs759739992 Chr.12: 108517107-108517107 on Build GRCh38
rs766675819 Chr.12: 108517325-108517326 on Build GRCh38
rs1026283174 Chr.12: 108517087-108517087 on Build GRCh38
rs143532175 Chr.12: 108517184-108517184 on Build GRCh38
rs764239107 Chr.12: 108517280-108517280 on Build GRCh38
rs557155225 Chr.12: 108516895-108516895 on Build GRCh38
rs375982873 Chr.12: 108517065-108517065 on Build GRCh38
rs762684803 Chr.12: 108517264-108517264 on Build GRCh38
rs774050555 Chr.12: 108517274-108517274 on Build GRCh38
rs779570764 Chr.12: 108517212-108517212 on Build GRCh38
rs767592137 Chr.12: 108517188-108517188 on Build GRCh38
rs928657845 Chr.12: 108516959-108516959 on Build GRCh38
rs34567952 Chr.12: 108517005-108517005 on Build GRCh38
rs762739948 Chr.12: 108517186-108517186 on Build GRCh38
rs775176391 Chr.12: 108516997-108516997 on Build GRCh38
rs915170666 Chr.12: 108516984-108516984 on Build GRCh38
rs146248286 Chr.12: 108517242-108517242 on Build GRCh38
rs371001608 Chr.12: 108517191-108517191 on Build GRCh38
rs780739968 Chr.12: 108517241-108517241 on Build GRCh38
rs764199332 Chr.12: 108517198-108517198 on Build GRCh38
rs1016465213 Chr.12: 108517113-108517113 on Build GRCh38
rs899391192 Chr.12: 108517056-108517056 on Build GRCh38
rs549166647 Chr.12: 108516818-108516818 on Build GRCh38
rs371618378 Chr.12: 108517038-108517038 on Build GRCh38
rs761827476 Chr.12: 108517194-108517194 on Build GRCh38

View Full Product Details