Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 24364058-24364629 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRHL3
Entrez Gene ID
57822
Gene Name
grainyhead like transcription factor 3
Gene Aliases
SOM, TFCP2L4, VWS2
UniGene
Hs.657920
Gene Chromosome Location
Chr.1: 24319322-24364482 on Build GRCh38
Transcripts
NM_198174.2 NM_001199012.1 NM_001199013.1 NM_001199014.1 NM_178122.4 XM_006711025.2 XM_011542403.2 XM_011542404.1 XM_017002768.1 XM_017002769.1 XM_017002770.1

Gene Symbol
STPG1
Entrez Gene ID
90529
Gene Name
sperm tail PG-rich repeat containing 1
Gene Aliases
C1orf201, MAPO2
UniGene
Hs.403187
Gene Chromosome Location
Chr.1: 24356999-24415533 on Build GRCh38
Transcripts
NM_198174.2 NM_001199012.1 NM_001199013.1 NM_001199014.1 NM_178122.4 XM_006711025.2 XM_011542403.2 XM_011542404.1 XM_017002768.1 XM_017002769.1 XM_017002770.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5763064 c.1727G>A p.R576H Chr.1: 24364217-24364217 on Build GRCh38
1340953 c.1731delC p.P579fs*11 Chr.1: 24364221-24364221 on Build GRCh38
3746890 c.1784T>A p.M595K Chr.1: 24364274-24364274 on Build GRCh38
3705706 c.1836C>A p.S612R Chr.1: 24364326-24364326 on Build GRCh38
5630831 c.1853G>C p.S618T Chr.1: 24364343-24364343 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1050156757 Chr.1: 24364179-24364179 on Build GRCh38
rs763952499 Chr.1: 24364207-24364207 on Build GRCh38
rs1040447178 Chr.1: 24364188-24364188 on Build GRCh38
rs961980539 Chr.1: 24364234-24364234 on Build GRCh38
rs542032426 Chr.1: 24364570-24364570 on Build GRCh38
rs538587157 Chr.1: 24364461-24364461 on Build GRCh38
rs200103413 Chr.1: 24364257-24364257 on Build GRCh38
rs74060321 Chr.1: 24364404-24364404 on Build GRCh38
rs903207110 Chr.1: 24364409-24364409 on Build GRCh38
rs1044963024 Chr.1: 24364462-24364462 on Build GRCh38
rs1013067741 Chr.1: 24364445-24364445 on Build GRCh38
rs773122253 Chr.1: 24364402-24364402 on Build GRCh38
rs377523202 Chr.1: 24364571-24364571 on Build GRCh38
rs1003406337 Chr.1: 24364504-24364504 on Build GRCh38
rs6694170 Chr.1: 24364186-24364186 on Build GRCh38
rs920830003 Chr.1: 24364351-24364351 on Build GRCh38
rs910197678 Chr.1: 24364183-24364183 on Build GRCh38
rs920462801 Chr.1: 24364166-24364166 on Build GRCh38
rs558169785 Chr.1: 24364365-24364365 on Build GRCh38
rs773826784 Chr.1: 24364167-24364167 on Build GRCh38
rs1050497646 Chr.1: 24364202-24364202 on Build GRCh38
rs151326764 Chr.1: 24364208-24364208 on Build GRCh38
rs747321006 Chr.1: 24364309-24364309 on Build GRCh38
rs141931022 Chr.1: 24364224-24364224 on Build GRCh38
rs573511242 Chr.1: 24364512-24364512 on Build GRCh38
rs1054623536 Chr.1: 24364209-24364209 on Build GRCh38
rs768212173 Chr.1: 24364150-24364150 on Build GRCh38
rs566408876 Chr.1: 24364418-24364418 on Build GRCh38
rs112469942 Chr.1: 24364275-24364275 on Build GRCh38
rs535901162 Chr.1: 24364225-24364225 on Build GRCh38
rs545809 Chr.1: 24364274-24364274 on Build GRCh38
rs576250609 Chr.1: 24364401-24364401 on Build GRCh38
rs1036593925 Chr.1: 24364345-24364345 on Build GRCh38
rs893404112 Chr.1: 24364442-24364442 on Build GRCh38
rs766651546 Chr.1: 24364177-24364177 on Build GRCh38
rs961700398 Chr.1: 24364544-24364544 on Build GRCh38
rs539431346 Chr.1: 24364364-24364364 on Build GRCh38
rs558326048 Chr.1: 24364474-24364474 on Build GRCh38
rs889257858 Chr.1: 24364387-24364387 on Build GRCh38
rs764307330 Chr.1: 24364405-24364405 on Build GRCh38
rs549102088 Chr.1: 24364151-24364151 on Build GRCh38
rs145783642 Chr.1: 24364356-24364356 on Build GRCh38
rs60585087 Chr.1: 24364131-24364131 on Build GRCh38
rs985891394 Chr.1: 24364609-24364609 on Build GRCh38
rs769150587 Chr.1: 24364347-24364347 on Build GRCh38
rs1023025621 Chr.1: 24364222-24364222 on Build GRCh38
rs951911760 Chr.1: 24364300-24364300 on Build GRCh38
rs1016021379 Chr.1: 24364226-24364226 on Build GRCh38
rs994944673 Chr.1: 24364201-24364201 on Build GRCh38
rs775916587 Chr.1: 24364313-24364313 on Build GRCh38
rs910262356 Chr.1: 24364359-24364359 on Build GRCh38
rs142435457 Chr.1: 24364174-24364174 on Build GRCh38
rs768127778 Chr.1: 24364269-24364284 on Build GRCh38
rs998813039 Chr.1: 24364223-24364223 on Build GRCh38
rs927881333 Chr.1: 24364097-24364097 on Build GRCh38
rs893330497 Chr.1: 24364216-24364216 on Build GRCh38
rs951965536 Chr.1: 24364586-24364586 on Build GRCh38
rs1015593534 Chr.1: 24364530-24364530 on Build GRCh38
rs561498763 Chr.1: 24364436-24364436 on Build GRCh38
rs773073578 Chr.1: 24364217-24364217 on Build GRCh38

View Full Product Details