Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 43980981-43981554 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B4GALT2
Entrez Gene ID
8704
Gene Name
beta-1,4-galactosyltransferase 2
Gene Aliases
B4Gal-T2, B4Gal-T3, beta4Gal-T2
UniGene
Hs.632403
Gene Chromosome Location
Chr.1: 43979202-43991171 on Build GRCh38
Transcripts
NM_001005417.2 NM_003780.4 NM_030587.2 XM_017002716.1 XM_017002717.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4664952 c.131C>T p.A44V Chr.1: 43981204-43981204 on Build GRCh38
6501664 c.218G>A p.R73H Chr.1: 43981291-43981291 on Build GRCh38
4664953 c.44C>T p.A15V Chr.1: 43981204-43981204 on Build GRCh38
6609434 c.96T>A p.F32L Chr.1: 43981256-43981256 on Build GRCh38
4610960 c.175_177delAGC p.S65delS Chr.1: 43981335-43981337 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs569291036 Chr.1: 43981311-43981311 on Build GRCh38
rs536172060 Chr.1: 43981192-43981192 on Build GRCh38
rs111233359 Chr.1: 43981132-43981132 on Build GRCh38
rs376991548 Chr.1: 43981256-43981256 on Build GRCh38
rs765815375 Chr.1: 43981492-43981492 on Build GRCh38
rs928961448 Chr.1: 43981399-43981399 on Build GRCh38
rs766581340 Chr.1: 43981512-43981512 on Build GRCh38
rs757948149 Chr.1: 43981139-43981139 on Build GRCh38
rs746116651 Chr.1: 43981171-43981171 on Build GRCh38
rs912110603 Chr.1: 43981416-43981416 on Build GRCh38
rs757787915 Chr.1: 43981323-43981323 on Build GRCh38
rs376096161 Chr.1: 43981119-43981119 on Build GRCh38
rs757515250 Chr.1: 43981242-43981242 on Build GRCh38
rs755229393 Chr.1: 43981429-43981429 on Build GRCh38
rs1021080409 Chr.1: 43981236-43981236 on Build GRCh38
rs762532434 Chr.1: 43981291-43981291 on Build GRCh38
rs767540187 Chr.1: 43981112-43981112 on Build GRCh38
rs367580012 Chr.1: 43981422-43981422 on Build GRCh38
rs148339623 Chr.1: 43981392-43981392 on Build GRCh38
rs767080506 Chr.1: 43981317-43981317 on Build GRCh38
rs754260512 Chr.1: 43981322-43981322 on Build GRCh38
rs751687755 Chr.1: 43981427-43981427 on Build GRCh38
rs959872350 Chr.1: 43981481-43981481 on Build GRCh38
rs774859229 Chr.1: 43981483-43981483 on Build GRCh38
rs759490732 Chr.1: 43981175-43981175 on Build GRCh38
rs779365326 Chr.1: 43981324-43981324 on Build GRCh38
rs1056470275 Chr.1: 43981199-43981199 on Build GRCh38
rs1030999156 Chr.1: 43981372-43981372 on Build GRCh38
rs368225760 Chr.1: 43981136-43981136 on Build GRCh38
rs763585302 Chr.1: 43981508-43981508 on Build GRCh38
rs902610690 Chr.1: 43981460-43981460 on Build GRCh38
rs756580465 Chr.1: 43981146-43981146 on Build GRCh38
rs138683109 Chr.1: 43981380-43981380 on Build GRCh38
rs1022750383 Chr.1: 43981359-43981359 on Build GRCh38
rs371553773 Chr.1: 43981194-43981194 on Build GRCh38
rs765910641 Chr.1: 43981411-43981411 on Build GRCh38
rs756643777 Chr.1: 43981210-43981210 on Build GRCh38
rs1010989612 Chr.1: 43981221-43981221 on Build GRCh38
rs766089438 Chr.1: 43981302-43981302 on Build GRCh38
rs551490191 Chr.1: 43981334-43981335 on Build GRCh38
rs980986533 Chr.1: 43981395-43981395 on Build GRCh38
rs776480818 Chr.1: 43981276-43981276 on Build GRCh38
rs41303407 Chr.1: 43981265-43981265 on Build GRCh38
rs747964707 Chr.1: 43981367-43981367 on Build GRCh38
rs748315206 Chr.1: 43981431-43981431 on Build GRCh38
rs882961 Chr.1: 43981412-43981412 on Build GRCh38
rs188669335 Chr.1: 43981530-43981530 on Build GRCh38
rs1045280898 Chr.1: 43981024-43981024 on Build GRCh38
rs150890298 Chr.1: 43981438-43981438 on Build GRCh38
rs571755531 Chr.1: 43981107-43981107 on Build GRCh38
rs1018673467 Chr.1: 43981333-43981333 on Build GRCh38
rs201975391 Chr.1: 43981432-43981432 on Build GRCh38
rs375270042 Chr.1: 43981441-43981441 on Build GRCh38
rs200716449 Chr.1: 43981373-43981373 on Build GRCh38
rs756246445 Chr.1: 43981268-43981268 on Build GRCh38
rs761552271 Chr.1: 43981181-43981181 on Build GRCh38
rs749123997 Chr.1: 43981374-43981374 on Build GRCh38
rs968474262 Chr.1: 43981157-43981157 on Build GRCh38
rs2353008 Chr.1: 43981384-43981384 on Build GRCh38
rs943486521 Chr.1: 43981189-43981189 on Build GRCh38
rs752759378 Chr.1: 43981335-43981337 on Build GRCh38
rs774767449 Chr.1: 43981152-43981152 on Build GRCh38
rs771816100 Chr.1: 43981482-43981482 on Build GRCh38
rs1047306273 Chr.1: 43981435-43981435 on Build GRCh38
rs764277669 Chr.1: 43981335-43981343 on Build GRCh38
rs769733983 Chr.1: 43981371-43981371 on Build GRCh38
rs781072642 Chr.1: 43981430-43981430 on Build GRCh38
rs767545341 Chr.1: 43981335-43981340 on Build GRCh38
rs562721930 Chr.1: 43981065-43981065 on Build GRCh38
rs779452219 Chr.1: 43981246-43981246 on Build GRCh38
rs774201196 Chr.1: 43981459-43981459 on Build GRCh38
rs994484819 Chr.1: 43981066-43981066 on Build GRCh38
rs758687714 Chr.1: 43981351-43981351 on Build GRCh38
rs139961353 Chr.1: 43981260-43981260 on Build GRCh38
rs762724852 Chr.1: 43981091-43981091 on Build GRCh38
rs775520119 Chr.1: 43981118-43981118 on Build GRCh38
rs1036041258 Chr.1: 43981020-43981020 on Build GRCh38
rs199512234 Chr.1: 43981365-43981365 on Build GRCh38
rs746022331 Chr.1: 43981252-43981252 on Build GRCh38
rs769533543 Chr.1: 43981290-43981290 on Build GRCh38
rs567382513 Chr.1: 43981019-43981019 on Build GRCh38
rs748611876 Chr.1: 43981141-43981141 on Build GRCh38
rs770551686 Chr.1: 43981377-43981377 on Build GRCh38
rs375735099 Chr.1: 43981241-43981241 on Build GRCh38
rs991399486 Chr.1: 43981484-43981484 on Build GRCh38
rs780266757 Chr.1: 43981262-43981262 on Build GRCh38
rs762398729 Chr.1: 43981403-43981403 on Build GRCh38
rs749684547 Chr.1: 43981151-43981151 on Build GRCh38
rs141394710 Chr.1: 43981126-43981126 on Build GRCh38
rs760299217 Chr.1: 43981486-43981486 on Build GRCh38
rs894020408 Chr.1: 43981204-43981204 on Build GRCh38
rs371421881 Chr.1: 43981391-43981391 on Build GRCh38
rs529773880 Chr.1: 43981158-43981158 on Build GRCh38
rs547278358 Chr.1: 43981176-43981176 on Build GRCh38
rs766166962 Chr.1: 43981102-43981102 on Build GRCh38
rs567524191 Chr.1: 43981183-43981183 on Build GRCh38
rs765806648 Chr.1: 43981193-43981193 on Build GRCh38
rs750026175 Chr.1: 43981184-43981184 on Build GRCh38
rs751751243 Chr.1: 43981052-43981052 on Build GRCh38
rs940311738 Chr.1: 43981463-43981463 on Build GRCh38
rs375329179 Chr.1: 43981140-43981140 on Build GRCh38
rs759264772 Chr.1: 43981390-43981390 on Build GRCh38
rs773804993 Chr.1: 43981498-43981498 on Build GRCh38
rs776479895 Chr.1: 43981180-43981180 on Build GRCh38
rs759733436 Chr.1: 43981105-43981105 on Build GRCh38
rs368130720 Chr.1: 43981446-43981446 on Build GRCh38
rs987553231 Chr.1: 43981414-43981414 on Build GRCh38
rs780462896 Chr.1: 43981363-43981363 on Build GRCh38
rs745454373 Chr.1: 43981480-43981480 on Build GRCh38
rs761829356 Chr.1: 43981281-43981281 on Build GRCh38
rs149892509 Chr.1: 43981335-43981335 on Build GRCh38
rs374651243 Chr.1: 43981421-43981421 on Build GRCh38
rs960237219 Chr.1: 43981266-43981266 on Build GRCh38
rs779430078 Chr.1: 43981510-43981511 on Build GRCh38
rs867144565 Chr.1: 43981125-43981125 on Build GRCh38
rs527442282 Chr.1: 43981175-43981175 on Build GRCh38
rs145753168 Chr.1: 43981366-43981366 on Build GRCh38
rs753587540 Chr.1: 43981127-43981127 on Build GRCh38
rs774216783 Chr.1: 43981174-43981175 on Build GRCh38

View Full Product Details