Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 115700197-115700792 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASQ2
Entrez Gene ID
845
Gene Name
calsequestrin 2
Gene Aliases
PDIB2
UniGene
Hs.57975
Gene Chromosome Location
Chr.1: 115700003-115768805 on Build GRCh38
Transcripts
NM_001232.3

SNP Detail

SNP ID Chromosome Location
rs753399691 Chr.1: 115700391-115700391 on Build GRCh38
rs1140238 Chr.1: 115700609-115700609 on Build GRCh38
rs890900923 Chr.1: 115700713-115700713 on Build GRCh38
rs963171199 Chr.1: 115700444-115700444 on Build GRCh38
rs1000749704 Chr.1: 115700356-115700356 on Build GRCh38
rs997002632 Chr.1: 115700376-115700376 on Build GRCh38
rs991581170 Chr.1: 115700471-115700471 on Build GRCh38
rs917427417 Chr.1: 115700258-115700258 on Build GRCh38
rs567171722 Chr.1: 115700347-115700348 on Build GRCh38
rs773802801 Chr.1: 115700506-115700507 on Build GRCh38
rs977468033 Chr.1: 115700616-115700616 on Build GRCh38
rs1055862952 Chr.1: 115700248-115700248 on Build GRCh38
rs950164742 Chr.1: 115700266-115700266 on Build GRCh38
rs568557039 Chr.1: 115700485-115700485 on Build GRCh38
rs528761232 Chr.1: 115700323-115700323 on Build GRCh38
rs973218233 Chr.1: 115700481-115700481 on Build GRCh38
rs1039829915 Chr.1: 115700689-115700689 on Build GRCh38
rs796966289 Chr.1: 115700718-115700718 on Build GRCh38
rs558093187 Chr.1: 115700703-115700703 on Build GRCh38
rs973989613 Chr.1: 115700424-115700424 on Build GRCh38
rs936117041 Chr.1: 115700668-115700668 on Build GRCh38
rs1030702246 Chr.1: 115700482-115700482 on Build GRCh38
rs954562708 Chr.1: 115700408-115700408 on Build GRCh38
rs370391826 Chr.1: 115700286-115700286 on Build GRCh38
rs766919745 Chr.1: 115700740-115700740 on Build GRCh38
rs937436184 Chr.1: 115700244-115700244 on Build GRCh38
rs917293815 Chr.1: 115700257-115700257 on Build GRCh38
rs926002665 Chr.1: 115700667-115700667 on Build GRCh38
rs1029332949 Chr.1: 115700386-115700386 on Build GRCh38
rs76074918 Chr.1: 115700236-115700236 on Build GRCh38
rs547458797 Chr.1: 115700312-115700312 on Build GRCh38
rs898305902 Chr.1: 115700764-115700764 on Build GRCh38
rs752752366 Chr.1: 115700757-115700757 on Build GRCh38
rs992778014 Chr.1: 115700599-115700599 on Build GRCh38
rs1050714474 Chr.1: 115700679-115700679 on Build GRCh38
rs564494904 Chr.1: 115700274-115700274 on Build GRCh38
rs11347859 Chr.1: 115700224-115700224 on Build GRCh38
rs773542486 Chr.1: 115700526-115700526 on Build GRCh38
rs184571412 Chr.1: 115700596-115700596 on Build GRCh38
rs149941897 Chr.1: 115700345-115700345 on Build GRCh38
rs1054540005 Chr.1: 115700346-115700346 on Build GRCh38
rs936267523 Chr.1: 115700593-115700593 on Build GRCh38
rs1029954002 Chr.1: 115700403-115700403 on Build GRCh38
rs950093877 Chr.1: 115700517-115700517 on Build GRCh38
rs946143966 Chr.1: 115700264-115700264 on Build GRCh38
rs887371827 Chr.1: 115700747-115700747 on Build GRCh38
rs766012096 Chr.1: 115700586-115700586 on Build GRCh38
rs7521023 Chr.1: 115700759-115700759 on Build GRCh38
rs536769594 Chr.1: 115700669-115700669 on Build GRCh38
rs1039862304 Chr.1: 115700384-115700384 on Build GRCh38
rs1004562136 Chr.1: 115700420-115700420 on Build GRCh38
rs900052305 Chr.1: 115700708-115700708 on Build GRCh38
rs72554068 Chr.1: 115700738-115700738 on Build GRCh38
rs1015062488 Chr.1: 115700433-115700433 on Build GRCh38
rs191366126 Chr.1: 115700542-115700542 on Build GRCh38
rs958695767 Chr.1: 115700437-115700437 on Build GRCh38
rs998419586 Chr.1: 115700401-115700401 on Build GRCh38
rs762615632 Chr.1: 115700556-115700556 on Build GRCh38
rs917180844 Chr.1: 115700605-115700605 on Build GRCh38
rs746341228 Chr.1: 115700338-115700338 on Build GRCh38
rs886045157 Chr.1: 115700223-115700224 on Build GRCh38

View Full Product Details