Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 114599233-114599671 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DENND2C
Entrez Gene ID
163259
Gene Name
DENN domain containing 2C
Gene Aliases
dJ1156J9.1
UniGene
Hs.654928
Gene Chromosome Location
Chr.1: 114584575-114670111 on Build GRCh38
Transcripts
NM_001256404.1 NM_198459.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4842975 c.2151G>A p.P717P Chr.1: 114599406-114599406 on Build GRCh38
6642320 c.2168C>T p.T723I Chr.1: 114599389-114599389 on Build GRCh38
5843127 c.2150C>T p.P717L Chr.1: 114599407-114599407 on Build GRCh38
4872633 c.2201A>G p.D734G Chr.1: 114599356-114599356 on Build GRCh38
4783917 c.2234G>T p.G745V Chr.1: 114599323-114599323 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373876760 Chr.1: 114599286-114599286 on Build GRCh38
rs781305601 Chr.1: 114599410-114599410 on Build GRCh38
rs778125043 Chr.1: 114599404-114599404 on Build GRCh38
rs200297482 Chr.1: 114599308-114599308 on Build GRCh38
rs763890588 Chr.1: 114599260-114599260 on Build GRCh38
rs766945962 Chr.1: 114599278-114599278 on Build GRCh38
rs752749984 Chr.1: 114599535-114599535 on Build GRCh38
rs770986312 Chr.1: 114599316-114599316 on Build GRCh38
rs745817508 Chr.1: 114599326-114599326 on Build GRCh38
rs1037821707 Chr.1: 114599569-114599569 on Build GRCh38
rs868456364 Chr.1: 114599538-114599538 on Build GRCh38
rs373650502 Chr.1: 114599430-114599430 on Build GRCh38
rs548792563 Chr.1: 114599406-114599406 on Build GRCh38
rs751215687 Chr.1: 114599452-114599453 on Build GRCh38
rs201976495 Chr.1: 114599282-114599282 on Build GRCh38
rs533733411 Chr.1: 114599372-114599372 on Build GRCh38
rs537763732 Chr.1: 114599616-114599616 on Build GRCh38
rs767604949 Chr.1: 114599505-114599505 on Build GRCh38
rs190920274 Chr.1: 114599543-114599543 on Build GRCh38
rs760063723 Chr.1: 114599374-114599374 on Build GRCh38
rs765682075 Chr.1: 114599376-114599376 on Build GRCh38
rs762792966 Chr.1: 114599352-114599352 on Build GRCh38
rs774350711 Chr.1: 114599498-114599498 on Build GRCh38
rs751285039 Chr.1: 114599266-114599266 on Build GRCh38
rs201065413 Chr.1: 114599359-114599359 on Build GRCh38
rs753234702 Chr.1: 114599380-114599380 on Build GRCh38
rs750364375 Chr.1: 114599379-114599379 on Build GRCh38
rs758785731 Chr.1: 114599301-114599301 on Build GRCh38
rs749249060 Chr.1: 114599483-114599483 on Build GRCh38
rs755502651 Chr.1: 114599283-114599283 on Build GRCh38
rs758697521 Chr.1: 114599386-114599386 on Build GRCh38
rs532994522 Chr.1: 114599303-114599303 on Build GRCh38
rs745986093 Chr.1: 114599414-114599414 on Build GRCh38
rs775405891 Chr.1: 114599351-114599351 on Build GRCh38
rs144027770 Chr.1: 114599325-114599325 on Build GRCh38
rs753107891 Chr.1: 114599294-114599294 on Build GRCh38
rs150571862 Chr.1: 114599361-114599361 on Build GRCh38
rs377079272 Chr.1: 114599502-114599502 on Build GRCh38
rs768553799 Chr.1: 114599484-114599484 on Build GRCh38
rs762836274 Chr.1: 114599462-114599462 on Build GRCh38
rs140807433 Chr.1: 114599368-114599368 on Build GRCh38
rs761318291 Chr.1: 114599267-114599267 on Build GRCh38
rs187642664 Chr.1: 114599363-114599363 on Build GRCh38
rs932965853 Chr.1: 114599560-114599560 on Build GRCh38
rs757490817 Chr.1: 114599407-114599407 on Build GRCh38
rs868376242 Chr.1: 114599392-114599392 on Build GRCh38
rs769661805 Chr.1: 114599327-114599327 on Build GRCh38
rs781191610 Chr.1: 114599321-114599321 on Build GRCh38

View Full Product Details