Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 151518556-151519154 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CGN
Entrez Gene ID
57530
Gene Name
cingulin
Gene Aliases
-
UniGene
Hs.591464
Gene Chromosome Location
Chr.1: 151510859-151538692 on Build GRCh38
Transcripts
NM_020770.2 XM_005245365.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6573756 c.443A>T p.D148V Chr.1: 151518962-151518962 on Build GRCh38
4140829 c.64T>G p.F22V Chr.1: 151518583-151518583 on Build GRCh38
367597 c.75G>A p.E25E Chr.1: 151518594-151518594 on Build GRCh38
1728877 c.352A>T p.S118C Chr.1: 151518871-151518871 on Build GRCh38
4881393 c.180C>T p.I60I Chr.1: 151518699-151518699 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773533878 Chr.1: 151518583-151518583 on Build GRCh38
rs771744604 Chr.1: 151519017-151519017 on Build GRCh38
rs773873933 Chr.1: 151518881-151518881 on Build GRCh38
rs912576558 Chr.1: 151518737-151518737 on Build GRCh38
rs144710868 Chr.1: 151518612-151518612 on Build GRCh38
rs372537987 Chr.1: 151518976-151518976 on Build GRCh38
rs757459390 Chr.1: 151518834-151518834 on Build GRCh38
rs773190405 Chr.1: 151518863-151518863 on Build GRCh38
rs764767311 Chr.1: 151518821-151518821 on Build GRCh38
rs768532403 Chr.1: 151518753-151518753 on Build GRCh38
rs142876758 Chr.1: 151518699-151518699 on Build GRCh38
rs745441103 Chr.1: 151519013-151519013 on Build GRCh38
rs375611149 Chr.1: 151519115-151519115 on Build GRCh38
rs757866674 Chr.1: 151518969-151518969 on Build GRCh38
rs370327717 Chr.1: 151518864-151518864 on Build GRCh38
rs776514568 Chr.1: 151518580-151518580 on Build GRCh38
rs779168182 Chr.1: 151518867-151518867 on Build GRCh38
rs995729163 Chr.1: 151518944-151518944 on Build GRCh38
rs373995070 Chr.1: 151518919-151518919 on Build GRCh38
rs201247201 Chr.1: 151519041-151519041 on Build GRCh38
rs753209352 Chr.1: 151518718-151518718 on Build GRCh38
rs768461464 Chr.1: 151518667-151518667 on Build GRCh38
rs777153053 Chr.1: 151519065-151519065 on Build GRCh38
rs563727271 Chr.1: 151519100-151519100 on Build GRCh38
rs375705109 Chr.1: 151518796-151518796 on Build GRCh38
rs770390194 Chr.1: 151518878-151518878 on Build GRCh38
rs768688972 Chr.1: 151518742-151518744 on Build GRCh38
rs779332921 Chr.1: 151518742-151518742 on Build GRCh38
rs777068899 Chr.1: 151519123-151519123 on Build GRCh38
rs776650337 Chr.1: 151518892-151518892 on Build GRCh38
rs941207167 Chr.1: 151518618-151518618 on Build GRCh38
rs181435993 Chr.1: 151518935-151518935 on Build GRCh38
rs750481885 Chr.1: 151518980-151518980 on Build GRCh38
rs780983147 Chr.1: 151518848-151518848 on Build GRCh38
rs764673258 Chr.1: 151518965-151518965 on Build GRCh38
rs762349450 Chr.1: 151519082-151519082 on Build GRCh38
rs199670131 Chr.1: 151518861-151518861 on Build GRCh38
rs750126203 Chr.1: 151518623-151518623 on Build GRCh38
rs745364025 Chr.1: 151519112-151519112 on Build GRCh38
rs778777875 Chr.1: 151519106-151519106 on Build GRCh38
rs767380672 Chr.1: 151518589-151518589 on Build GRCh38
rs905632358 Chr.1: 151519028-151519028 on Build GRCh38
rs764771682 Chr.1: 151518620-151518620 on Build GRCh38
rs746607714 Chr.1: 151519117-151519117 on Build GRCh38
rs747240975 Chr.1: 151518674-151518675 on Build GRCh38
rs866489297 Chr.1: 151519125-151519125 on Build GRCh38
rs747961301 Chr.1: 151518998-151518998 on Build GRCh38
rs376940080 Chr.1: 151518637-151518637 on Build GRCh38
rs749481156 Chr.1: 151518988-151518988 on Build GRCh38
rs775452629 Chr.1: 151518595-151518595 on Build GRCh38
rs1031204033 Chr.1: 151518721-151518721 on Build GRCh38
rs369021272 Chr.1: 151518586-151518586 on Build GRCh38
rs924424773 Chr.1: 151518767-151518767 on Build GRCh38
rs779242955 Chr.1: 151518836-151518836 on Build GRCh38
rs747898806 Chr.1: 151518859-151518859 on Build GRCh38
rs373220207 Chr.1: 151518686-151518686 on Build GRCh38
rs770868795 Chr.1: 151519002-151519002 on Build GRCh38
rs140767773 Chr.1: 151519049-151519049 on Build GRCh38
rs746775445 Chr.1: 151518747-151518747 on Build GRCh38
rs756846779 Chr.1: 151518619-151518619 on Build GRCh38
rs576438381 Chr.1: 151518777-151518777 on Build GRCh38
rs765995264 Chr.1: 151518782-151518782 on Build GRCh38
rs768433517 Chr.1: 151519118-151519118 on Build GRCh38
rs556631043 Chr.1: 151518771-151518771 on Build GRCh38
rs141717082 Chr.1: 151518613-151518613 on Build GRCh38
rs777578389 Chr.1: 151519001-151519001 on Build GRCh38
rs747284823 Chr.1: 151518641-151518641 on Build GRCh38
rs766600191 Chr.1: 151519087-151519087 on Build GRCh38
rs559686107 Chr.1: 151519135-151519135 on Build GRCh38
rs907430303 Chr.1: 151518642-151518642 on Build GRCh38
rs772871077 Chr.1: 151519111-151519111 on Build GRCh38
rs1006810957 Chr.1: 151518685-151518685 on Build GRCh38
rs753956199 Chr.1: 151518828-151518828 on Build GRCh38
rs751915500 Chr.1: 151518700-151518700 on Build GRCh38
rs371149064 Chr.1: 151518770-151518770 on Build GRCh38
rs764760450 Chr.1: 151518684-151518684 on Build GRCh38
rs755421633 Chr.1: 151518701-151518701 on Build GRCh38
rs918400696 Chr.1: 151518628-151518628 on Build GRCh38
rs985514918 Chr.1: 151518706-151518706 on Build GRCh38
rs144617400 Chr.1: 151519105-151519105 on Build GRCh38
rs775576011 Chr.1: 151518985-151518985 on Build GRCh38
rs140740467 Chr.1: 151518653-151518653 on Build GRCh38
rs775922111 Chr.1: 151518953-151518953 on Build GRCh38
rs769631086 Chr.1: 151518766-151518766 on Build GRCh38
rs566214963 Chr.1: 151518664-151518664 on Build GRCh38
rs144269614 Chr.1: 151518741-151518741 on Build GRCh38
rs372997590 Chr.1: 151518773-151518773 on Build GRCh38
rs770226846 Chr.1: 151519125-151519133 on Build GRCh38
rs142903144 Chr.1: 151518842-151518842 on Build GRCh38
rs777855550 Chr.1: 151518731-151518731 on Build GRCh38
rs780306512 Chr.1: 151518664-151518666 on Build GRCh38
rs369567501 Chr.1: 151518652-151518652 on Build GRCh38
rs954122778 Chr.1: 151518994-151518994 on Build GRCh38
rs143545092 Chr.1: 151519114-151519114 on Build GRCh38
rs748248097 Chr.1: 151519042-151519043 on Build GRCh38
rs751933887 Chr.1: 151518629-151518629 on Build GRCh38
rs762679619 Chr.1: 151518813-151518813 on Build GRCh38
rs775360877 Chr.1: 151518662-151518662 on Build GRCh38
rs530916904 Chr.1: 151518820-151518820 on Build GRCh38
rs760634522 Chr.1: 151518598-151518598 on Build GRCh38
rs149546521 Chr.1: 151518644-151518644 on Build GRCh38
rs545428620 Chr.1: 151518803-151518803 on Build GRCh38
rs761194235 Chr.1: 151518680-151518680 on Build GRCh38
rs755945240 Chr.1: 151518999-151518999 on Build GRCh38
rs765843724 Chr.1: 151519083-151519083 on Build GRCh38
rs780206061 Chr.1: 151518993-151518993 on Build GRCh38
rs199915302 Chr.1: 151518625-151518625 on Build GRCh38
rs749386763 Chr.1: 151518736-151518736 on Build GRCh38
rs773787926 Chr.1: 151519004-151519004 on Build GRCh38
rs150772007 Chr.1: 151518640-151518640 on Build GRCh38
rs200463676 Chr.1: 151519134-151519134 on Build GRCh38
rs776676596 Chr.1: 151518672-151518672 on Build GRCh38
rs200877437 Chr.1: 151519099-151519099 on Build GRCh38
rs761066980 Chr.1: 151519024-151519024 on Build GRCh38
rs112666836 Chr.1: 151518765-151518765 on Build GRCh38
rs892920338 Chr.1: 151518784-151518784 on Build GRCh38
rs751254718 Chr.1: 151518791-151518791 on Build GRCh38
rs372302522 Chr.1: 151519086-151519086 on Build GRCh38
rs756601182 Chr.1: 151518730-151518730 on Build GRCh38
rs746384237 Chr.1: 151518879-151518879 on Build GRCh38
rs1003052721 Chr.1: 151518918-151518918 on Build GRCh38
rs781521435 Chr.1: 151518715-151518715 on Build GRCh38
rs780346325 Chr.1: 151518626-151518626 on Build GRCh38
rs754432896 Chr.1: 151518967-151518967 on Build GRCh38
rs775057269 Chr.1: 151519022-151519022 on Build GRCh38
rs547118877 Chr.1: 151519051-151519051 on Build GRCh38
rs911278116 Chr.1: 151518923-151518923 on Build GRCh38
rs746059831 Chr.1: 151518838-151518838 on Build GRCh38
rs769896963 Chr.1: 151518581-151518581 on Build GRCh38
rs1016990584 Chr.1: 151518819-151518819 on Build GRCh38
rs758578154 Chr.1: 151518981-151518981 on Build GRCh38
rs367655363 Chr.1: 151519101-151519101 on Build GRCh38
rs141096784 Chr.1: 151518960-151518960 on Build GRCh38
rs368325227 Chr.1: 151518655-151518655 on Build GRCh38

View Full Product Details