Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 233434016-233434565 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DGKD
Entrez Gene ID
8527
Gene Name
diacylglycerol kinase delta
Gene Aliases
DGKdelta, dgkd-2
UniGene
Hs.471675
Gene Chromosome Location
Chr.2: 233354507-233472098 on Build GRCh38
Transcripts
NM_003648.2 NM_152879.2 XM_005246107.3 XM_005246108.1 XM_011512028.2 XM_011512029.2 XM_011512030.2 XM_011512031.2 XM_011512032.2 XM_011512033.2 XM_011512035.1 XM_011512037.2 XM_011512039.2 XM_011512040.2 XM_017005138.1 XM_017005139.1 XM_017005140.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
256753 c.397G>T p.E133* Chr.2: 233434428-233434428 on Build GRCh38
6855424 c.312G>T p.E104D Chr.2: 233434475-233434475 on Build GRCh38
6157595 c.356C>T p.T119I Chr.2: 233434387-233434387 on Build GRCh38
6157596 c.224C>T p.T75I Chr.2: 233434387-233434387 on Build GRCh38
1306554 c.428C>T p.T143I Chr.2: 233434459-233434459 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777537370 Chr.2: 233434518-233434518 on Build GRCh38
rs1054155277 Chr.2: 233434192-233434192 on Build GRCh38
rs956240179 Chr.2: 233434350-233434350 on Build GRCh38
rs780735498 Chr.2: 233434519-233434520 on Build GRCh38
rs555106460 Chr.2: 233434186-233434186 on Build GRCh38
rs539408707 Chr.2: 233434058-233434058 on Build GRCh38
rs543778517 Chr.2: 233434292-233434292 on Build GRCh38
rs917323934 Chr.2: 233434052-233434052 on Build GRCh38
rs764750461 Chr.2: 233434088-233434088 on Build GRCh38
rs752549994 Chr.2: 233434502-233434502 on Build GRCh38
rs889430672 Chr.2: 233434299-233434299 on Build GRCh38
rs760314847 Chr.2: 233434475-233434475 on Build GRCh38
rs149371433 Chr.2: 233434353-233434353 on Build GRCh38
rs1000857452 Chr.2: 233434317-233434317 on Build GRCh38
rs557822875 Chr.2: 233434257-233434257 on Build GRCh38
rs113187512 Chr.2: 233434303-233434303 on Build GRCh38
rs927290517 Chr.2: 233434068-233434068 on Build GRCh38
rs1035236705 Chr.2: 233434272-233434272 on Build GRCh38
rs781381899 Chr.2: 233434396-233434396 on Build GRCh38
rs1048276134 Chr.2: 233434110-233434110 on Build GRCh38
rs765850789 Chr.2: 233434492-233434494 on Build GRCh38
rs780030279 Chr.2: 233434530-233434530 on Build GRCh38
rs751049319 Chr.2: 233434495-233434496 on Build GRCh38
rs931885911 Chr.2: 233434179-233434179 on Build GRCh38
rs750624748 Chr.2: 233434511-233434511 on Build GRCh38
rs756153562 Chr.2: 233434409-233434409 on Build GRCh38
rs758678361 Chr.2: 233434297-233434297 on Build GRCh38
rs1004047699 Chr.2: 233434134-233434134 on Build GRCh38
rs564205238 Chr.2: 233434449-233434449 on Build GRCh38
rs760097406 Chr.2: 233434370-233434370 on Build GRCh38
rs767255891 Chr.2: 233434364-233434364 on Build GRCh38
rs528777926 Chr.2: 233434198-233434198 on Build GRCh38
rs368596307 Chr.2: 233434059-233434059 on Build GRCh38
rs900383083 Chr.2: 233434114-233434114 on Build GRCh38
rs975954671 Chr.2: 233434393-233434393 on Build GRCh38
rs527390856 Chr.2: 233434419-233434419 on Build GRCh38
rs1020006429 Chr.2: 233434255-233434255 on Build GRCh38
rs757979130 Chr.2: 233434487-233434488 on Build GRCh38
rs371847901 Chr.2: 233434438-233434438 on Build GRCh38
rs757528886 Chr.2: 233434395-233434395 on Build GRCh38
rs368190456 Chr.2: 233434250-233434250 on Build GRCh38
rs753930177 Chr.2: 233434531-233434531 on Build GRCh38
rs754613323 Chr.2: 233434499-233434499 on Build GRCh38
rs765141701 Chr.2: 233434500-233434500 on Build GRCh38
rs577997502 Chr.2: 233434263-233434263 on Build GRCh38
rs370747782 Chr.2: 233434067-233434067 on Build GRCh38
rs921756816 Chr.2: 233434456-233434456 on Build GRCh38
rs868267059 Chr.2: 233434379-233434379 on Build GRCh38
rs368023821 Chr.2: 233434375-233434375 on Build GRCh38
rs867076712 Chr.2: 233434421-233434421 on Build GRCh38
rs746029609 Chr.2: 233434406-233434406 on Build GRCh38
rs931912377 Chr.2: 233434478-233434478 on Build GRCh38
rs997876420 Chr.2: 233434201-233434201 on Build GRCh38
rs772740562 Chr.2: 233434467-233434467 on Build GRCh38
rs569907243 Chr.2: 233434109-233434109 on Build GRCh38
rs771916422 Chr.2: 233434450-233434450 on Build GRCh38
rs371795469 Chr.2: 233434326-233434326 on Build GRCh38
rs535322056 Chr.2: 233434165-233434165 on Build GRCh38
rs751031480 Chr.2: 233434385-233434385 on Build GRCh38
rs1040325717 Chr.2: 233434104-233434104 on Build GRCh38
rs375241645 Chr.2: 233434526-233434526 on Build GRCh38
rs192817809 Chr.2: 233434374-233434374 on Build GRCh38
rs776163093 Chr.2: 233434488-233434488 on Build GRCh38
rs138608963 Chr.2: 233434328-233434328 on Build GRCh38
rs759513205 Chr.2: 233434494-233434494 on Build GRCh38
rs888282574 Chr.2: 233434124-233434124 on Build GRCh38
rs752461831 Chr.2: 233434383-233434383 on Build GRCh38
rs1028425605 Chr.2: 233434240-233434240 on Build GRCh38
rs774904512 Chr.2: 233434439-233434439 on Build GRCh38
rs1024365787 Chr.2: 233434454-233434454 on Build GRCh38
rs1036817990 Chr.2: 233434172-233434172 on Build GRCh38
rs199985859 Chr.2: 233434428-233434428 on Build GRCh38
rs765043872 Chr.2: 233434380-233434380 on Build GRCh38
rs902693359 Chr.2: 233434256-233434256 on Build GRCh38
rs907920013 Chr.2: 233434367-233434370 on Build GRCh38
rs144740758 Chr.2: 233434206-233434206 on Build GRCh38
rs371849616 Chr.2: 233434477-233434477 on Build GRCh38
rs1003632796 Chr.2: 233434269-233434269 on Build GRCh38
rs930624260 Chr.2: 233434501-233434501 on Build GRCh38
rs952056513 Chr.2: 233434352-233434352 on Build GRCh38
rs991019816 Chr.2: 233434276-233434276 on Build GRCh38

View Full Product Details