Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 57518390-57518959 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTCFL
Entrez Gene ID
140690
Gene Name
CCCTC-binding factor like
Gene Aliases
BORIS, CT27, CTCF-T, HMGB1L1, dJ579F20.2
UniGene
Hs.131543
Gene Chromosome Location
Chr.20: 57495965-57525652 on Build GRCh38
Transcripts
NM_001269040.1 NM_001269041.1 NM_001269042.1 NM_001269043.1 NM_001269044.1 NM_001269045.1 NM_001269046.1 NM_001269047.1 NM_001269048.1 NM_001269049.1 NM_001269050.1 NM_001269051.1 NM_001269052.1 NM_001269054.1 NM_001269055.1 NM_080618.3 NR_072975.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4897908 c.1015G>A p.E339K Chr.20: 57518802-57518802 on Build GRCh38
7363156 c.1049C>A p.A350D Chr.20: 57518768-57518768 on Build GRCh38
4293306 c.1049C>A p.A350D Chr.20: 57518768-57518768 on Build GRCh38
4293308 c.1015G>T p.E339* Chr.20: 57518802-57518802 on Build GRCh38
4812464 c.988C>T p.R330* Chr.20: 57518829-57518829 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760586764 Chr.20: 57518637-57518637 on Build GRCh38
rs986169986 Chr.20: 57518854-57518854 on Build GRCh38
rs762652848 Chr.20: 57518907-57518907 on Build GRCh38
rs571289232 Chr.20: 57518714-57518714 on Build GRCh38
rs750108384 Chr.20: 57518905-57518905 on Build GRCh38
rs529143854 Chr.20: 57518632-57518632 on Build GRCh38
rs200386629 Chr.20: 57518797-57518797 on Build GRCh38
rs932032823 Chr.20: 57518419-57518419 on Build GRCh38
rs758963630 Chr.20: 57518620-57518620 on Build GRCh38
rs143392924 Chr.20: 57518868-57518868 on Build GRCh38
rs867930389 Chr.20: 57518768-57518768 on Build GRCh38
rs550119711 Chr.20: 57518519-57518519 on Build GRCh38
rs1031351757 Chr.20: 57518795-57518795 on Build GRCh38
rs749958483 Chr.20: 57518761-57518761 on Build GRCh38
rs763928247 Chr.20: 57518713-57518713 on Build GRCh38
rs536770951 Chr.20: 57518766-57518766 on Build GRCh38
rs186835244 Chr.20: 57518646-57518646 on Build GRCh38
rs768537858 Chr.20: 57518883-57518883 on Build GRCh38
rs776430323 Chr.20: 57518896-57518896 on Build GRCh38
rs565533385 Chr.20: 57518652-57518652 on Build GRCh38
rs764875553 Chr.20: 57518749-57518750 on Build GRCh38
rs76451390 Chr.20: 57518422-57518422 on Build GRCh38
rs777120322 Chr.20: 57518651-57518651 on Build GRCh38
rs536304823 Chr.20: 57518658-57518658 on Build GRCh38
rs146122711 Chr.20: 57518832-57518832 on Build GRCh38
rs376829541 Chr.20: 57518869-57518869 on Build GRCh38
rs956619364 Chr.20: 57518819-57518819 on Build GRCh38
rs555474647 Chr.20: 57518794-57518794 on Build GRCh38
rs542500600 Chr.20: 57518833-57518833 on Build GRCh38
rs141558238 Chr.20: 57518565-57518565 on Build GRCh38
rs866938612 Chr.20: 57518753-57518753 on Build GRCh38
rs745326932 Chr.20: 57518610-57518610 on Build GRCh38
rs756884877 Chr.20: 57518593-57518593 on Build GRCh38
rs779610413 Chr.20: 57518780-57518780 on Build GRCh38
rs995538553 Chr.20: 57518856-57518856 on Build GRCh38
rs368021548 Chr.20: 57518668-57518670 on Build GRCh38
rs760649442 Chr.20: 57518875-57518875 on Build GRCh38
rs761534619 Chr.20: 57518900-57518900 on Build GRCh38
rs953501676 Chr.20: 57518851-57518851 on Build GRCh38
rs773315013 Chr.20: 57518515-57518518 on Build GRCh38
rs760812766 Chr.20: 57518829-57518829 on Build GRCh38
rs755518010 Chr.20: 57518805-57518805 on Build GRCh38
rs534534312 Chr.20: 57518686-57518686 on Build GRCh38
rs1035528642 Chr.20: 57518914-57518914 on Build GRCh38
rs904121155 Chr.20: 57518556-57518556 on Build GRCh38
rs929641612 Chr.20: 57518647-57518647 on Build GRCh38
rs1000816044 Chr.20: 57518767-57518767 on Build GRCh38
rs773449360 Chr.20: 57518680-57518680 on Build GRCh38
rs760652104 Chr.20: 57518704-57518704 on Build GRCh38
rs776725257 Chr.20: 57518667-57518669 on Build GRCh38
rs1027288714 Chr.20: 57518894-57518894 on Build GRCh38
rs770977683 Chr.20: 57518845-57518845 on Build GRCh38
rs938439803 Chr.20: 57518443-57518443 on Build GRCh38
rs183111207 Chr.20: 57518903-57518903 on Build GRCh38
rs774424509 Chr.20: 57518579-57518580 on Build GRCh38
rs909368503 Chr.20: 57518514-57518514 on Build GRCh38
rs759501293 Chr.20: 57518675-57518677 on Build GRCh38
rs373623490 Chr.20: 57518859-57518859 on Build GRCh38
rs770566739 Chr.20: 57518662-57518662 on Build GRCh38
rs778431920 Chr.20: 57518604-57518604 on Build GRCh38
rs1044391472 Chr.20: 57518509-57518509 on Build GRCh38
rs928342063 Chr.20: 57518432-57518432 on Build GRCh38
rs781718246 Chr.20: 57518821-57518821 on Build GRCh38
rs761481492 Chr.20: 57518740-57518740 on Build GRCh38
rs767495424 Chr.20: 57518687-57518687 on Build GRCh38
rs768088535 Chr.20: 57518454-57518454 on Build GRCh38
rs963862738 Chr.20: 57518879-57518879 on Build GRCh38
rs780363646 Chr.20: 57518630-57518630 on Build GRCh38
rs769651750 Chr.20: 57518427-57518428 on Build GRCh38
rs915613300 Chr.20: 57518473-57518473 on Build GRCh38
rs533280274 Chr.20: 57518490-57518490 on Build GRCh38
rs192655640 Chr.20: 57518702-57518702 on Build GRCh38
rs765853313 Chr.20: 57518930-57518930 on Build GRCh38

View Full Product Details