Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 33488051-33488638 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC28
Entrez Gene ID
54943
Gene Name
DnaJ heat shock protein family (Hsp40) member C28
Gene Aliases
C21orf55, C21orf78
UniGene
Hs.458308
Gene Chromosome Location
Chr.21: 33488055-33491723 on Build GRCh38
Transcripts
NM_001040192.2 NM_001320746.1 NM_017833.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1666188 c.1073T>G p.L358R Chr.21: 33488321-33488321 on Build GRCh38
4383178 c.926A>T p.K309M Chr.21: 33488468-33488468 on Build GRCh38
4295549 c.942T>G p.F314L Chr.21: 33488452-33488452 on Build GRCh38
419254 c.1110G>C p.K370N Chr.21: 33488284-33488284 on Build GRCh38
5858539 c.761delC p.P254fs*10 Chr.21: 33488633-33488633 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777657441 Chr.21: 33488552-33488552 on Build GRCh38
rs779358335 Chr.21: 33488314-33488314 on Build GRCh38
rs953496920 Chr.21: 33488303-33488303 on Build GRCh38
rs907623229 Chr.21: 33488611-33488611 on Build GRCh38
rs373953010 Chr.21: 33488117-33488117 on Build GRCh38
rs759028742 Chr.21: 33488466-33488466 on Build GRCh38
rs756688991 Chr.21: 33488424-33488424 on Build GRCh38
rs768858415 Chr.21: 33488468-33488471 on Build GRCh38
rs754237121 Chr.21: 33488409-33488409 on Build GRCh38
rs182194192 Chr.21: 33488465-33488465 on Build GRCh38
rs375527360 Chr.21: 33488275-33488275 on Build GRCh38
rs534560693 Chr.21: 33488200-33488200 on Build GRCh38
rs750848168 Chr.21: 33488602-33488602 on Build GRCh38
rs190424708 Chr.21: 33488411-33488411 on Build GRCh38
rs1010543640 Chr.21: 33488254-33488254 on Build GRCh38
rs372123876 Chr.21: 33488257-33488257 on Build GRCh38
rs751729241 Chr.21: 33488513-33488513 on Build GRCh38
rs763992633 Chr.21: 33488251-33488251 on Build GRCh38
rs758965751 Chr.21: 33488509-33488509 on Build GRCh38
rs749448909 Chr.21: 33488450-33488450 on Build GRCh38
rs778647123 Chr.21: 33488381-33488381 on Build GRCh38
rs776588830 Chr.21: 33488366-33488366 on Build GRCh38
rs199590962 Chr.21: 33488595-33488595 on Build GRCh38
rs763196074 Chr.21: 33488603-33488603 on Build GRCh38
rs754570640 Chr.21: 33488286-33488286 on Build GRCh38
rs548149621 Chr.21: 33488307-33488309 on Build GRCh38
rs745998189 Chr.21: 33488339-33488339 on Build GRCh38
rs775370742 Chr.21: 33488345-33488345 on Build GRCh38
rs769843998 Chr.21: 33488546-33488546 on Build GRCh38
rs930807962 Chr.21: 33488109-33488109 on Build GRCh38
rs757788305 Chr.21: 33488307-33488307 on Build GRCh38
rs139852262 Chr.21: 33488443-33488447 on Build GRCh38
rs748990753 Chr.21: 33488558-33488558 on Build GRCh38
rs780212708 Chr.21: 33488416-33488416 on Build GRCh38
rs1019340762 Chr.21: 33488349-33488349 on Build GRCh38
rs151326173 Chr.21: 33488532-33488532 on Build GRCh38
rs774911317 Chr.21: 33488594-33488594 on Build GRCh38
rs922937556 Chr.21: 33488569-33488569 on Build GRCh38
rs770736409 Chr.21: 33488458-33488458 on Build GRCh38
rs772826369 Chr.21: 33488455-33488455 on Build GRCh38
rs763805342 Chr.21: 33488248-33488248 on Build GRCh38
rs748768239 Chr.21: 33488272-33488273 on Build GRCh38
rs778117320 Chr.21: 33488302-33488302 on Build GRCh38
rs372061825 Chr.21: 33488274-33488274 on Build GRCh38
rs564175696 Chr.21: 33488541-33488541 on Build GRCh38
rs749616309 Chr.21: 33488304-33488304 on Build GRCh38
rs966900592 Chr.21: 33488517-33488517 on Build GRCh38
rs78121368 Chr.21: 33488526-33488526 on Build GRCh38
rs962782459 Chr.21: 33488161-33488161 on Build GRCh38
rs147512528 Chr.21: 33488405-33488405 on Build GRCh38
rs756498209 Chr.21: 33488189-33488189 on Build GRCh38
rs761873408 Chr.21: 33488261-33488261 on Build GRCh38
rs745934033 Chr.21: 33488219-33488219 on Build GRCh38
rs752100416 Chr.21: 33488610-33488610 on Build GRCh38
rs1002608190 Chr.21: 33488173-33488173 on Build GRCh38
rs763604079 Chr.21: 33488601-33488601 on Build GRCh38
rs546367322 Chr.21: 33488237-33488237 on Build GRCh38
rs373451141 Chr.21: 33488193-33488193 on Build GRCh38
rs770955229 Chr.21: 33488566-33488566 on Build GRCh38
rs751212470 Chr.21: 33488285-33488285 on Build GRCh38
rs748504016 Chr.21: 33488183-33488183 on Build GRCh38
rs920732882 Chr.21: 33488080-33488080 on Build GRCh38
rs769048458 Chr.21: 33488422-33488422 on Build GRCh38
rs373310996 Chr.21: 33488357-33488357 on Build GRCh38
rs775774536 Chr.21: 33488238-33488238 on Build GRCh38
rs771365218 Chr.21: 33488203-33488203 on Build GRCh38
rs772175920 Chr.21: 33488342-33488342 on Build GRCh38
rs762955572 Chr.21: 33488457-33488457 on Build GRCh38
rs552657108 Chr.21: 33488262-33488262 on Build GRCh38
rs752329100 Chr.21: 33488470-33488470 on Build GRCh38
rs747036845 Chr.21: 33488418-33488418 on Build GRCh38
rs781567691 Chr.21: 33488385-33488386 on Build GRCh38
rs146746889 Chr.21: 33488587-33488587 on Build GRCh38
rs201546020 Chr.21: 33488370-33488370 on Build GRCh38
rs780402290 Chr.21: 33488511-33488511 on Build GRCh38
rs139967340 Chr.21: 33488449-33488449 on Build GRCh38
rs370459473 Chr.21: 33488120-33488120 on Build GRCh38
rs765928995 Chr.21: 33488374-33488374 on Build GRCh38
rs1048134965 Chr.21: 33488145-33488145 on Build GRCh38
rs774449561 Chr.21: 33488216-33488216 on Build GRCh38
rs200537192 Chr.21: 33488279-33488279 on Build GRCh38
rs757834589 Chr.21: 33488180-33488180 on Build GRCh38
rs200406763 Chr.21: 33488605-33488605 on Build GRCh38
rs748024199 Chr.21: 33488445-33488445 on Build GRCh38
rs758762635 Chr.21: 33488413-33488413 on Build GRCh38
rs142250208 Chr.21: 33488124-33488124 on Build GRCh38
rs754660713 Chr.21: 33488387-33488387 on Build GRCh38
rs564467953 Chr.21: 33488378-33488378 on Build GRCh38
rs771581769 Chr.21: 33488590-33488590 on Build GRCh38
rs142171090 Chr.21: 33488483-33488483 on Build GRCh38
rs570372214 Chr.21: 33488155-33488155 on Build GRCh38
rs760259315 Chr.21: 33488477-33488477 on Build GRCh38
rs750721974 Chr.21: 33488390-33488391 on Build GRCh38
rs781534131 Chr.21: 33488436-33488436 on Build GRCh38
rs760442498 Chr.21: 33488239-33488239 on Build GRCh38
rs765636057 Chr.21: 33488480-33488480 on Build GRCh38
rs3834674 Chr.21: 33488450-33488454 on Build GRCh38
rs778313340 Chr.21: 33488191-33488191 on Build GRCh38
rs747258791 Chr.21: 33488354-33488354 on Build GRCh38
rs904487470 Chr.21: 33488576-33488576 on Build GRCh38
rs773198313 Chr.21: 33488371-33488371 on Build GRCh38
rs960713125 Chr.21: 33488468-33488468 on Build GRCh38
rs1033798340 Chr.21: 33488190-33488190 on Build GRCh38
rs759102103 Chr.21: 33488589-33488589 on Build GRCh38

View Full Product Details