Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 26467588-26468131 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYYR1
Entrez Gene ID
116159
Gene Name
cysteine and tyrosine rich 1
Gene Aliases
C21orf95
UniGene
Hs.37445
Gene Chromosome Location
Chr.21: 26466209-26573404 on Build GRCh38
Transcripts
NM_001320768.1 NM_052954.3 NR_135472.1 NR_135515.1 NR_135516.1

Gene Symbol
CYYR1-AS1
Entrez Gene ID
100996571
Gene Name
cysteine and tyrosine rich 1 antisense RNA 1
Gene Aliases
-
UniGene
Hs.570432
Gene Chromosome Location
Chr.21: 26393635-26569252 on Build GRCh38
Transcripts
NM_001320768.1 NM_052954.3 NR_135472.1 NR_135515.1 NR_135516.1

SNP Detail

SNP ID Chromosome Location
rs939809570 Chr.21: 26467620-26467621 on Build GRCh38
rs547792014 Chr.21: 26467794-26467794 on Build GRCh38
rs891700981 Chr.21: 26467672-26467672 on Build GRCh38
rs115409952 Chr.21: 26467945-26467945 on Build GRCh38
rs1027711481 Chr.21: 26467772-26467772 on Build GRCh38
rs1040304034 Chr.21: 26467923-26467923 on Build GRCh38
rs1046277077 Chr.21: 26467621-26467621 on Build GRCh38
rs775068702 Chr.21: 26467842-26467842 on Build GRCh38
rs952285194 Chr.21: 26468044-26468044 on Build GRCh38
rs1008856880 Chr.21: 26467716-26467716 on Build GRCh38
rs531771598 Chr.21: 26467689-26467689 on Build GRCh38
rs561183072 Chr.21: 26467822-26467822 on Build GRCh38
rs745379605 Chr.21: 26467785-26467785 on Build GRCh38
rs931986979 Chr.21: 26467939-26467939 on Build GRCh38
rs554451224 Chr.21: 26467993-26467993 on Build GRCh38
rs957331733 Chr.21: 26468072-26468072 on Build GRCh38
rs219642 Chr.21: 26467901-26467901 on Build GRCh38
rs981540394 Chr.21: 26468080-26468080 on Build GRCh38
rs773348781 Chr.21: 26468084-26468084 on Build GRCh38
rs1002128356 Chr.21: 26467656-26467656 on Build GRCh38
rs558534976 Chr.21: 26467838-26467838 on Build GRCh38
rs767529730 Chr.21: 26468052-26468052 on Build GRCh38
rs1010558004 Chr.21: 26467669-26467669 on Build GRCh38
rs576597536 Chr.21: 26468043-26468043 on Build GRCh38
rs191044338 Chr.21: 26467990-26467990 on Build GRCh38
rs887906433 Chr.21: 26467779-26467779 on Build GRCh38
rs537617941 Chr.21: 26467873-26467873 on Build GRCh38
rs542914924 Chr.21: 26467735-26467735 on Build GRCh38
rs893466231 Chr.21: 26467668-26467668 on Build GRCh38
rs1014230985 Chr.21: 26468027-26468027 on Build GRCh38
rs566819993 Chr.21: 26467825-26467825 on Build GRCh38
rs2830239 Chr.21: 26467978-26467978 on Build GRCh38
rs1008993293 Chr.21: 26467738-26467738 on Build GRCh38
rs935824516 Chr.21: 26467635-26467635 on Build GRCh38
rs1023949425 Chr.21: 26467811-26467811 on Build GRCh38
rs1010505585 Chr.21: 26467908-26467908 on Build GRCh38
rs927815660 Chr.21: 26467846-26467846 on Build GRCh38
rs1032719415 Chr.21: 26468066-26468066 on Build GRCh38
rs114334580 Chr.21: 26467874-26467874 on Build GRCh38
rs760824288 Chr.21: 26468111-26468111 on Build GRCh38
rs147569763 Chr.21: 26467909-26467909 on Build GRCh38
rs546801413 Chr.21: 26467975-26467975 on Build GRCh38
rs553346111 Chr.21: 26467935-26467935 on Build GRCh38
rs887854045 Chr.21: 26467969-26467969 on Build GRCh38
rs879143865 Chr.21: 26467892-26467892 on Build GRCh38
rs144992751 Chr.21: 26467665-26467665 on Build GRCh38
rs181926961 Chr.21: 26467637-26467637 on Build GRCh38
rs182829858 Chr.21: 26468065-26468065 on Build GRCh38
rs568432845 Chr.21: 26467761-26467761 on Build GRCh38
rs1019121038 Chr.21: 26467747-26467747 on Build GRCh38
rs935801470 Chr.21: 26467867-26467867 on Build GRCh38
rs768291643 Chr.21: 26467925-26467925 on Build GRCh38
rs575311560 Chr.21: 26467727-26467727 on Build GRCh38
rs569039808 Chr.21: 26467845-26467845 on Build GRCh38
rs77041278 Chr.21: 26467787-26467787 on Build GRCh38
rs1005005674 Chr.21: 26467973-26467973 on Build GRCh38
rs143100369 Chr.21: 26468056-26468056 on Build GRCh38
rs142060580 Chr.21: 26468020-26468020 on Build GRCh38
rs186242798 Chr.21: 26467827-26467827 on Build GRCh38
rs371812343 Chr.21: 26467789-26467789 on Build GRCh38
rs996185888 Chr.21: 26467748-26467748 on Build GRCh38

View Full Product Details