Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 20086212-20086751 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DGCR8
Entrez Gene ID
54487
Gene Name
DGCR8 microprocessor complex subunit
Gene Aliases
C22orf12, DGCRK6, Gy1, pasha
UniGene
Hs.643452
Gene Chromosome Location
Chr.22: 20080232-20111877 on Build GRCh38
Transcripts
NM_001190326.1 NM_022720.6

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6196873 c.276T>A p.S92R Chr.22: 20086239-20086239 on Build GRCh38
4677161 c.401A>C p.K134T Chr.22: 20086364-20086364 on Build GRCh38
2892677 c.495C>T p.G165G Chr.22: 20086458-20086458 on Build GRCh38
6211292 c.459C>T p.S153S Chr.22: 20086422-20086422 on Build GRCh38
3800039 c.267G>A p.P89P Chr.22: 20086230-20086230 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs929456723 Chr.22: 20086618-20086618 on Build GRCh38
rs1056127835 Chr.22: 20086261-20086261 on Build GRCh38
rs774123790 Chr.22: 20086280-20086280 on Build GRCh38
rs758424877 Chr.22: 20086339-20086339 on Build GRCh38
rs754945301 Chr.22: 20086536-20086536 on Build GRCh38
rs755158789 Chr.22: 20086251-20086251 on Build GRCh38
rs768183516 Chr.22: 20086279-20086279 on Build GRCh38
rs1025753898 Chr.22: 20086287-20086287 on Build GRCh38
rs1012463946 Chr.22: 20086275-20086275 on Build GRCh38
rs916217530 Chr.22: 20086474-20086474 on Build GRCh38
rs999724184 Chr.22: 20086611-20086611 on Build GRCh38
rs1053854267 Chr.22: 20086244-20086244 on Build GRCh38
rs118025402 Chr.22: 20086396-20086396 on Build GRCh38
rs766698132 Chr.22: 20086311-20086311 on Build GRCh38
rs541503891 Chr.22: 20086489-20086489 on Build GRCh38
rs147225345 Chr.22: 20086459-20086459 on Build GRCh38
rs948298108 Chr.22: 20086476-20086476 on Build GRCh38
rs761772756 Chr.22: 20086510-20086510 on Build GRCh38
rs201435288 Chr.22: 20086239-20086239 on Build GRCh38
rs150234024 Chr.22: 20086258-20086258 on Build GRCh38
rs780100516 Chr.22: 20086364-20086364 on Build GRCh38
rs369938408 Chr.22: 20086327-20086327 on Build GRCh38
rs748162875 Chr.22: 20086253-20086253 on Build GRCh38
rs769062987 Chr.22: 20086670-20086670 on Build GRCh38
rs376763977 Chr.22: 20086411-20086411 on Build GRCh38
rs536949587 Chr.22: 20086473-20086473 on Build GRCh38
rs1056538434 Chr.22: 20086460-20086460 on Build GRCh38
rs376007260 Chr.22: 20086437-20086437 on Build GRCh38
rs745740923 Chr.22: 20086346-20086346 on Build GRCh38
rs367625866 Chr.22: 20086647-20086647 on Build GRCh38
rs753674680 Chr.22: 20086529-20086529 on Build GRCh38
rs767654784 Chr.22: 20086517-20086517 on Build GRCh38
rs751003558 Chr.22: 20086237-20086237 on Build GRCh38
rs907653514 Chr.22: 20086421-20086421 on Build GRCh38
rs766869709 Chr.22: 20086248-20086248 on Build GRCh38
rs763168480 Chr.22: 20086404-20086404 on Build GRCh38
rs377706846 Chr.22: 20086301-20086301 on Build GRCh38
rs747821585 Chr.22: 20086389-20086389 on Build GRCh38
rs770524071 Chr.22: 20086467-20086467 on Build GRCh38
rs754347281 Chr.22: 20086600-20086602 on Build GRCh38
rs779195256 Chr.22: 20086690-20086691 on Build GRCh38
rs747733681 Chr.22: 20086426-20086426 on Build GRCh38
rs754677487 Chr.22: 20086424-20086424 on Build GRCh38
rs372483452 Chr.22: 20086338-20086338 on Build GRCh38
rs370659006 Chr.22: 20086622-20086622 on Build GRCh38
rs777187411 Chr.22: 20086706-20086706 on Build GRCh38
rs142171508 Chr.22: 20086623-20086623 on Build GRCh38
rs758334658 Chr.22: 20086614-20086614 on Build GRCh38
rs755366185 Chr.22: 20086646-20086647 on Build GRCh38
rs775155216 Chr.22: 20086269-20086269 on Build GRCh38
rs182736423 Chr.22: 20086250-20086250 on Build GRCh38
rs764663100 Chr.22: 20086667-20086667 on Build GRCh38
rs764011063 Chr.22: 20086711-20086711 on Build GRCh38
rs149913549 Chr.22: 20086680-20086680 on Build GRCh38
rs144722138 Chr.22: 20086397-20086397 on Build GRCh38
rs778785404 Chr.22: 20086398-20086398 on Build GRCh38
rs778791032 Chr.22: 20086560-20086560 on Build GRCh38
rs370350957 Chr.22: 20086392-20086392 on Build GRCh38
rs201293912 Chr.22: 20086400-20086400 on Build GRCh38
rs777536932 Chr.22: 20086615-20086615 on Build GRCh38
rs143802353 Chr.22: 20086414-20086414 on Build GRCh38
rs1045028033 Chr.22: 20086540-20086540 on Build GRCh38
rs139146240 Chr.22: 20086635-20086635 on Build GRCh38
rs35987994 Chr.22: 20086483-20086483 on Build GRCh38
rs755427841 Chr.22: 20086326-20086326 on Build GRCh38
rs767182028 Chr.22: 20086290-20086290 on Build GRCh38
rs761372486 Chr.22: 20086289-20086289 on Build GRCh38
rs199753950 Chr.22: 20086567-20086567 on Build GRCh38
rs774504190 Chr.22: 20086679-20086679 on Build GRCh38
rs745828702 Chr.22: 20086260-20086260 on Build GRCh38
rs376960661 Chr.22: 20086347-20086347 on Build GRCh38
rs138332207 Chr.22: 20086407-20086407 on Build GRCh38
rs749037488 Chr.22: 20086372-20086372 on Build GRCh38
rs547571792 Chr.22: 20086387-20086387 on Build GRCh38
rs776046259 Chr.22: 20086636-20086636 on Build GRCh38
rs746681654 Chr.22: 20086616-20086616 on Build GRCh38
rs769539602 Chr.22: 20086266-20086266 on Build GRCh38
rs754166512 Chr.22: 20086325-20086325 on Build GRCh38
rs374194876 Chr.22: 20086257-20086257 on Build GRCh38
rs761292966 Chr.22: 20086302-20086302 on Build GRCh38
rs867948445 Chr.22: 20086532-20086532 on Build GRCh38
rs773757856 Chr.22: 20086382-20086382 on Build GRCh38
rs765262070 Chr.22: 20086701-20086701 on Build GRCh38
rs192598875 Chr.22: 20086401-20086401 on Build GRCh38
rs751823713 Chr.22: 20086500-20086500 on Build GRCh38
rs780763970 Chr.22: 20086725-20086725 on Build GRCh38
rs751629283 Chr.22: 20086405-20086405 on Build GRCh38
rs773358048 Chr.22: 20086686-20086686 on Build GRCh38
rs34301880 Chr.22: 20086632-20086632 on Build GRCh38
rs746513636 Chr.22: 20086462-20086462 on Build GRCh38
rs775847917 Chr.22: 20086487-20086487 on Build GRCh38
rs778403931 Chr.22: 20086388-20086388 on Build GRCh38
rs766811802 Chr.22: 20086504-20086506 on Build GRCh38
rs767022640 Chr.22: 20086451-20086451 on Build GRCh38
rs778796867 Chr.22: 20086425-20086425 on Build GRCh38
rs555922599 Chr.22: 20086477-20086477 on Build GRCh38
rs989729711 Chr.22: 20086464-20086464 on Build GRCh38
rs761204926 Chr.22: 20086393-20086393 on Build GRCh38
rs748916061 Chr.22: 20086274-20086274 on Build GRCh38
rs371515546 Chr.22: 20086693-20086693 on Build GRCh38
rs771665333 Chr.22: 20086391-20086391 on Build GRCh38
rs777864546 Chr.22: 20086343-20086343 on Build GRCh38
rs138886652 Chr.22: 20086291-20086291 on Build GRCh38
rs886170342 Chr.22: 20086241-20086241 on Build GRCh38
rs778992846 Chr.22: 20086252-20086252 on Build GRCh38
rs779829769 Chr.22: 20086408-20086408 on Build GRCh38
rs758218351 Chr.22: 20086707-20086707 on Build GRCh38
rs764298892 Chr.22: 20086490-20086490 on Build GRCh38
rs767432583 Chr.22: 20086681-20086681 on Build GRCh38
rs751104719 Chr.22: 20086714-20086714 on Build GRCh38

View Full Product Details