Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 19888249-19888801 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFHB
Entrez Gene ID
151651
Gene Name
EF-hand domain family member B
Gene Aliases
CFAP21
UniGene
Hs.733920
Gene Chromosome Location
Chr.3: 19879472-19947036 on Build GRCh38
Transcripts
NM_144715.3 XM_005264889.1 XM_011533382.1 XM_011533383.2 XM_011533385.2 XM_017005741.1 XM_017005743.1 XM_017005744.1 XM_017005745.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4680319 c.1744G>A p.D582N Chr.3: 19888633-19888633 on Build GRCh38
2948281 c.1905A>C p.K635N Chr.3: 19888472-19888472 on Build GRCh38
5632598 c.1821C>T p.D607D Chr.3: 19888556-19888556 on Build GRCh38
2948283 c.1885A>G p.K629E Chr.3: 19888492-19888492 on Build GRCh38
4769964 c.1867G>T p.A623S Chr.3: 19888510-19888510 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs139405916 Chr.3: 19888604-19888604 on Build GRCh38
rs761961963 Chr.3: 19888539-19888539 on Build GRCh38
rs762051850 Chr.3: 19888442-19888442 on Build GRCh38
rs988792045 Chr.3: 19888574-19888574 on Build GRCh38
rs932467885 Chr.3: 19888646-19888646 on Build GRCh38
rs1015233546 Chr.3: 19888483-19888483 on Build GRCh38
rs1036644573 Chr.3: 19888712-19888712 on Build GRCh38
rs536360265 Chr.3: 19888366-19888366 on Build GRCh38
rs952716481 Chr.3: 19888625-19888625 on Build GRCh38
rs553536568 Chr.3: 19888569-19888569 on Build GRCh38
rs776556971 Chr.3: 19888648-19888648 on Build GRCh38
rs763447735 Chr.3: 19888575-19888575 on Build GRCh38
rs746207312 Chr.3: 19888624-19888624 on Build GRCh38
rs76305623 Chr.3: 19888638-19888638 on Build GRCh38
rs758886256 Chr.3: 19888626-19888626 on Build GRCh38
rs143556620 Chr.3: 19888499-19888499 on Build GRCh38
rs895533023 Chr.3: 19888773-19888773 on Build GRCh38
rs371381180 Chr.3: 19888711-19888711 on Build GRCh38
rs111751840 Chr.3: 19888683-19888683 on Build GRCh38
rs148888292 Chr.3: 19888321-19888321 on Build GRCh38
rs1043405454 Chr.3: 19888496-19888496 on Build GRCh38
rs767174997 Chr.3: 19888700-19888700 on Build GRCh38
rs747406287 Chr.3: 19888633-19888633 on Build GRCh38
rs1049562332 Chr.3: 19888676-19888676 on Build GRCh38
rs763182761 Chr.3: 19888696-19888696 on Build GRCh38
rs978689145 Chr.3: 19888726-19888726 on Build GRCh38
rs780492640 Chr.3: 19888628-19888628 on Build GRCh38
rs1001290013 Chr.3: 19888302-19888302 on Build GRCh38
rs752217869 Chr.3: 19888467-19888467 on Build GRCh38
rs35769819 Chr.3: 19888326-19888327 on Build GRCh38
rs765497812 Chr.3: 19888543-19888543 on Build GRCh38
rs186260258 Chr.3: 19888380-19888380 on Build GRCh38
rs746293003 Chr.3: 19888510-19888510 on Build GRCh38
rs919929617 Chr.3: 19888390-19888390 on Build GRCh38
rs777363241 Chr.3: 19888490-19888490 on Build GRCh38
rs547236720 Chr.3: 19888317-19888317 on Build GRCh38
rs952859371 Chr.3: 19888395-19888395 on Build GRCh38
rs1013720913 Chr.3: 19888541-19888541 on Build GRCh38
rs768324052 Chr.3: 19888587-19888587 on Build GRCh38
rs751369107 Chr.3: 19888462-19888462 on Build GRCh38
rs575985322 Chr.3: 19888411-19888411 on Build GRCh38
rs936967356 Chr.3: 19888752-19888752 on Build GRCh38
rs776619228 Chr.3: 19888630-19888633 on Build GRCh38
rs1034123966 Chr.3: 19888305-19888305 on Build GRCh38
rs995297385 Chr.3: 19888505-19888505 on Build GRCh38
rs985489547 Chr.3: 19888404-19888404 on Build GRCh38
rs1027670980 Chr.3: 19888596-19888596 on Build GRCh38
rs143232041 Chr.3: 19888619-19888619 on Build GRCh38
rs755625111 Chr.3: 19888485-19888485 on Build GRCh38
rs776774625 Chr.3: 19888537-19888537 on Build GRCh38
rs943852225 Chr.3: 19888426-19888426 on Build GRCh38
rs925874936 Chr.3: 19888745-19888745 on Build GRCh38
rs746603079 Chr.3: 19888409-19888411 on Build GRCh38
rs891021212 Chr.3: 19888686-19888686 on Build GRCh38
rs144314217 Chr.3: 19888617-19888617 on Build GRCh38
rs1025153767 Chr.3: 19888558-19888558 on Build GRCh38
rs760789974 Chr.3: 19888522-19888522 on Build GRCh38
rs1055469716 Chr.3: 19888765-19888765 on Build GRCh38
rs866107748 Chr.3: 19888717-19888717 on Build GRCh38
rs781168489 Chr.3: 19888466-19888466 on Build GRCh38
rs748289194 Chr.3: 19888680-19888680 on Build GRCh38
rs868683716 Chr.3: 19888339-19888339 on Build GRCh38
rs4858714 Chr.3: 19888511-19888511 on Build GRCh38
rs759365986 Chr.3: 19888554-19888554 on Build GRCh38
rs574334089 Chr.3: 19888611-19888611 on Build GRCh38
rs775905775 Chr.3: 19888520-19888520 on Build GRCh38
rs1025207475 Chr.3: 19888361-19888361 on Build GRCh38
rs775627295 Chr.3: 19888401-19888401 on Build GRCh38
rs536684675 Chr.3: 19888429-19888429 on Build GRCh38
rs113536030 Chr.3: 19888562-19888562 on Build GRCh38
rs913108329 Chr.3: 19888470-19888470 on Build GRCh38
rs767466744 Chr.3: 19888561-19888561 on Build GRCh38
rs752526302 Chr.3: 19888568-19888568 on Build GRCh38
rs976559215 Chr.3: 19888434-19888434 on Build GRCh38
rs754058323 Chr.3: 19888427-19888427 on Build GRCh38
rs774128621 Chr.3: 19888546-19888546 on Build GRCh38
rs779367298 Chr.3: 19888501-19888501 on Build GRCh38
rs866664842 Chr.3: 19888481-19888481 on Build GRCh38
rs141867051 Chr.3: 19888465-19888465 on Build GRCh38
rs148022259 Chr.3: 19888707-19888707 on Build GRCh38
rs558784131 Chr.3: 19888453-19888453 on Build GRCh38
rs768575907 Chr.3: 19888526-19888526 on Build GRCh38
rs748855589 Chr.3: 19888493-19888493 on Build GRCh38
rs760733942 Chr.3: 19888403-19888403 on Build GRCh38
rs191556621 Chr.3: 19888684-19888684 on Build GRCh38

View Full Product Details