Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 38478945-38479436 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACVR2B
Entrez Gene ID
93
Gene Name
activin A receptor type 2B
Gene Aliases
ACTRIIB, ActR-IIB, HTX4
UniGene
Hs.174273
Gene Chromosome Location
Chr.3: 38454299-38493142 on Build GRCh38
Transcripts
NM_001106.3 XM_005265583.3 XM_017007514.1 XM_017007515.1 XM_017007516.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6164371 c.684G>C p.Q228H Chr.3: 38479145-38479145 on Build GRCh38
6593259 c.699C>G p.I233M Chr.3: 38479160-38479160 on Build GRCh38
3357706 c.676T>C p.S226P Chr.3: 38479137-38479137 on Build GRCh38
1566789 c.691C>T p.R231W Chr.3: 38479152-38479152 on Build GRCh38
4659220 c.772G>A p.E258K Chr.3: 38479233-38479233 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780335146 Chr.3: 38479292-38479292 on Build GRCh38
rs773064086 Chr.3: 38479089-38479089 on Build GRCh38
rs376462721 Chr.3: 38479139-38479139 on Build GRCh38
rs768528311 Chr.3: 38479388-38479388 on Build GRCh38
rs186009413 Chr.3: 38479119-38479119 on Build GRCh38
rs543794 Chr.3: 38479322-38479322 on Build GRCh38
rs557220149 Chr.3: 38479003-38479003 on Build GRCh38
rs908792189 Chr.3: 38479049-38479049 on Build GRCh38
rs200389337 Chr.3: 38479087-38479087 on Build GRCh38
rs370618639 Chr.3: 38479303-38479303 on Build GRCh38
rs543751289 Chr.3: 38478992-38478992 on Build GRCh38
rs762748057 Chr.3: 38479100-38479100 on Build GRCh38
rs746287509 Chr.3: 38478991-38478991 on Build GRCh38
rs559220080 Chr.3: 38479097-38479097 on Build GRCh38
rs755621345 Chr.3: 38479115-38479115 on Build GRCh38
rs550140682 Chr.3: 38479353-38479353 on Build GRCh38
rs776493809 Chr.3: 38479276-38479277 on Build GRCh38
rs145988028 Chr.3: 38479205-38479205 on Build GRCh38
rs34730163 Chr.3: 38479379-38479380 on Build GRCh38
rs749214138 Chr.3: 38479199-38479199 on Build GRCh38
rs770766278 Chr.3: 38479203-38479203 on Build GRCh38
rs575664601 Chr.3: 38479066-38479066 on Build GRCh38
rs908442251 Chr.3: 38479153-38479153 on Build GRCh38
rs540729310 Chr.3: 38479236-38479236 on Build GRCh38
rs754436899 Chr.3: 38479233-38479233 on Build GRCh38
rs894650369 Chr.3: 38479385-38479385 on Build GRCh38
rs773938193 Chr.3: 38479102-38479102 on Build GRCh38
rs754829682 Chr.3: 38479336-38479336 on Build GRCh38
rs944251733 Chr.3: 38479069-38479069 on Build GRCh38
rs370988039 Chr.3: 38479118-38479118 on Build GRCh38
rs893771246 Chr.3: 38479105-38479105 on Build GRCh38
rs746734251 Chr.3: 38479370-38479370 on Build GRCh38
rs190959821 Chr.3: 38479387-38479387 on Build GRCh38
rs752285756 Chr.3: 38479113-38479113 on Build GRCh38
rs912203722 Chr.3: 38478977-38478977 on Build GRCh38
rs55945487 Chr.3: 38479029-38479029 on Build GRCh38
rs564309911 Chr.3: 38479237-38479237 on Build GRCh38
rs975562450 Chr.3: 38479023-38479023 on Build GRCh38
rs1047304657 Chr.3: 38479047-38479047 on Build GRCh38
rs781213507 Chr.3: 38479318-38479318 on Build GRCh38
rs779306516 Chr.3: 38479138-38479138 on Build GRCh38
rs970606066 Chr.3: 38479152-38479152 on Build GRCh38
rs759214358 Chr.3: 38479206-38479206 on Build GRCh38
rs569952032 Chr.3: 38479384-38479384 on Build GRCh38
rs1041595310 Chr.3: 38479078-38479078 on Build GRCh38
rs534516 Chr.3: 38479150-38479150 on Build GRCh38
rs747841186 Chr.3: 38479299-38479300 on Build GRCh38
rs747010144 Chr.3: 38479082-38479082 on Build GRCh38
rs757764446 Chr.3: 38479132-38479132 on Build GRCh38
rs533731 Chr.3: 38479083-38479083 on Build GRCh38
rs780014318 Chr.3: 38479163-38479163 on Build GRCh38
rs1021898981 Chr.3: 38479404-38479404 on Build GRCh38
rs147640696 Chr.3: 38479184-38479184 on Build GRCh38
rs751764494 Chr.3: 38479297-38479297 on Build GRCh38
rs533158678 Chr.3: 38479287-38479287 on Build GRCh38
rs149589197 Chr.3: 38479211-38479211 on Build GRCh38
rs767032109 Chr.3: 38479110-38479110 on Build GRCh38
rs111619406 Chr.3: 38479024-38479024 on Build GRCh38
rs2075705 Chr.3: 38478984-38478984 on Build GRCh38
rs768662356 Chr.3: 38479175-38479175 on Build GRCh38
rs140053752 Chr.3: 38479180-38479180 on Build GRCh38
rs527396511 Chr.3: 38479232-38479232 on Build GRCh38
rs760321356 Chr.3: 38479212-38479212 on Build GRCh38
rs776031478 Chr.3: 38479053-38479053 on Build GRCh38
rs750829648 Chr.3: 38479268-38479268 on Build GRCh38
rs931113335 Chr.3: 38479114-38479114 on Build GRCh38
rs997314327 Chr.3: 38479101-38479101 on Build GRCh38
rs541402369 Chr.3: 38479104-38479104 on Build GRCh38
rs983108008 Chr.3: 38479329-38479329 on Build GRCh38
rs758668102 Chr.3: 38479147-38479147 on Build GRCh38
rs976963607 Chr.3: 38479402-38479402 on Build GRCh38
rs544872 Chr.3: 38479160-38479160 on Build GRCh38
rs34815229 Chr.3: 38479148-38479148 on Build GRCh38

View Full Product Details