Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 39333456-39334053 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCR8
Entrez Gene ID
1237
Gene Name
C-C motif chemokine receptor 8
Gene Aliases
CC-CKR-8, CCR-8, CDw198, CKRL1, CMKBR8, CMKBRL2, CY6, GPRCY6, TER1
UniGene
Hs.113222
Gene Chromosome Location
Chr.3: 39329706-39333680 on Build GRCh38
Transcripts
NM_005201.3

SNP Detail

SNP ID Chromosome Location
rs998905182 Chr.3: 39333711-39333711 on Build GRCh38
rs572840979 Chr.3: 39333607-39333607 on Build GRCh38
rs758539947 Chr.3: 39333925-39333925 on Build GRCh38
rs1054566174 Chr.3: 39334025-39334025 on Build GRCh38
rs191453375 Chr.3: 39333779-39333779 on Build GRCh38
rs974441597 Chr.3: 39333875-39333875 on Build GRCh38
rs532581916 Chr.3: 39333900-39333900 on Build GRCh38
rs552601424 Chr.3: 39333604-39333604 on Build GRCh38
rs765704760 Chr.3: 39333699-39333699 on Build GRCh38
rs907168602 Chr.3: 39334022-39334022 on Build GRCh38
rs146639474 Chr.3: 39333750-39333750 on Build GRCh38
rs184013272 Chr.3: 39333808-39333808 on Build GRCh38
rs868388623 Chr.3: 39333518-39333518 on Build GRCh38
rs932837555 Chr.3: 39333632-39333632 on Build GRCh38
rs1023059271 Chr.3: 39333566-39333566 on Build GRCh38
rs191884340 Chr.3: 39333719-39333719 on Build GRCh38
rs777124317 Chr.3: 39333626-39333626 on Build GRCh38
rs990275193 Chr.3: 39333754-39333754 on Build GRCh38
rs552275320 Chr.3: 39333912-39333912 on Build GRCh38
rs1033314595 Chr.3: 39333517-39333517 on Build GRCh38
rs1012825557 Chr.3: 39333757-39333757 on Build GRCh38
rs766239394 Chr.3: 39333649-39333649 on Build GRCh38
rs867474269 Chr.3: 39333506-39333506 on Build GRCh38
rs866922979 Chr.3: 39333505-39333505 on Build GRCh38
rs912728125 Chr.3: 39333667-39333667 on Build GRCh38
rs961354094 Chr.3: 39333584-39333584 on Build GRCh38
rs2088668 Chr.3: 39333924-39333924 on Build GRCh38
rs374147848 Chr.3: 39333738-39333739 on Build GRCh38
rs971199628 Chr.3: 39333767-39333767 on Build GRCh38
rs934353937 Chr.3: 39334024-39334024 on Build GRCh38
rs76294985 Chr.3: 39333976-39333976 on Build GRCh38
rs141400466 Chr.3: 39333991-39333991 on Build GRCh38
rs971502818 Chr.3: 39333570-39333570 on Build GRCh38
rs774969927 Chr.3: 39333913-39333913 on Build GRCh38
rs547120095 Chr.3: 39333798-39333798 on Build GRCh38
rs1042563135 Chr.3: 39333855-39333855 on Build GRCh38
rs971275755 Chr.3: 39333679-39333679 on Build GRCh38
rs35383683 Chr.3: 39333937-39333938 on Build GRCh38
rs996659138 Chr.3: 39333521-39333521 on Build GRCh38
rs867248966 Chr.3: 39333543-39333543 on Build GRCh38
rs957247698 Chr.3: 39333526-39333526 on Build GRCh38
rs781162005 Chr.3: 39334026-39334026 on Build GRCh38
rs1027491829 Chr.3: 39333523-39333523 on Build GRCh38
rs1022842291 Chr.3: 39333760-39333760 on Build GRCh38
rs991694500 Chr.3: 39333535-39333535 on Build GRCh38
rs544668169 Chr.3: 39333610-39333610 on Build GRCh38
rs565216750 Chr.3: 39333677-39333677 on Build GRCh38
rs574173549 Chr.3: 39333590-39333590 on Build GRCh38
rs1040120045 Chr.3: 39333980-39333980 on Build GRCh38
rs184116659 Chr.3: 39333723-39333723 on Build GRCh38
rs902871898 Chr.3: 39333709-39333709 on Build GRCh38
rs760045721 Chr.3: 39333681-39333681 on Build GRCh38
rs111800208 Chr.3: 39333721-39333721 on Build GRCh38
rs186682298 Chr.3: 39333739-39333739 on Build GRCh38
rs559630340 Chr.3: 39333935-39333935 on Build GRCh38
rs981520144 Chr.3: 39333572-39333572 on Build GRCh38
rs762693542 Chr.3: 39333810-39333810 on Build GRCh38
rs145197183 Chr.3: 39333577-39333577 on Build GRCh38
rs981108373 Chr.3: 39333690-39333690 on Build GRCh38
rs924217259 Chr.3: 39333868-39333868 on Build GRCh38

View Full Product Details