Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 24037486-24038072 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FKBP1B
Entrez Gene ID
2281
Gene Name
FK506 binding protein 1B
Gene Aliases
FKBP12.6, FKBP1L, OTK4, PKBP1L, PPIase
UniGene
Hs.709461
Gene Chromosome Location
Chr.2: 24033205-24067743 on Build GRCh38
Transcripts
XM_017003594.1 NM_001142319.1 NM_025203.2 XM_017005029.1

Gene Symbol
WDCP
Entrez Gene ID
80304
Gene Name
WD repeat and coiled coil containing
Gene Aliases
C2orf44, PP384
UniGene
Hs.24624
Gene Chromosome Location
Chr.2: 24029336-24047426 on Build GRCh38
Transcripts
XM_017003594.1 NM_001142319.1 NM_025203.2 XM_017005029.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5615053 c.1630C>T p.Q544* Chr.2: 24037865-24037865 on Build GRCh38
6091155 c.1594G>A p.D532N Chr.2: 24037901-24037901 on Build GRCh38
4624727 c.1451C>T p.T484I Chr.2: 24038044-24038044 on Build GRCh38
4093125 c.1655A>G p.K552R Chr.2: 24037840-24037840 on Build GRCh38
7136874 c.1568C>T p.S523L Chr.2: 24037927-24037927 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748703105 Chr.2: 24037950-24037950 on Build GRCh38
rs971052835 Chr.2: 24037743-24037743 on Build GRCh38
rs187108244 Chr.2: 24037734-24037734 on Build GRCh38
rs772816479 Chr.2: 24037874-24037874 on Build GRCh38
rs752505884 Chr.2: 24037832-24037832 on Build GRCh38
rs150149695 Chr.2: 24037708-24037708 on Build GRCh38
rs893367506 Chr.2: 24037706-24037706 on Build GRCh38
rs777903295 Chr.2: 24037629-24037629 on Build GRCh38
rs761234536 Chr.2: 24037689-24037689 on Build GRCh38
rs564772738 Chr.2: 24037651-24037651 on Build GRCh38
rs903262119 Chr.2: 24037919-24037919 on Build GRCh38
rs906839816 Chr.2: 24038026-24038026 on Build GRCh38
rs761425312 Chr.2: 24037717-24037717 on Build GRCh38
rs201244599 Chr.2: 24037858-24037858 on Build GRCh38
rs760598586 Chr.2: 24037905-24037905 on Build GRCh38
rs749914359 Chr.2: 24037579-24037579 on Build GRCh38
rs775986747 Chr.2: 24037685-24037685 on Build GRCh38
rs750714575 Chr.2: 24037841-24037841 on Build GRCh38
rs995324153 Chr.2: 24038047-24038047 on Build GRCh38
rs762848759 Chr.2: 24037724-24037724 on Build GRCh38
rs759201973 Chr.2: 24037804-24037804 on Build GRCh38
rs781724848 Chr.2: 24037864-24037864 on Build GRCh38
rs758591083 Chr.2: 24037842-24037842 on Build GRCh38
rs549738600 Chr.2: 24037650-24037650 on Build GRCh38
rs750056319 Chr.2: 24037720-24037720 on Build GRCh38
rs200402344 Chr.2: 24037791-24037791 on Build GRCh38
rs773834149 Chr.2: 24037886-24037886 on Build GRCh38
rs114984909 Chr.2: 24037524-24037524 on Build GRCh38
rs765004509 Chr.2: 24037718-24037718 on Build GRCh38
rs996871058 Chr.2: 24037623-24037623 on Build GRCh38
rs777764829 Chr.2: 24037747-24037747 on Build GRCh38
rs149277361 Chr.2: 24037839-24037839 on Build GRCh38
rs998945870 Chr.2: 24037543-24037543 on Build GRCh38
rs1051188302 Chr.2: 24037625-24037625 on Build GRCh38
rs747512895 Chr.2: 24037763-24037763 on Build GRCh38
rs149090512 Chr.2: 24038045-24038045 on Build GRCh38
rs1055013606 Chr.2: 24037939-24037939 on Build GRCh38
rs917547586 Chr.2: 24037797-24037797 on Build GRCh38
rs747426487 Chr.2: 24037848-24037848 on Build GRCh38
rs769487963 Chr.2: 24037873-24037873 on Build GRCh38
rs1023183784 Chr.2: 24038005-24038005 on Build GRCh38
rs775552056 Chr.2: 24037904-24037904 on Build GRCh38
rs778374387 Chr.2: 24037968-24037968 on Build GRCh38
rs756027777 Chr.2: 24037607-24037607 on Build GRCh38
rs749964768 Chr.2: 24037608-24037608 on Build GRCh38
rs934852576 Chr.2: 24037860-24037860 on Build GRCh38
rs781398981 Chr.2: 24037946-24037946 on Build GRCh38
rs750482403 Chr.2: 24037926-24037926 on Build GRCh38
rs758426886 Chr.2: 24037938-24037938 on Build GRCh38
rs763024905 Chr.2: 24038023-24038023 on Build GRCh38
rs934033904 Chr.2: 24037616-24037616 on Build GRCh38
rs756665495 Chr.2: 24037955-24037955 on Build GRCh38
rs748995402 Chr.2: 24037877-24037877 on Build GRCh38
rs1033127827 Chr.2: 24037570-24037570 on Build GRCh38
rs779108340 Chr.2: 24037754-24037754 on Build GRCh38
rs746259142 Chr.2: 24037679-24037679 on Build GRCh38
rs891081722 Chr.2: 24037982-24037982 on Build GRCh38
rs778019108 Chr.2: 24037886-24037903 on Build GRCh38
rs748838053 Chr.2: 24037985-24037985 on Build GRCh38
rs1026024389 Chr.2: 24037769-24037769 on Build GRCh38
rs774504322 Chr.2: 24037654-24037654 on Build GRCh38
rs767156449 Chr.2: 24037898-24037898 on Build GRCh38
rs764044769 Chr.2: 24038013-24038013 on Build GRCh38
rs893220813 Chr.2: 24037575-24037575 on Build GRCh38
rs374870648 Chr.2: 24037908-24037908 on Build GRCh38
rs756155854 Chr.2: 24037697-24037700 on Build GRCh38
rs899850148 Chr.2: 24037568-24037568 on Build GRCh38
rs768617221 Chr.2: 24037753-24037753 on Build GRCh38
rs775118738 Chr.2: 24038008-24038008 on Build GRCh38
rs771967127 Chr.2: 24037996-24037996 on Build GRCh38
rs140144798 Chr.2: 24037871-24037871 on Build GRCh38
rs767517933 Chr.2: 24037831-24037831 on Build GRCh38
rs772480257 Chr.2: 24037759-24037759 on Build GRCh38
rs769188247 Chr.2: 24037772-24037772 on Build GRCh38
rs1040088870 Chr.2: 24037556-24037556 on Build GRCh38
rs770729263 Chr.2: 24037879-24037879 on Build GRCh38
rs751347149 Chr.2: 24037795-24037795 on Build GRCh38
rs921231266 Chr.2: 24037863-24037863 on Build GRCh38
rs537197051 Chr.2: 24037942-24037942 on Build GRCh38
rs368866309 Chr.2: 24037847-24037847 on Build GRCh38
rs754911589 Chr.2: 24037745-24037745 on Build GRCh38
rs138431832 Chr.2: 24037778-24037778 on Build GRCh38
rs200971514 Chr.2: 24037777-24037777 on Build GRCh38
rs779887230 Chr.2: 24037627-24037627 on Build GRCh38
rs773013311 Chr.2: 24037783-24037783 on Build GRCh38
rs746029896 Chr.2: 24037755-24037755 on Build GRCh38
rs147244110 Chr.2: 24037990-24037990 on Build GRCh38
rs1002493346 Chr.2: 24038039-24038039 on Build GRCh38
rs115270382 Chr.2: 24037992-24037992 on Build GRCh38
rs772354340 Chr.2: 24037681-24037681 on Build GRCh38
rs369415808 Chr.2: 24037940-24037940 on Build GRCh38
rs757340328 Chr.2: 24037835-24037835 on Build GRCh38
rs375161885 Chr.2: 24037626-24037626 on Build GRCh38
rs776514314 Chr.2: 24038018-24038018 on Build GRCh38
rs769364997 Chr.2: 24037779-24037779 on Build GRCh38
rs752845901 Chr.2: 24037665-24037667 on Build GRCh38
rs766221326 Chr.2: 24037725-24037725 on Build GRCh38
rs774062500 Chr.2: 24037991-24037991 on Build GRCh38
rs367867532 Chr.2: 24037895-24037895 on Build GRCh38
rs776179142 Chr.2: 24037715-24037715 on Build GRCh38

View Full Product Details