Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 3670934-3671315 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALLC
Entrez Gene ID
55821
Gene Name
allantoicase
Gene Aliases
ALC
UniGene
Hs.97899
Gene Chromosome Location
Chr.2: 3645368-3702671 on Build GRCh38
Transcripts
NM_018436.3 XM_011510370.2 XM_017004495.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1742745 c.24A>T p.V8V Chr.2: 3671181-3671181 on Build GRCh38
5474482 c.21C>A p.S7S Chr.2: 3671178-3671178 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs867754274 Chr.2: 3671186-3671186 on Build GRCh38
rs773501884 Chr.2: 3671135-3671135 on Build GRCh38
rs776275565 Chr.2: 3671207-3671207 on Build GRCh38
rs558798889 Chr.2: 3670992-3670992 on Build GRCh38
rs368868282 Chr.2: 3671131-3671131 on Build GRCh38
rs745662430 Chr.2: 3671114-3671114 on Build GRCh38
rs1016998522 Chr.2: 3671163-3671163 on Build GRCh38
rs773732874 Chr.2: 3671236-3671236 on Build GRCh38
rs541622966 Chr.2: 3671126-3671126 on Build GRCh38
rs1011159569 Chr.2: 3671073-3671073 on Build GRCh38
rs892662135 Chr.2: 3671072-3671072 on Build GRCh38
rs146521098 Chr.2: 3671133-3671133 on Build GRCh38
rs962917038 Chr.2: 3671022-3671022 on Build GRCh38
rs758297888 Chr.2: 3671183-3671183 on Build GRCh38
rs59726279 Chr.2: 3671065-3671065 on Build GRCh38
rs960792066 Chr.2: 3671109-3671109 on Build GRCh38
rs761121178 Chr.2: 3671103-3671103 on Build GRCh38
rs946831682 Chr.2: 3671257-3671257 on Build GRCh38
rs757096252 Chr.2: 3671110-3671110 on Build GRCh38
rs763324177 Chr.2: 3671142-3671142 on Build GRCh38
rs1002880277 Chr.2: 3671282-3671282 on Build GRCh38
rs942761399 Chr.2: 3671262-3671262 on Build GRCh38
rs373415935 Chr.2: 3671178-3671178 on Build GRCh38
rs914063288 Chr.2: 3671255-3671255 on Build GRCh38
rs753814303 Chr.2: 3671168-3671168 on Build GRCh38
rs911073877 Chr.2: 3671239-3671239 on Build GRCh38
rs975746309 Chr.2: 3671169-3671169 on Build GRCh38
rs1044035483 Chr.2: 3671074-3671074 on Build GRCh38
rs13409104 Chr.2: 3671102-3671102 on Build GRCh38
rs1040448491 Chr.2: 3671038-3671038 on Build GRCh38
rs984597619 Chr.2: 3671145-3671145 on Build GRCh38
rs771791796 Chr.2: 3671115-3671115 on Build GRCh38
rs200344138 Chr.2: 3671134-3671134 on Build GRCh38
rs369154044 Chr.2: 3671176-3671176 on Build GRCh38
rs564050552 Chr.2: 3671233-3671233 on Build GRCh38
rs570267579 Chr.2: 3671089-3671089 on Build GRCh38
rs933866560 Chr.2: 3671060-3671060 on Build GRCh38
rs910212697 Chr.2: 3671012-3671012 on Build GRCh38
rs990444601 Chr.2: 3670989-3670989 on Build GRCh38
rs746720707 Chr.2: 3671193-3671193 on Build GRCh38
rs576308283 Chr.2: 3671156-3671156 on Build GRCh38
rs573131939 Chr.2: 3670959-3670959 on Build GRCh38
rs745607491 Chr.2: 3671179-3671179 on Build GRCh38
rs572139191 Chr.2: 3671279-3671279 on Build GRCh38
rs1054576476 Chr.2: 3671285-3671285 on Build GRCh38
rs60644898 Chr.2: 3671056-3671056 on Build GRCh38
rs768034036 Chr.2: 3671205-3671205 on Build GRCh38
rs111876121 Chr.2: 3671011-3671011 on Build GRCh38
rs767685543 Chr.2: 3671165-3671165 on Build GRCh38
rs747665598 Chr.2: 3671225-3671225 on Build GRCh38
rs779539122 Chr.2: 3671192-3671192 on Build GRCh38
rs546978029 Chr.2: 3671232-3671232 on Build GRCh38
rs951947743 Chr.2: 3671002-3671002 on Build GRCh38
rs201176267 Chr.2: 3671271-3671271 on Build GRCh38
rs943005816 Chr.2: 3671018-3671018 on Build GRCh38
rs375517332 Chr.2: 3671123-3671123 on Build GRCh38
rs961075703 Chr.2: 3670971-3670971 on Build GRCh38
rs577140234 Chr.2: 3671037-3671037 on Build GRCh38
rs905418108 Chr.2: 3671280-3671280 on Build GRCh38
rs757050465 Chr.2: 3671152-3671152 on Build GRCh38
rs377010333 Chr.2: 3671159-3671159 on Build GRCh38
rs964617242 Chr.2: 3671161-3671161 on Build GRCh38
rs919115878 Chr.2: 3670975-3670975 on Build GRCh38
rs140963429 Chr.2: 3671292-3671292 on Build GRCh38
rs866663518 Chr.2: 3671185-3671185 on Build GRCh38
rs923022183 Chr.2: 3671055-3671055 on Build GRCh38
rs140543052 Chr.2: 3670972-3670972 on Build GRCh38
rs776966958 Chr.2: 3671139-3671139 on Build GRCh38
rs778918019 Chr.2: 3671111-3671111 on Build GRCh38
rs942792386 Chr.2: 3671090-3671090 on Build GRCh38

View Full Product Details