Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 57584031-57584630 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARF4
Entrez Gene ID
378
Gene Name
ADP ribosylation factor 4
Gene Aliases
ARF2
UniGene
Hs.652183
Gene Chromosome Location
Chr.3: 57571363-57597488 on Build GRCh38
Transcripts
NM_001660.3 NM_001322176.1

Gene Symbol
PDE12
Entrez Gene ID
201626
Gene Name
phosphodiesterase 12
Gene Aliases
2'-PDE, 2-PDE
UniGene
Hs.572993
Gene Chromosome Location
Chr.3: 57556247-57656480 on Build GRCh38
Transcripts
NM_001660.3 NM_001322176.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5627660 c.104A>G p.Y35C Chr.3: 57584428-57584428 on Build GRCh38
6427175 c.127G>T p.V43F Chr.3: 57584405-57584405 on Build GRCh38
3596712 c.76G>A p.D26N Chr.3: 57584456-57584456 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs945491746 Chr.3: 57584328-57584328 on Build GRCh38
rs545291448 Chr.3: 57584257-57584257 on Build GRCh38
rs373000414 Chr.3: 57584363-57584363 on Build GRCh38
rs554630232 Chr.3: 57584116-57584116 on Build GRCh38
rs143859407 Chr.3: 57584520-57584520 on Build GRCh38
rs772124941 Chr.3: 57584473-57584473 on Build GRCh38
rs936279687 Chr.3: 57584092-57584092 on Build GRCh38
rs918122110 Chr.3: 57584309-57584309 on Build GRCh38
rs758832713 Chr.3: 57584365-57584365 on Build GRCh38
rs767425593 Chr.3: 57584055-57584055 on Build GRCh38
rs945145848 Chr.3: 57584072-57584072 on Build GRCh38
rs926921307 Chr.3: 57584106-57584106 on Build GRCh38
rs776740900 Chr.3: 57584488-57584488 on Build GRCh38
rs562854111 Chr.3: 57584251-57584251 on Build GRCh38
rs549957320 Chr.3: 57584534-57584534 on Build GRCh38
rs574177230 Chr.3: 57584155-57584155 on Build GRCh38
rs369343914 Chr.3: 57584474-57584474 on Build GRCh38
rs993552083 Chr.3: 57584232-57584232 on Build GRCh38
rs766157937 Chr.3: 57584480-57584480 on Build GRCh38
rs569864431 Chr.3: 57584554-57584554 on Build GRCh38
rs1056364232 Chr.3: 57584176-57584176 on Build GRCh38
rs749700809 Chr.3: 57584467-57584469 on Build GRCh38
rs747316487 Chr.3: 57584388-57584388 on Build GRCh38
rs888273088 Chr.3: 57584258-57584258 on Build GRCh38
rs753990618 Chr.3: 57584069-57584069 on Build GRCh38
rs368936399 Chr.3: 57584359-57584359 on Build GRCh38
rs527669778 Chr.3: 57584348-57584348 on Build GRCh38
rs748926137 Chr.3: 57584410-57584410 on Build GRCh38
rs1042150526 Chr.3: 57584086-57584086 on Build GRCh38
rs755833114 Chr.3: 57584396-57584396 on Build GRCh38
rs779606925 Chr.3: 57584406-57584406 on Build GRCh38
rs985166899 Chr.3: 57584267-57584267 on Build GRCh38
rs992419211 Chr.3: 57584303-57584303 on Build GRCh38
rs6763507 Chr.3: 57584215-57584215 on Build GRCh38
rs757806290 Chr.3: 57584477-57584479 on Build GRCh38
rs748165241 Chr.3: 57584462-57584462 on Build GRCh38
rs981429985 Chr.3: 57584088-57584088 on Build GRCh38
rs1049886628 Chr.3: 57584596-57584596 on Build GRCh38
rs538976103 Chr.3: 57584556-57584556 on Build GRCh38
rs1018590262 Chr.3: 57584284-57584284 on Build GRCh38
rs903526555 Chr.3: 57584091-57584091 on Build GRCh38
rs915370671 Chr.3: 57584061-57584061 on Build GRCh38
rs188811931 Chr.3: 57584351-57584351 on Build GRCh38
rs202163662 Chr.3: 57584369-57584369 on Build GRCh38
rs1049681565 Chr.3: 57584102-57584102 on Build GRCh38
rs541710360 Chr.3: 57584149-57584149 on Build GRCh38
rs773931742 Chr.3: 57584439-57584439 on Build GRCh38
rs992373982 Chr.3: 57584595-57584595 on Build GRCh38
rs768064782 Chr.3: 57584431-57584431 on Build GRCh38
rs952065475 Chr.3: 57584265-57584265 on Build GRCh38
rs115703281 Chr.3: 57584558-57584558 on Build GRCh38
rs760354843 Chr.3: 57584478-57584478 on Build GRCh38
rs1008129194 Chr.3: 57584126-57584126 on Build GRCh38
rs565571096 Chr.3: 57584571-57584571 on Build GRCh38
rs929354179 Chr.3: 57584242-57584242 on Build GRCh38
rs771791127 Chr.3: 57584108-57584108 on Build GRCh38
rs540803118 Chr.3: 57584060-57584060 on Build GRCh38
rs773138173 Chr.3: 57584059-57584059 on Build GRCh38
rs998523861 Chr.3: 57584319-57584319 on Build GRCh38
rs1033672313 Chr.3: 57584286-57584286 on Build GRCh38
rs117242785 Chr.3: 57584302-57584302 on Build GRCh38
rs140177104 Chr.3: 57584254-57584254 on Build GRCh38
rs1029307685 Chr.3: 57584349-57584349 on Build GRCh38

View Full Product Details