Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 58653471-58653904 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM3D
Entrez Gene ID
131177
Gene Name
family with sequence similarity 3 member D
Gene Aliases
EF7, OIT1
UniGene
Hs.61265
Gene Chromosome Location
Chr.3: 58633943-58666843 on Build GRCh38
Transcripts
NM_138805.2 XM_005264862.4 XM_005264863.4 XM_005264864.4 XM_006712965.3 XM_006712967.3 XM_011533348.2 XM_011533349.2 XM_011533350.2 XM_011533351.2 XM_011533352.2 XM_011533353.2 XM_011533354.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4119694 c.88A>C p.S30R Chr.3: 58653707-58653707 on Build GRCh38
1206338 c.70C>T p.R24* Chr.3: 58653725-58653725 on Build GRCh38
1206337 c.109C>T p.P37S Chr.3: 58653686-58653686 on Build GRCh38
5596688 c.27C>T p.L9L Chr.3: 58653768-58653768 on Build GRCh38
3596891 c.59G>A p.W20* Chr.3: 58653736-58653736 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781585060 Chr.3: 58653820-58653832 on Build GRCh38
rs571861350 Chr.3: 58653711-58653711 on Build GRCh38
rs140921588 Chr.3: 58653692-58653692 on Build GRCh38
rs759568298 Chr.3: 58653718-58653718 on Build GRCh38
rs756119580 Chr.3: 58653654-58653654 on Build GRCh38
rs959493701 Chr.3: 58653871-58653871 on Build GRCh38
rs763454381 Chr.3: 58653869-58653869 on Build GRCh38
rs1008272199 Chr.3: 58653570-58653570 on Build GRCh38
rs954606799 Chr.3: 58653544-58653544 on Build GRCh38
rs13097314 Chr.3: 58653691-58653691 on Build GRCh38
rs1051520378 Chr.3: 58653826-58653826 on Build GRCh38
rs959609753 Chr.3: 58653660-58653660 on Build GRCh38
rs890297316 Chr.3: 58653841-58653841 on Build GRCh38
rs749230899 Chr.3: 58653790-58653790 on Build GRCh38
rs536657345 Chr.3: 58653779-58653779 on Build GRCh38
rs778301723 Chr.3: 58653773-58653773 on Build GRCh38
rs771609102 Chr.3: 58653785-58653785 on Build GRCh38
rs1007393336 Chr.3: 58653864-58653864 on Build GRCh38
rs372221279 Chr.3: 58653622-58653622 on Build GRCh38
rs556235777 Chr.3: 58653851-58653851 on Build GRCh38
rs373992269 Chr.3: 58653744-58653744 on Build GRCh38
rs375435708 Chr.3: 58653526-58653526 on Build GRCh38
rs184205265 Chr.3: 58653621-58653621 on Build GRCh38
rs955487113 Chr.3: 58653549-58653549 on Build GRCh38
rs1007445775 Chr.3: 58653553-58653553 on Build GRCh38
rs929069217 Chr.3: 58653815-58653815 on Build GRCh38
rs774015496 Chr.3: 58653687-58653687 on Build GRCh38
rs754489900 Chr.3: 58653772-58653772 on Build GRCh38
rs199708473 Chr.3: 58653646-58653646 on Build GRCh38
rs752889941 Chr.3: 58653624-58653624 on Build GRCh38
rs775429985 Chr.3: 58653828-58653828 on Build GRCh38
rs867454790 Chr.3: 58653781-58653781 on Build GRCh38
rs1030642572 Chr.3: 58653500-58653500 on Build GRCh38
rs1017526605 Chr.3: 58653604-58653604 on Build GRCh38
rs201543938 Chr.3: 58653683-58653683 on Build GRCh38
rs540712972 Chr.3: 58653506-58653506 on Build GRCh38
rs202112915 Chr.3: 58653653-58653653 on Build GRCh38
rs547933815 Chr.3: 58653742-58653742 on Build GRCh38
rs1028277428 Chr.3: 58653504-58653504 on Build GRCh38
rs777162177 Chr.3: 58653716-58653716 on Build GRCh38
rs878883392 Chr.3: 58653836-58653836 on Build GRCh38
rs531865043 Chr.3: 58653682-58653682 on Build GRCh38
rs767094622 Chr.3: 58653634-58653634 on Build GRCh38
rs373040244 Chr.3: 58653794-58653794 on Build GRCh38
rs780201218 Chr.3: 58653669-58653669 on Build GRCh38
rs769843494 Chr.3: 58653827-58653827 on Build GRCh38
rs370181272 Chr.3: 58653701-58653701 on Build GRCh38
rs201578543 Chr.3: 58653642-58653642 on Build GRCh38
rs777070117 Chr.3: 58653825-58653825 on Build GRCh38
rs780113389 Chr.3: 58653758-58653758 on Build GRCh38
rs567619603 Chr.3: 58653748-58653748 on Build GRCh38
rs1020997969 Chr.3: 58653573-58653573 on Build GRCh38
rs993371130 Chr.3: 58653493-58653493 on Build GRCh38
rs765250738 Chr.3: 58653724-58653724 on Build GRCh38
rs761384923 Chr.3: 58653546-58653546 on Build GRCh38
rs35045921 Chr.3: 58653738-58653738 on Build GRCh38
rs151211035 Chr.3: 58653690-58653690 on Build GRCh38
rs1051595730 Chr.3: 58653804-58653804 on Build GRCh38
rs925553942 Chr.3: 58653643-58653643 on Build GRCh38

View Full Product Details