Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 54569697-54570225 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACNA2D3
Entrez Gene ID
55799
Gene Name
calcium voltage-gated channel auxiliary subunit alpha2delta 3
Gene Aliases
HSA272268
UniGene
Hs.656687
Gene Chromosome Location
Chr.3: 54122594-55074557 on Build GRCh38
Transcripts
NM_018398.2 XM_005265318.2 XM_011533946.2 XM_011533947.2 XM_011533948.2 XM_011533949.2 XM_011533950.2 XM_011533951.2 XM_011533952.2 XM_011533953.2 XM_017006850.1 XM_017006851.1 XM_017006852.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
50699 c.754A>T p.T252S Chr.3: 54569970-54569970 on Build GRCh38
3916456 c.839C>T p.S280L Chr.3: 54570055-54570055 on Build GRCh38
3916455 c.688G>A p.E230K Chr.3: 54569806-54569806 on Build GRCh38
290303 c.729C>T p.N243N Chr.3: 54569847-54569847 on Build GRCh38
1047363 c.868G>T p.D290Y Chr.3: 54570084-54570084 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780215990 Chr.3: 54570024-54570024 on Build GRCh38
rs761102907 Chr.3: 54570055-54570055 on Build GRCh38
rs555199325 Chr.3: 54569849-54569849 on Build GRCh38
rs774768835 Chr.3: 54569819-54569819 on Build GRCh38
rs370687758 Chr.3: 54570116-54570116 on Build GRCh38
rs778781450 Chr.3: 54570050-54570050 on Build GRCh38
rs941677767 Chr.3: 54569742-54569742 on Build GRCh38
rs544975041 Chr.3: 54569779-54569779 on Build GRCh38
rs781216988 Chr.3: 54569989-54569989 on Build GRCh38
rs753647554 Chr.3: 54570128-54570128 on Build GRCh38
rs763051376 Chr.3: 54569810-54569810 on Build GRCh38
rs752449908 Chr.3: 54569977-54569977 on Build GRCh38
rs564710377 Chr.3: 54569944-54569944 on Build GRCh38
rs554525616 Chr.3: 54570181-54570181 on Build GRCh38
rs753517872 Chr.3: 54569831-54569831 on Build GRCh38
rs768762688 Chr.3: 54569907-54569907 on Build GRCh38
rs956246816 Chr.3: 54569899-54569899 on Build GRCh38
rs752219073 Chr.3: 54569836-54569836 on Build GRCh38
rs377267211 Chr.3: 54569987-54569987 on Build GRCh38
rs566029082 Chr.3: 54570125-54570125 on Build GRCh38
rs377346997 Chr.3: 54570111-54570111 on Build GRCh38
rs2251788 Chr.3: 54570161-54570161 on Build GRCh38
rs778802466 Chr.3: 54570036-54570036 on Build GRCh38
rs374420641 Chr.3: 54570094-54570094 on Build GRCh38
rs757954461 Chr.3: 54569978-54569978 on Build GRCh38
rs770683091 Chr.3: 54569750-54569750 on Build GRCh38
rs771464959 Chr.3: 54570069-54570069 on Build GRCh38
rs768082736 Chr.3: 54569930-54569930 on Build GRCh38
rs375997429 Chr.3: 54570160-54570160 on Build GRCh38
rs776905893 Chr.3: 54569774-54569774 on Build GRCh38
rs749577299 Chr.3: 54570027-54570027 on Build GRCh38
rs576155670 Chr.3: 54569727-54569727 on Build GRCh38
rs916530780 Chr.3: 54570202-54570202 on Build GRCh38
rs2359855 Chr.3: 54570002-54570002 on Build GRCh38
rs755951660 Chr.3: 54570003-54570003 on Build GRCh38
rs753020399 Chr.3: 54569718-54569718 on Build GRCh38
rs267599905 Chr.3: 54569844-54569844 on Build GRCh38
rs375873375 Chr.3: 54569916-54569916 on Build GRCh38
rs868035069 Chr.3: 54570114-54570114 on Build GRCh38
rs199884248 Chr.3: 54569984-54569984 on Build GRCh38
rs774915735 Chr.3: 54569910-54569910 on Build GRCh38
rs779227226 Chr.3: 54570148-54570148 on Build GRCh38
rs748132030 Chr.3: 54570151-54570151 on Build GRCh38
rs775712611 Chr.3: 54569886-54569886 on Build GRCh38
rs549815783 Chr.3: 54570028-54570028 on Build GRCh38
rs562712182 Chr.3: 54570134-54570134 on Build GRCh38
rs532692490 Chr.3: 54570096-54570096 on Build GRCh38
rs767838008 Chr.3: 54569824-54569824 on Build GRCh38
rs923437886 Chr.3: 54570041-54570041 on Build GRCh38
rs746962852 Chr.3: 54569862-54569862 on Build GRCh38
rs371006961 Chr.3: 54570070-54570070 on Build GRCh38
rs78831319 Chr.3: 54569882-54569882 on Build GRCh38
rs376472819 Chr.3: 54569835-54569835 on Build GRCh38
rs372114533 Chr.3: 54569796-54569796 on Build GRCh38
rs183970474 Chr.3: 54569894-54569894 on Build GRCh38
rs750989660 Chr.3: 54570109-54570109 on Build GRCh38
rs772141915 Chr.3: 54570135-54570135 on Build GRCh38
rs137869064 Chr.3: 54569866-54569866 on Build GRCh38
rs374837722 Chr.3: 54570144-54570144 on Build GRCh38
rs370946415 Chr.3: 54570119-54570119 on Build GRCh38
rs767334425 Chr.3: 54570115-54570115 on Build GRCh38
rs540193185 Chr.3: 54569934-54569934 on Build GRCh38
rs764535638 Chr.3: 54569817-54569817 on Build GRCh38
rs570177370 Chr.3: 54570044-54570044 on Build GRCh38
rs754620838 Chr.3: 54569834-54569834 on Build GRCh38
rs780790446 Chr.3: 54569880-54569880 on Build GRCh38
rs764938897 Chr.3: 54569949-54569949 on Build GRCh38
rs1028375232 Chr.3: 54569720-54569720 on Build GRCh38
rs1037610671 Chr.3: 54569777-54569777 on Build GRCh38
rs773382247 Chr.3: 54569941-54569941 on Build GRCh38
rs748264157 Chr.3: 54570038-54570038 on Build GRCh38
rs1007414748 Chr.3: 54570121-54570121 on Build GRCh38
rs772419843 Chr.3: 54570040-54570040 on Build GRCh38
rs568406396 Chr.3: 54570188-54570188 on Build GRCh38
rs992539234 Chr.3: 54569883-54569883 on Build GRCh38
rs372465588 Chr.3: 54569985-54569985 on Build GRCh38

View Full Product Details