Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 73766886-73767411 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUSP11
Entrez Gene ID
8446
Gene Name
dual specificity phosphatase 11
Gene Aliases
PIR1
UniGene
Hs.14611
Gene Chromosome Location
Chr.2: 73762184-73780157 on Build GRCh38
Transcripts
NM_003584.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3668991 c.557G>A p.R186Q Chr.2: 73766888-73766888 on Build GRCh38
7177266 c.505G>A p.D169N Chr.2: 73767197-73767197 on Build GRCh38
4431849 c.687C>G p.F229L Chr.2: 73766899-73766899 on Build GRCh38
4442117 c.698G>C p.R233P Chr.2: 73766888-73766888 on Build GRCh38
4095666 c.658G>A p.V220M Chr.2: 73767185-73767185 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770852987 Chr.2: 73766923-73766923 on Build GRCh38
rs775514973 Chr.2: 73766929-73766929 on Build GRCh38
rs751948878 Chr.2: 73767256-73767256 on Build GRCh38
rs201994650 Chr.2: 73767156-73767156 on Build GRCh38
rs139965270 Chr.2: 73767018-73767018 on Build GRCh38
rs1019942389 Chr.2: 73767352-73767352 on Build GRCh38
rs1046388639 Chr.2: 73766957-73766957 on Build GRCh38
rs2567599 Chr.2: 73766947-73766947 on Build GRCh38
rs747146433 Chr.2: 73766921-73766921 on Build GRCh38
rs758807109 Chr.2: 73767244-73767244 on Build GRCh38
rs373870876 Chr.2: 73766936-73766936 on Build GRCh38
rs1017895281 Chr.2: 73767011-73767011 on Build GRCh38
rs920643981 Chr.2: 73767351-73767351 on Build GRCh38
rs533793318 Chr.2: 73767155-73767155 on Build GRCh38
rs549718175 Chr.2: 73767029-73767029 on Build GRCh38
rs976541524 Chr.2: 73767016-73767016 on Build GRCh38
rs571327243 Chr.2: 73767042-73767042 on Build GRCh38
rs899269022 Chr.2: 73767053-73767053 on Build GRCh38
rs755515518 Chr.2: 73767134-73767134 on Build GRCh38
rs974398127 Chr.2: 73767114-73767114 on Build GRCh38
rs778199022 Chr.2: 73767328-73767328 on Build GRCh38
rs770070348 Chr.2: 73766938-73766938 on Build GRCh38
rs774330083 Chr.2: 73766937-73766937 on Build GRCh38
rs764428840 Chr.2: 73767253-73767253 on Build GRCh38
rs144123904 Chr.2: 73767185-73767185 on Build GRCh38
rs967572842 Chr.2: 73767363-73767363 on Build GRCh38
rs1009603418 Chr.2: 73767092-73767093 on Build GRCh38
rs763086609 Chr.2: 73767249-73767249 on Build GRCh38
rs200463622 Chr.2: 73767229-73767229 on Build GRCh38
rs753138604 Chr.2: 73767243-73767243 on Build GRCh38
rs949343170 Chr.2: 73767145-73767145 on Build GRCh38
rs774389362 Chr.2: 73767204-73767204 on Build GRCh38
rs544329640 Chr.2: 73767379-73767379 on Build GRCh38
rs565101040 Chr.2: 73767291-73767291 on Build GRCh38
rs773046905 Chr.2: 73767230-73767230 on Build GRCh38
rs967135226 Chr.2: 73767056-73767056 on Build GRCh38
rs900864052 Chr.2: 73767304-73767304 on Build GRCh38
rs772326055 Chr.2: 73766913-73766913 on Build GRCh38
rs372338986 Chr.2: 73767060-73767060 on Build GRCh38
rs557803282 Chr.2: 73767091-73767091 on Build GRCh38
rs777812114 Chr.2: 73766918-73766918 on Build GRCh38
rs776164591 Chr.2: 73767240-73767240 on Build GRCh38
rs766021045 Chr.2: 73767238-73767238 on Build GRCh38
rs553990085 Chr.2: 73766970-73766970 on Build GRCh38
rs773367798 Chr.2: 73766939-73766957 on Build GRCh38
rs747146824 Chr.2: 73767166-73767166 on Build GRCh38
rs772671081 Chr.2: 73766955-73766955 on Build GRCh38
rs760245494 Chr.2: 73767113-73767113 on Build GRCh38
rs1018597929 Chr.2: 73767094-73767095 on Build GRCh38
rs747960782 Chr.2: 73767214-73767214 on Build GRCh38
rs566639432 Chr.2: 73767099-73767099 on Build GRCh38
rs930750584 Chr.2: 73767361-73767361 on Build GRCh38
rs1006769857 Chr.2: 73767289-73767289 on Build GRCh38
rs376076420 Chr.2: 73767186-73767186 on Build GRCh38
rs1020429718 Chr.2: 73767050-73767050 on Build GRCh38
rs567459635 Chr.2: 73767012-73767012 on Build GRCh38
rs373082460 Chr.2: 73767139-73767139 on Build GRCh38
rs758654024 Chr.2: 73767143-73767143 on Build GRCh38
rs906552333 Chr.2: 73766972-73766972 on Build GRCh38
rs778063146 Chr.2: 73767147-73767147 on Build GRCh38
rs1047888023 Chr.2: 73767378-73767378 on Build GRCh38
rs761679490 Chr.2: 73766943-73766943 on Build GRCh38
rs201273120 Chr.2: 73767242-73767242 on Build GRCh38
rs763042301 Chr.2: 73766930-73766930 on Build GRCh38
rs991036776 Chr.2: 73767105-73767105 on Build GRCh38
rs573739709 Chr.2: 73767303-73767303 on Build GRCh38
rs971322076 Chr.2: 73767273-73767273 on Build GRCh38
rs998281894 Chr.2: 73767336-73767336 on Build GRCh38
rs753339056 Chr.2: 73767126-73767126 on Build GRCh38
rs1055131886 Chr.2: 73767315-73767315 on Build GRCh38
rs181544719 Chr.2: 73767074-73767074 on Build GRCh38
rs964316960 Chr.2: 73767112-73767112 on Build GRCh38
rs765862553 Chr.2: 73767124-73767124 on Build GRCh38
rs757492914 Chr.2: 73767181-73767181 on Build GRCh38
rs964980701 Chr.2: 73767015-73767015 on Build GRCh38
rs760307760 Chr.2: 73767234-73767234 on Build GRCh38
rs768764291 Chr.2: 73767196-73767196 on Build GRCh38

View Full Product Details