Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 165749515-165750090 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GALNT3
Entrez Gene ID
2591
Gene Name
polypeptide N-acetylgalactosaminyltransferase 3
Gene Aliases
GalNAc-T3, HFTC, HHS
UniGene
Hs.170986
Gene Chromosome Location
Chr.2: 165747803-165796352 on Build GRCh38
Transcripts
NM_004482.3 XM_005246449.1 XM_011510929.1 XM_017003770.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6669095 c.1704C>T p.L568L Chr.2: 165749817-165749817 on Build GRCh38
4086268 c.1717G>A p.A573T Chr.2: 165749804-165749804 on Build GRCh38
476144 c.1689T>C p.H563H Chr.2: 165749832-165749832 on Build GRCh38
6088042 c.1633G>A p.E545K Chr.2: 165749888-165749888 on Build GRCh38
4086269 c.1657C>T p.R553W Chr.2: 165749864-165749864 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750333904 Chr.2: 165749809-165749809 on Build GRCh38
rs534533721 Chr.2: 165749974-165749974 on Build GRCh38
rs549196682 Chr.2: 165750030-165750030 on Build GRCh38
rs775040032 Chr.2: 165749837-165749837 on Build GRCh38
rs1014973678 Chr.2: 165749608-165749608 on Build GRCh38
rs937533631 Chr.2: 165749904-165749904 on Build GRCh38
rs556498644 Chr.2: 165750047-165750047 on Build GRCh38
rs143974566 Chr.2: 165749632-165749632 on Build GRCh38
rs777834339 Chr.2: 165749817-165749817 on Build GRCh38
rs377447501 Chr.2: 165749914-165749914 on Build GRCh38
rs373125597 Chr.2: 165749897-165749897 on Build GRCh38
rs774689220 Chr.2: 165749727-165749727 on Build GRCh38
rs760159041 Chr.2: 165749842-165749842 on Build GRCh38
rs1036787279 Chr.2: 165749989-165749989 on Build GRCh38
rs548932359 Chr.2: 165749645-165749645 on Build GRCh38
rs759069735 Chr.2: 165749728-165749728 on Build GRCh38
rs763643230 Chr.2: 165749748-165749748 on Build GRCh38
rs994867188 Chr.2: 165749996-165749996 on Build GRCh38
rs35167480 Chr.2: 165749961-165749964 on Build GRCh38
rs370837483 Chr.2: 165749939-165749939 on Build GRCh38
rs895816414 Chr.2: 165749556-165749556 on Build GRCh38
rs886042853 Chr.2: 165749827-165749827 on Build GRCh38
rs373927295 Chr.2: 165749942-165749942 on Build GRCh38
rs137853087 Chr.2: 165749747-165749747 on Build GRCh38
rs781352133 Chr.2: 165749811-165749811 on Build GRCh38
rs771512636 Chr.2: 165749730-165749730 on Build GRCh38
rs571016917 Chr.2: 165749825-165749825 on Build GRCh38
rs745565747 Chr.2: 165749652-165749652 on Build GRCh38
rs375337784 Chr.2: 165749708-165749708 on Build GRCh38
rs994604367 Chr.2: 165749619-165749619 on Build GRCh38
rs763048224 Chr.2: 165749885-165749885 on Build GRCh38
rs776110662 Chr.2: 165749857-165749857 on Build GRCh38
rs765255078 Chr.2: 165749767-165749767 on Build GRCh38
rs749600839 Chr.2: 165749721-165749721 on Build GRCh38
rs540438658 Chr.2: 165749614-165749614 on Build GRCh38
rs756116505 Chr.2: 165749907-165749907 on Build GRCh38
rs1056014685 Chr.2: 165749910-165749910 on Build GRCh38
rs1027729343 Chr.2: 165750017-165750017 on Build GRCh38
rs376449936 Chr.2: 165749822-165749822 on Build GRCh38
rs766405349 Chr.2: 165749892-165749892 on Build GRCh38
rs370027205 Chr.2: 165749778-165749778 on Build GRCh38
rs529197007 Chr.2: 165749644-165749644 on Build GRCh38
rs764878326 Chr.2: 165749864-165749864 on Build GRCh38
rs986001440 Chr.2: 165749686-165749689 on Build GRCh38
rs753718380 Chr.2: 165749763-165749763 on Build GRCh38
rs775378938 Chr.2: 165749757-165749765 on Build GRCh38
rs1018828690 Chr.2: 165750059-165750059 on Build GRCh38
rs201855584 Chr.2: 165749863-165749863 on Build GRCh38
rs888097996 Chr.2: 165749594-165749594 on Build GRCh38
rs148063331 Chr.2: 165749764-165749764 on Build GRCh38
rs1005213008 Chr.2: 165750024-165750024 on Build GRCh38
rs979205397 Chr.2: 165749874-165749874 on Build GRCh38
rs768399418 Chr.2: 165749845-165749845 on Build GRCh38
rs146521644 Chr.2: 165749816-165749816 on Build GRCh38
rs755143985 Chr.2: 165749635-165749636 on Build GRCh38
rs779319452 Chr.2: 165749932-165749932 on Build GRCh38
rs945605902 Chr.2: 165749856-165749856 on Build GRCh38
rs531415480 Chr.2: 165749810-165749810 on Build GRCh38
rs181609425 Chr.2: 165750035-165750035 on Build GRCh38
rs567582522 Chr.2: 165749948-165749948 on Build GRCh38
rs770283734 Chr.2: 165749698-165749698 on Build GRCh38
rs768648366 Chr.2: 165750011-165750011 on Build GRCh38
rs11900021 Chr.2: 165749935-165749935 on Build GRCh38
rs371376082 Chr.2: 165749702-165749702 on Build GRCh38
rs757646735 Chr.2: 165749922-165749922 on Build GRCh38
rs772619399 Chr.2: 165749936-165749936 on Build GRCh38
rs1014084427 Chr.2: 165750040-165750040 on Build GRCh38
rs760388118 Chr.2: 165749738-165749738 on Build GRCh38
rs1014076262 Chr.2: 165749671-165749671 on Build GRCh38
rs771517567 Chr.2: 165749726-165749726 on Build GRCh38
rs373524164 Chr.2: 165749813-165749813 on Build GRCh38
rs966375161 Chr.2: 165750060-165750060 on Build GRCh38
rs771159363 Chr.2: 165749828-165749828 on Build GRCh38
rs377579661 Chr.2: 165749830-165749830 on Build GRCh38
rs775129956 Chr.2: 165749731-165749731 on Build GRCh38
rs1014675213 Chr.2: 165749608-165749610 on Build GRCh38
rs533204322 Chr.2: 165749600-165749600 on Build GRCh38
rs534078627 Chr.2: 165749912-165749912 on Build GRCh38
rs370190472 Chr.2: 165749832-165749832 on Build GRCh38
rs750387449 Chr.2: 165749876-165749876 on Build GRCh38
rs888173408 Chr.2: 165749995-165749995 on Build GRCh38
rs965701373 Chr.2: 165749777-165749777 on Build GRCh38
rs267606841 Chr.2: 165749801-165749801 on Build GRCh38

View Full Product Details