Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 59326378-59326832 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DAAM1
Entrez Gene ID
23002
Gene Name
dishevelled associated activator of morphogenesis 1
Gene Aliases
-
UniGene
Hs.19156
Gene Chromosome Location
Chr.14: 59188611-59371405 on Build GRCh38
Transcripts
NM_001270520.1 NM_014992.2 XM_005267430.1 XM_005267431.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5792248 c.1275A>C p.T425T Chr.14: 59326610-59326610 on Build GRCh38
956609 c.1308A>G p.V436V Chr.14: 59326643-59326643 on Build GRCh38
6639647 c.1304T>C p.V435A Chr.14: 59326639-59326639 on Build GRCh38
4940796 c.1275A>G p.T425T Chr.14: 59326610-59326610 on Build GRCh38
5636858 c.1245G>A p.Q415Q Chr.14: 59326580-59326580 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769580191 Chr.14: 59326481-59326481 on Build GRCh38
rs200254943 Chr.14: 59326480-59326481 on Build GRCh38
rs557818452 Chr.14: 59326586-59326586 on Build GRCh38
rs944896399 Chr.14: 59326754-59326754 on Build GRCh38
rs772529022 Chr.14: 59326523-59326523 on Build GRCh38
rs779557769 Chr.14: 59326713-59326713 on Build GRCh38
rs139756687 Chr.14: 59326535-59326535 on Build GRCh38
rs765957580 Chr.14: 59326501-59326501 on Build GRCh38
rs754945270 Chr.14: 59326603-59326603 on Build GRCh38
rs772796087 Chr.14: 59326484-59326485 on Build GRCh38
rs145334981 Chr.14: 59326641-59326641 on Build GRCh38
rs192299759 Chr.14: 59326560-59326560 on Build GRCh38
rs946837282 Chr.14: 59326663-59326663 on Build GRCh38
rs553128687 Chr.14: 59326441-59326441 on Build GRCh38
rs776856269 Chr.14: 59326538-59326538 on Build GRCh38
rs1038750161 Chr.14: 59326695-59326695 on Build GRCh38
rs575281671 Chr.14: 59326532-59326532 on Build GRCh38
rs780482467 Chr.14: 59326666-59326666 on Build GRCh38
rs924136921 Chr.14: 59326672-59326672 on Build GRCh38
rs769463822 Chr.14: 59326515-59326515 on Build GRCh38
rs754857237 Chr.14: 59326469-59326469 on Build GRCh38
rs749038803 Chr.14: 59326645-59326645 on Build GRCh38
rs765384602 Chr.14: 59326555-59326555 on Build GRCh38
rs531045209 Chr.14: 59326436-59326436 on Build GRCh38
rs528040399 Chr.14: 59326671-59326671 on Build GRCh38
rs763079479 Chr.14: 59326581-59326581 on Build GRCh38
rs892053169 Chr.14: 59326409-59326409 on Build GRCh38
rs751251823 Chr.14: 59326598-59326598 on Build GRCh38
rs748611308 Chr.14: 59326763-59326763 on Build GRCh38
rs943224427 Chr.14: 59326691-59326691 on Build GRCh38
rs182045251 Chr.14: 59326809-59326809 on Build GRCh38
rs776772228 Chr.14: 59326494-59326498 on Build GRCh38
rs761945053 Chr.14: 59326545-59326545 on Build GRCh38
rs915309762 Chr.14: 59326643-59326643 on Build GRCh38
rs34486635 Chr.14: 59326686-59326686 on Build GRCh38
rs1017464433 Chr.14: 59326492-59326492 on Build GRCh38
rs745703389 Chr.14: 59326694-59326694 on Build GRCh38
rs752222616 Chr.14: 59326640-59326640 on Build GRCh38
rs777230458 Chr.14: 59326685-59326685 on Build GRCh38
rs748920761 Chr.14: 59326521-59326521 on Build GRCh38
rs150072573 Chr.14: 59326537-59326537 on Build GRCh38
rs559861041 Chr.14: 59326741-59326741 on Build GRCh38
rs768988634 Chr.14: 59326536-59326536 on Build GRCh38
rs887572661 Chr.14: 59326527-59326527 on Build GRCh38
rs574951795 Chr.14: 59326778-59326778 on Build GRCh38
rs770689660 Chr.14: 59326458-59326458 on Build GRCh38
rs970252079 Chr.14: 59326471-59326471 on Build GRCh38
rs902767299 Chr.14: 59326398-59326398 on Build GRCh38
rs756849853 Chr.14: 59326655-59326655 on Build GRCh38
rs557105973 Chr.14: 59326491-59326491 on Build GRCh38
rs769237491 Chr.14: 59326674-59326674 on Build GRCh38
rs767398359 Chr.14: 59326611-59326611 on Build GRCh38
rs919136078 Chr.14: 59326576-59326576 on Build GRCh38
rs925710817 Chr.14: 59326761-59326761 on Build GRCh38
rs189214118 Chr.14: 59326489-59326489 on Build GRCh38
rs369360632 Chr.14: 59326519-59326519 on Build GRCh38
rs568164601 Chr.14: 59326452-59326452 on Build GRCh38

View Full Product Details