Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 73689393-73689677 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAL1
Entrez Gene ID
83544
Gene Name
dynein axonemal light chain 1
Gene Aliases
C14orf168, CILD16
UniGene
Hs.271270
Gene Chromosome Location
Chr.14: 73644875-73703728 on Build GRCh38
Transcripts
NM_001201366.1 NM_031427.3 XM_017021679.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2137293 c.442G>T p.V148L Chr.14: 73689425-73689425 on Build GRCh38
1477757 c.449A>G p.N150S Chr.14: 73689432-73689432 on Build GRCh38
1587597 c.493G>T p.E165* Chr.14: 73689476-73689476 on Build GRCh38
5938344 c.427G>A p.E143K Chr.14: 73689410-73689410 on Build GRCh38
4819923 c.412G>A p.E138K Chr.14: 73689395-73689395 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs138833457 Chr.14: 73689568-73689568 on Build GRCh38
rs781170779 Chr.14: 73689483-73689483 on Build GRCh38
rs756764101 Chr.14: 73689419-73689420 on Build GRCh38
rs62004949 Chr.14: 73689613-73689613 on Build GRCh38
rs912352605 Chr.14: 73689535-73689535 on Build GRCh38
rs1037739634 Chr.14: 73689425-73689425 on Build GRCh38
rs754325250 Chr.14: 73689462-73689462 on Build GRCh38
rs972321310 Chr.14: 73689474-73689474 on Build GRCh38
rs35284335 Chr.14: 73689500-73689500 on Build GRCh38
rs181061488 Chr.14: 73689531-73689531 on Build GRCh38
rs745686826 Chr.14: 73689491-73689491 on Build GRCh38
rs73297422 Chr.14: 73689542-73689542 on Build GRCh38
rs564719742 Chr.14: 73689615-73689615 on Build GRCh38
rs387907021 Chr.14: 73689432-73689432 on Build GRCh38
rs957923125 Chr.14: 73689552-73689552 on Build GRCh38
rs72721725 Chr.14: 73689526-73689526 on Build GRCh38
rs780459682 Chr.14: 73689445-73689445 on Build GRCh38
rs572609061 Chr.14: 73689596-73689596 on Build GRCh38
rs867585871 Chr.14: 73689507-73689507 on Build GRCh38
rs372572996 Chr.14: 73689469-73689469 on Build GRCh38
rs774402568 Chr.14: 73689533-73689533 on Build GRCh38
rs966412495 Chr.14: 73689643-73689643 on Build GRCh38
rs190778454 Chr.14: 73689473-73689473 on Build GRCh38
rs150636700 Chr.14: 73689475-73689475 on Build GRCh38
rs776888880 Chr.14: 73689452-73689452 on Build GRCh38
rs770681880 Chr.14: 73689481-73689481 on Build GRCh38
rs942407437 Chr.14: 73689541-73689541 on Build GRCh38

View Full Product Details