Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 96264149-96264727 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BDKRB1
Entrez Gene ID
623
Gene Name
bradykinin receptor B1
Gene Aliases
B1BKR, B1R, BDKRB2, BKB1R, BKR1, BRADYB1
UniGene
Hs.525572
Gene Chromosome Location
Chr.14: 96256210-96264763 on Build GRCh38
Transcripts
NM_000710.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3815623 c.617T>C p.L206S Chr.14: 96264299-96264299 on Build GRCh38
6611149 c.491_492insG p.L167fs*30 Chr.14: 96264173-96264174 on Build GRCh38
6611147 c.926C>T p.P309L Chr.14: 96264608-96264608 on Build GRCh38
1640184 c.721G>T p.G241W Chr.14: 96264403-96264403 on Build GRCh38
5589807 c.636C>T p.L212L Chr.14: 96264318-96264318 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753420449 Chr.14: 96264336-96264336 on Build GRCh38
rs952768042 Chr.14: 96264301-96264301 on Build GRCh38
rs761782388 Chr.14: 96264323-96264323 on Build GRCh38
rs774873105 Chr.14: 96264469-96264469 on Build GRCh38
rs768607326 Chr.14: 96264297-96264297 on Build GRCh38
rs759325778 Chr.14: 96264465-96264465 on Build GRCh38
rs753395833 Chr.14: 96264252-96264252 on Build GRCh38
rs779585922 Chr.14: 96264175-96264175 on Build GRCh38
rs748553489 Chr.14: 96264265-96264265 on Build GRCh38
rs8004609 Chr.14: 96264632-96264632 on Build GRCh38
rs548082324 Chr.14: 96264551-96264551 on Build GRCh38
rs774234943 Chr.14: 96264315-96264315 on Build GRCh38
rs773048836 Chr.14: 96264332-96264332 on Build GRCh38
rs779487244 Chr.14: 96264204-96264204 on Build GRCh38
rs764543589 Chr.14: 96264508-96264508 on Build GRCh38
rs937276546 Chr.14: 96264535-96264535 on Build GRCh38
rs772887692 Chr.14: 96264399-96264399 on Build GRCh38
rs115980193 Chr.14: 96264556-96264556 on Build GRCh38
rs566696550 Chr.14: 96264553-96264553 on Build GRCh38
rs143432375 Chr.14: 96264663-96264663 on Build GRCh38
rs757269829 Chr.14: 96264660-96264660 on Build GRCh38
rs925794298 Chr.14: 96264617-96264617 on Build GRCh38
rs576380894 Chr.14: 96264610-96264610 on Build GRCh38
rs747328339 Chr.14: 96264371-96264371 on Build GRCh38
rs1014202764 Chr.14: 96264169-96264169 on Build GRCh38
rs754137271 Chr.14: 96264641-96264641 on Build GRCh38
rs752754542 Chr.14: 96264650-96264650 on Build GRCh38
rs988263764 Chr.14: 96264515-96264515 on Build GRCh38
rs767946457 Chr.14: 96264624-96264624 on Build GRCh38
rs61744359 Chr.14: 96264329-96264329 on Build GRCh38
rs540558937 Chr.14: 96264436-96264436 on Build GRCh38
rs140574024 Chr.14: 96264222-96264222 on Build GRCh38
rs905716111 Chr.14: 96264211-96264211 on Build GRCh38
rs769273138 Chr.14: 96264202-96264202 on Build GRCh38
rs188387870 Chr.14: 96264243-96264243 on Build GRCh38
rs145322761 Chr.14: 96264526-96264526 on Build GRCh38
rs200342765 Chr.14: 96264208-96264208 on Build GRCh38
rs927230105 Chr.14: 96264535-96264535 on Build GRCh38
rs369351922 Chr.14: 96264375-96264375 on Build GRCh38
rs150459277 Chr.14: 96264447-96264447 on Build GRCh38
rs765990196 Chr.14: 96264334-96264334 on Build GRCh38
rs979445300 Chr.14: 96264309-96264309 on Build GRCh38
rs979559676 Chr.14: 96264480-96264480 on Build GRCh38
rs753566189 Chr.14: 96264338-96264340 on Build GRCh38
rs770984603 Chr.14: 96264560-96264560 on Build GRCh38
rs757437458 Chr.14: 96264374-96264374 on Build GRCh38
rs45528332 Chr.14: 96264403-96264403 on Build GRCh38
rs752150596 Chr.14: 96264514-96264514 on Build GRCh38
rs560358743 Chr.14: 96264333-96264333 on Build GRCh38
rs773723733 Chr.14: 96264612-96264612 on Build GRCh38
rs112973857 Chr.14: 96264615-96264615 on Build GRCh38
rs750046887 Chr.14: 96264477-96264477 on Build GRCh38
rs372328639 Chr.14: 96264217-96264217 on Build GRCh38
rs769502145 Chr.14: 96264467-96264467 on Build GRCh38
rs573449408 Chr.14: 96264410-96264410 on Build GRCh38
rs762479141 Chr.14: 96264481-96264481 on Build GRCh38
rs373428829 Chr.14: 96264688-96264688 on Build GRCh38
rs143823168 Chr.14: 96264253-96264253 on Build GRCh38
rs762941065 Chr.14: 96264223-96264223 on Build GRCh38
rs146837856 Chr.14: 96264355-96264355 on Build GRCh38
rs372538907 Chr.14: 96264340-96264340 on Build GRCh38
rs76887699 Chr.14: 96264241-96264241 on Build GRCh38
rs752244721 Chr.14: 96264407-96264407 on Build GRCh38
rs756016302 Chr.14: 96264671-96264671 on Build GRCh38
rs376804785 Chr.14: 96264370-96264370 on Build GRCh38
rs2229459 Chr.14: 96264431-96264431 on Build GRCh38
rs755140763 Chr.14: 96264643-96264643 on Build GRCh38
rs943944986 Chr.14: 96264197-96264197 on Build GRCh38
rs925485029 Chr.14: 96264498-96264498 on Build GRCh38
rs375483074 Chr.14: 96264244-96264244 on Build GRCh38
rs374961307 Chr.14: 96264432-96264432 on Build GRCh38
rs868223942 Chr.14: 96264377-96264378 on Build GRCh38
rs755771607 Chr.14: 96264248-96264248 on Build GRCh38
rs757606075 Chr.14: 96264426-96264426 on Build GRCh38
rs371690606 Chr.14: 96264376-96264376 on Build GRCh38
rs758922069 Chr.14: 96264365-96264365 on Build GRCh38
rs368729494 Chr.14: 96264177-96264177 on Build GRCh38
rs573131210 Chr.14: 96264672-96264672 on Build GRCh38
rs778510396 Chr.14: 96264451-96264452 on Build GRCh38
rs201208442 Chr.14: 96264448-96264448 on Build GRCh38
rs550229352 Chr.14: 96264521-96264521 on Build GRCh38
rs779513626 Chr.14: 96264264-96264264 on Build GRCh38
rs112987981 Chr.14: 96264294-96264294 on Build GRCh38
rs113666900 Chr.14: 96264409-96264409 on Build GRCh38
rs897385154 Chr.14: 96264430-96264430 on Build GRCh38
rs145002930 Chr.14: 96264209-96264209 on Build GRCh38
rs147649798 Chr.14: 96264214-96264214 on Build GRCh38
rs781224046 Chr.14: 96264661-96264661 on Build GRCh38
rs749323359 Chr.14: 96264442-96264442 on Build GRCh38
rs202216190 Chr.14: 96264390-96264390 on Build GRCh38
rs143735378 Chr.14: 96264402-96264402 on Build GRCh38
rs757505732 Chr.14: 96264337-96264337 on Build GRCh38
rs368279365 Chr.14: 96264272-96264272 on Build GRCh38
rs557384669 Chr.14: 96264619-96264619 on Build GRCh38
rs771176101 Chr.14: 96264277-96264277 on Build GRCh38
rs1045664666 Chr.14: 96264362-96264362 on Build GRCh38
rs756835528 Chr.14: 96264437-96264439 on Build GRCh38
rs897431721 Chr.14: 96264687-96264687 on Build GRCh38
rs763792076 Chr.14: 96264191-96264192 on Build GRCh38
rs762484208 Chr.14: 96264589-96264589 on Build GRCh38
rs781508498 Chr.14: 96264427-96264427 on Build GRCh38
rs566659692 Chr.14: 96264266-96264266 on Build GRCh38
rs142201911 Chr.14: 96264444-96264444 on Build GRCh38
rs571639374 Chr.14: 96264583-96264583 on Build GRCh38
rs965789691 Chr.14: 96264513-96264513 on Build GRCh38
rs766620117 Chr.14: 96264633-96264633 on Build GRCh38
rs557512070 Chr.14: 96264386-96264386 on Build GRCh38
rs868023682 Chr.14: 96264279-96264279 on Build GRCh38
rs780193100 Chr.14: 96264527-96264527 on Build GRCh38
rs760605422 Chr.14: 96264174-96264174 on Build GRCh38
rs764894215 Chr.14: 96264406-96264406 on Build GRCh38
rs775171266 Chr.14: 96264576-96264576 on Build GRCh38
rs765863931 Chr.14: 96264250-96264250 on Build GRCh38
rs917950992 Chr.14: 96264342-96264342 on Build GRCh38
rs764807889 Chr.14: 96264347-96264347 on Build GRCh38
rs1004605032 Chr.14: 96264574-96264574 on Build GRCh38
rs763956811 Chr.14: 96264224-96264224 on Build GRCh38
rs749897476 Chr.14: 96264690-96264702 on Build GRCh38
rs762289233 Chr.14: 96264516-96264516 on Build GRCh38
rs139028840 Chr.14: 96264443-96264443 on Build GRCh38
rs147132377 Chr.14: 96264679-96264679 on Build GRCh38
rs751373446 Chr.14: 96264232-96264232 on Build GRCh38
rs769590341 Chr.14: 96264569-96264569 on Build GRCh38
rs146437850 Chr.14: 96264694-96264694 on Build GRCh38
rs767266886 Chr.14: 96264328-96264328 on Build GRCh38
rs33925361 Chr.14: 96264381-96264381 on Build GRCh38
rs748095029 Chr.14: 96264385-96264385 on Build GRCh38
rs781135413 Chr.14: 96264562-96264562 on Build GRCh38
rs750696339 Chr.14: 96264428-96264428 on Build GRCh38
rs117760968 Chr.14: 96264568-96264568 on Build GRCh38
rs1046021772 Chr.14: 96264533-96264533 on Build GRCh38
rs184307206 Chr.14: 96264631-96264631 on Build GRCh38
rs1024542247 Chr.14: 96264182-96264182 on Build GRCh38
rs765424214 Chr.14: 96264648-96264648 on Build GRCh38
rs561648647 Chr.14: 96264172-96264172 on Build GRCh38
rs768189659 Chr.14: 96264692-96264692 on Build GRCh38
rs200369442 Chr.14: 96264382-96264382 on Build GRCh38

View Full Product Details