Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 89162614-89163110 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXN3
Entrez Gene ID
1112
Gene Name
forkhead box N3
Gene Aliases
C14orf116, CHES1, PRO1635
UniGene
Hs.434286
Gene Chromosome Location
Chr.14: 89156172-89619150 on Build GRCh38
Transcripts
NM_001085471.1 NM_005197.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6488081 c.1257G>C p.K419N Chr.14: 89162630-89162630 on Build GRCh38
5553382 c.1096G>A p.E366K Chr.14: 89162725-89162725 on Build GRCh38
1207444 c.1090G>A p.D364N Chr.14: 89162797-89162797 on Build GRCh38
4964426 c.999G>T p.E333D Chr.14: 89162888-89162888 on Build GRCh38
6666433 c.1199A>G p.Q400R Chr.14: 89162622-89162622 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759916715 Chr.14: 89162733-89162733 on Build GRCh38
rs764982279 Chr.14: 89162856-89162856 on Build GRCh38
rs377576571 Chr.14: 89162978-89162978 on Build GRCh38
rs141530191 Chr.14: 89162746-89162746 on Build GRCh38
rs3742680 Chr.14: 89163046-89163046 on Build GRCh38
rs571898798 Chr.14: 89162924-89162924 on Build GRCh38
rs567621014 Chr.14: 89163040-89163040 on Build GRCh38
rs776092725 Chr.14: 89162744-89162744 on Build GRCh38
rs774484775 Chr.14: 89162844-89162844 on Build GRCh38
rs61729601 Chr.14: 89162690-89162690 on Build GRCh38
rs767596671 Chr.14: 89162853-89162853 on Build GRCh38
rs777509511 Chr.14: 89162874-89162874 on Build GRCh38
rs768905331 Chr.14: 89162804-89162805 on Build GRCh38
rs12883195 Chr.14: 89162843-89162843 on Build GRCh38
rs146154259 Chr.14: 89162730-89162730 on Build GRCh38
rs752481081 Chr.14: 89162862-89162862 on Build GRCh38
rs756781928 Chr.14: 89162896-89162896 on Build GRCh38
rs146981725 Chr.14: 89162641-89162641 on Build GRCh38
rs746570619 Chr.14: 89162801-89162801 on Build GRCh38
rs1006645159 Chr.14: 89163031-89163031 on Build GRCh38
rs780937118 Chr.14: 89162831-89162831 on Build GRCh38
rs542281263 Chr.14: 89162798-89162798 on Build GRCh38
rs751640543 Chr.14: 89162748-89162748 on Build GRCh38
rs769069776 Chr.14: 89162934-89162934 on Build GRCh38
rs3191540 Chr.14: 89162724-89162724 on Build GRCh38
rs1018322027 Chr.14: 89163043-89163043 on Build GRCh38
rs770769761 Chr.14: 89162729-89162729 on Build GRCh38
rs556664977 Chr.14: 89163054-89163054 on Build GRCh38
rs200553485 Chr.14: 89162834-89162834 on Build GRCh38
rs776920246 Chr.14: 89162957-89162957 on Build GRCh38
rs760264825 Chr.14: 89162660-89162660 on Build GRCh38
rs772625675 Chr.14: 89162725-89162725 on Build GRCh38
rs748983466 Chr.14: 89163011-89163011 on Build GRCh38
rs201853361 Chr.14: 89162650-89162650 on Build GRCh38
rs866692680 Chr.14: 89162919-89162919 on Build GRCh38
rs762081118 Chr.14: 89162750-89162750 on Build GRCh38
rs763494456 Chr.14: 89162841-89162841 on Build GRCh38
rs200918416 Chr.14: 89162651-89162651 on Build GRCh38
rs751531394 Chr.14: 89162891-89162891 on Build GRCh38
rs762602017 Chr.14: 89162963-89162963 on Build GRCh38
rs769619389 Chr.14: 89162909-89162909 on Build GRCh38
rs139766020 Chr.14: 89162950-89162950 on Build GRCh38
rs1002334452 Chr.14: 89163045-89163045 on Build GRCh38
rs150528870 Chr.14: 89162792-89162792 on Build GRCh38
rs746637122 Chr.14: 89162726-89162726 on Build GRCh38
rs542397192 Chr.14: 89162949-89162949 on Build GRCh38
rs143108328 Chr.14: 89162708-89162708 on Build GRCh38
rs758356116 Chr.14: 89162865-89162865 on Build GRCh38
rs990448447 Chr.14: 89163066-89163066 on Build GRCh38
rs766287423 Chr.14: 89162913-89162913 on Build GRCh38
rs1050340410 Chr.14: 89162654-89162654 on Build GRCh38
rs541972213 Chr.14: 89162973-89162973 on Build GRCh38
rs772189579 Chr.14: 89162648-89162656 on Build GRCh38
rs780882135 Chr.14: 89162899-89162899 on Build GRCh38
rs747219009 Chr.14: 89162805-89162807 on Build GRCh38
rs745487150 Chr.14: 89162903-89162903 on Build GRCh38
rs74076955 Chr.14: 89162644-89162644 on Build GRCh38
rs150209397 Chr.14: 89162648-89162648 on Build GRCh38
rs773317819 Chr.14: 89162944-89162944 on Build GRCh38
rs113598579 Chr.14: 89162762-89162762 on Build GRCh38
rs138472417 Chr.14: 89162702-89162702 on Build GRCh38
rs149662874 Chr.14: 89162850-89162850 on Build GRCh38
rs199849564 Chr.14: 89162656-89162656 on Build GRCh38
rs147363460 Chr.14: 89162855-89162855 on Build GRCh38
rs754903884 Chr.14: 89162713-89162713 on Build GRCh38
rs774027873 Chr.14: 89162971-89162972 on Build GRCh38
rs890916639 Chr.14: 89163004-89163004 on Build GRCh38
rs768113760 Chr.14: 89163064-89163064 on Build GRCh38
rs750592549 Chr.14: 89162854-89162854 on Build GRCh38
rs527731955 Chr.14: 89162997-89162997 on Build GRCh38
rs763973847 Chr.14: 89162977-89162977 on Build GRCh38
rs369372657 Chr.14: 89162709-89162709 on Build GRCh38
rs757461952 Chr.14: 89162704-89162704 on Build GRCh38
rs767792979 Chr.14: 89162755-89162755 on Build GRCh38
rs775759046 Chr.14: 89162840-89162840 on Build GRCh38
rs12883064 Chr.14: 89162813-89162813 on Build GRCh38
rs1804717 Chr.14: 89162878-89162878 on Build GRCh38
rs770297981 Chr.14: 89162734-89162734 on Build GRCh38
rs779504673 Chr.14: 89163006-89163006 on Build GRCh38
rs544199861 Chr.14: 89162940-89162940 on Build GRCh38
rs778753018 Chr.14: 89162714-89162714 on Build GRCh38
rs547772835 Chr.14: 89163001-89163001 on Build GRCh38
rs45535535 Chr.14: 89162657-89162657 on Build GRCh38
rs777311000 Chr.14: 89162800-89162800 on Build GRCh38
rs776821245 Chr.14: 89162805-89162813 on Build GRCh38
rs750647040 Chr.14: 89162756-89162756 on Build GRCh38
rs533857726 Chr.14: 89162742-89162750 on Build GRCh38
rs149344132 Chr.14: 89162698-89162698 on Build GRCh38
rs765915065 Chr.14: 89162673-89162673 on Build GRCh38
rs780453992 Chr.14: 89162927-89162927 on Build GRCh38
rs896770352 Chr.14: 89163053-89163053 on Build GRCh38
rs375910548 Chr.14: 89162747-89162747 on Build GRCh38
rs755891097 Chr.14: 89162642-89162642 on Build GRCh38
rs143673672 Chr.14: 89162699-89162699 on Build GRCh38
rs375351484 Chr.14: 89162669-89162669 on Build GRCh38
rs371586722 Chr.14: 89162667-89162667 on Build GRCh38
rs750011321 Chr.14: 89162998-89162998 on Build GRCh38
rs139705919 Chr.14: 89162645-89162645 on Build GRCh38
rs562765734 Chr.14: 89162816-89162816 on Build GRCh38
rs373179687 Chr.14: 89162720-89162720 on Build GRCh38
rs754709521 Chr.14: 89162757-89162757 on Build GRCh38
rs748675321 Chr.14: 89162943-89162943 on Build GRCh38
rs139532153 Chr.14: 89162807-89162807 on Build GRCh38
rs775082179 Chr.14: 89163026-89163026 on Build GRCh38

View Full Product Details