Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 42387674-42387965 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPN3
Entrez Gene ID
825
Gene Name
calpain 3
Gene Aliases
CANP3, CANPL3, LGMD2, LGMD2A, nCL-1, p94
UniGene
Hs.143261
Gene Chromosome Location
Chr.15: 42359500-42412317 on Build GRCh38
Transcripts
NM_000070.2 NM_024344.1 NM_173087.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4054697 c.337T>A p.F113I Chr.15: 42387852-42387852 on Build GRCh38
5793537 c.557A>G p.N186S Chr.15: 42387811-42387811 on Build GRCh38
6232006 c.259G>A p.V87M Chr.15: 42387774-42387774 on Build GRCh38
6232005 c.520G>A p.V174M Chr.15: 42387774-42387774 on Build GRCh38
3501280 c.575C>T p.T192I Chr.15: 42387829-42387829 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs398123149 Chr.15: 42387834-42387834 on Build GRCh38
rs775983864 Chr.15: 42387869-42387869 on Build GRCh38
rs763719290 Chr.15: 42387862-42387862 on Build GRCh38
rs752890866 Chr.15: 42387696-42387696 on Build GRCh38
rs757648915 Chr.15: 42387705-42387705 on Build GRCh38
rs144383442 Chr.15: 42387779-42387779 on Build GRCh38
rs886043108 Chr.15: 42387874-42387874 on Build GRCh38
rs758795961 Chr.15: 42387820-42387820 on Build GRCh38
rs780728918 Chr.15: 42387836-42387836 on Build GRCh38
rs35889956 Chr.15: 42387805-42387805 on Build GRCh38
rs746436020 Chr.15: 42387730-42387730 on Build GRCh38
rs148855999 Chr.15: 42387838-42387838 on Build GRCh38
rs886043429 Chr.15: 42387790-42387790 on Build GRCh38
rs111830217 Chr.15: 42387918-42387918 on Build GRCh38
rs886042296 Chr.15: 42387754-42387754 on Build GRCh38
rs766782470 Chr.15: 42387909-42387909 on Build GRCh38
rs747144192 Chr.15: 42387743-42387743 on Build GRCh38
rs775504869 Chr.15: 42387854-42387854 on Build GRCh38
rs992545025 Chr.15: 42387767-42387767 on Build GRCh38
rs763449646 Chr.15: 42387809-42387809 on Build GRCh38
rs863224964 Chr.15: 42387752-42387752 on Build GRCh38
rs779415824 Chr.15: 42387710-42387710 on Build GRCh38
rs727503837 Chr.15: 42387852-42387866 on Build GRCh38
rs371721054 Chr.15: 42387747-42387747 on Build GRCh38
rs765012463 Chr.15: 42387881-42387881 on Build GRCh38
rs539315630 Chr.15: 42387814-42387814 on Build GRCh38
rs1039308482 Chr.15: 42387885-42387885 on Build GRCh38
rs144299553 Chr.15: 42387735-42387735 on Build GRCh38
rs201344810 Chr.15: 42387740-42387740 on Build GRCh38
rs886043143 Chr.15: 42387835-42387835 on Build GRCh38
rs1001356986 Chr.15: 42387868-42387868 on Build GRCh38
rs752172949 Chr.15: 42387924-42387924 on Build GRCh38
rs371166254 Chr.15: 42387847-42387847 on Build GRCh38
rs781125690 Chr.15: 42387841-42387841 on Build GRCh38
rs776924358 Chr.15: 42387764-42387764 on Build GRCh38
rs774114705 Chr.15: 42387780-42387780 on Build GRCh38
rs767477904 Chr.15: 42387792-42387792 on Build GRCh38
rs750787615 Chr.15: 42387815-42387815 on Build GRCh38
rs748248802 Chr.15: 42387774-42387774 on Build GRCh38
rs80338800 Chr.15: 42387804-42387804 on Build GRCh38
rs541597520 Chr.15: 42387872-42387872 on Build GRCh38
rs771800732 Chr.15: 42387848-42387848 on Build GRCh38
rs17593 Chr.15: 42387860-42387860 on Build GRCh38
rs794727615 Chr.15: 42387787-42387787 on Build GRCh38
rs147808529 Chr.15: 42387806-42387806 on Build GRCh38
rs756847946 Chr.15: 42387811-42387811 on Build GRCh38
rs199718635 Chr.15: 42387843-42387843 on Build GRCh38
rs758845197 Chr.15: 42387898-42387898 on Build GRCh38
rs201660362 Chr.15: 42387889-42387889 on Build GRCh38
rs749375799 Chr.15: 42387813-42387813 on Build GRCh38
rs768426565 Chr.15: 42387844-42387844 on Build GRCh38
rs886042965 Chr.15: 42387788-42387788 on Build GRCh38

View Full Product Details