Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 57604089-57604569 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GCOM1
Entrez Gene ID
145781
Gene Name
GRINL1A complex locus 1
Gene Aliases
GRINL1A, Gcom2, MYZAP, MYZAP-POLR2M, gcom
UniGene
Hs.437256
Gene Chromosome Location
Chr.15: 57591904-57717557 on Build GRCh38
Transcripts
NM_001018090.6 NM_001018091.6 NM_001285900.3 NR_104367.2 NR_104368.2 NR_104369.2 NR_104370.2 NR_104371.3 NM_001018100.4 NM_152451.7

Gene Symbol
MYZAP
Entrez Gene ID
100820829
Gene Name
myocardial zonula adherens protein
Gene Aliases
GCOM1, Gup, MYOZAP
UniGene
Hs.437256
Gene Chromosome Location
Chr.15: 57591904-57685364 on Build GRCh38
Transcripts
NM_001018090.6 NM_001018091.6 NM_001285900.3 NR_104367.2 NR_104368.2 NR_104369.2 NR_104370.2 NR_104371.3 NM_001018100.4 NM_152451.7

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3706646 c.94C>T p.R32W Chr.15: 57604287-57604287 on Build GRCh38
3502673 c.124C>T p.P42S Chr.15: 57604317-57604317 on Build GRCh38
3502670 c.114G>A p.E38E Chr.15: 57604307-57604307 on Build GRCh38
5444203 c.157G>T p.E53* Chr.15: 57604350-57604350 on Build GRCh38
963298 c.152G>T p.R51I Chr.15: 57604345-57604345 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs940980447 Chr.15: 57604542-57604542 on Build GRCh38
rs777508957 Chr.15: 57604433-57604434 on Build GRCh38
rs766525527 Chr.15: 57604397-57604397 on Build GRCh38
rs201832214 Chr.15: 57604295-57604295 on Build GRCh38
rs750539768 Chr.15: 57604246-57604246 on Build GRCh38
rs945164752 Chr.15: 57604322-57604322 on Build GRCh38
rs918819102 Chr.15: 57604550-57604550 on Build GRCh38
rs751584573 Chr.15: 57604339-57604339 on Build GRCh38
rs1018221612 Chr.15: 57604477-57604477 on Build GRCh38
rs927675791 Chr.15: 57604329-57604329 on Build GRCh38
rs372422965 Chr.15: 57604539-57604539 on Build GRCh38
rs7163748 Chr.15: 57604113-57604113 on Build GRCh38
rs527888455 Chr.15: 57604521-57604522 on Build GRCh38
rs746935570 Chr.15: 57604379-57604379 on Build GRCh38
rs756472292 Chr.15: 57604360-57604360 on Build GRCh38
rs375862743 Chr.15: 57604387-57604387 on Build GRCh38
rs188111223 Chr.15: 57604506-57604506 on Build GRCh38
rs202126161 Chr.15: 57604251-57604251 on Build GRCh38
rs182971331 Chr.15: 57604480-57604480 on Build GRCh38
rs756910527 Chr.15: 57604365-57604365 on Build GRCh38
rs150909622 Chr.15: 57604288-57604288 on Build GRCh38
rs575630447 Chr.15: 57604448-57604448 on Build GRCh38
rs190913965 Chr.15: 57604131-57604131 on Build GRCh38
rs752298820 Chr.15: 57604202-57604202 on Build GRCh38
rs755467129 Chr.15: 57604263-57604263 on Build GRCh38
rs941587639 Chr.15: 57604384-57604384 on Build GRCh38
rs553660183 Chr.15: 57604369-57604369 on Build GRCh38
rs376423450 Chr.15: 57604349-57604349 on Build GRCh38
rs191523528 Chr.15: 57604398-57604398 on Build GRCh38
rs766416861 Chr.15: 57604258-57604258 on Build GRCh38
rs748949826 Chr.15: 57604362-57604362 on Build GRCh38
rs757568596 Chr.15: 57604433-57604433 on Build GRCh38
rs371899236 Chr.15: 57604300-57604300 on Build GRCh38
rs2453091 Chr.15: 57604460-57604460 on Build GRCh38
rs1038628488 Chr.15: 57604547-57604547 on Build GRCh38
rs755810342 Chr.15: 57604276-57604276 on Build GRCh38
rs781553829 Chr.15: 57604268-57604268 on Build GRCh38
rs766505405 Chr.15: 57604332-57604332 on Build GRCh38
rs528840699 Chr.15: 57604164-57604164 on Build GRCh38
rs112322135 Chr.15: 57604478-57604478 on Build GRCh38
rs1010780654 Chr.15: 57604486-57604486 on Build GRCh38
rs559981914 Chr.15: 57604520-57604520 on Build GRCh38
rs573471782 Chr.15: 57604374-57604374 on Build GRCh38
rs978317886 Chr.15: 57604511-57604511 on Build GRCh38
rs1017361789 Chr.15: 57604430-57604430 on Build GRCh38
rs571053346 Chr.15: 57604359-57604359 on Build GRCh38
rs1005749772 Chr.15: 57604167-57604167 on Build GRCh38
rs748398018 Chr.15: 57604269-57604269 on Build GRCh38
rs915088281 Chr.15: 57604306-57604306 on Build GRCh38
rs772067728 Chr.15: 57604296-57604296 on Build GRCh38
rs183081206 Chr.15: 57604231-57604231 on Build GRCh38
rs1022610592 Chr.15: 57604505-57604505 on Build GRCh38
rs142253131 Chr.15: 57604284-57604284 on Build GRCh38
rs115438075 Chr.15: 57604368-57604368 on Build GRCh38
rs762098850 Chr.15: 57604225-57604225 on Build GRCh38
rs764535335 Chr.15: 57604328-57604328 on Build GRCh38
rs763928814 Chr.15: 57604302-57604302 on Build GRCh38
rs775386543 Chr.15: 57604299-57604299 on Build GRCh38
rs768404027 Chr.15: 57604386-57604386 on Build GRCh38
rs895389220 Chr.15: 57604146-57604146 on Build GRCh38
rs755483534 Chr.15: 57604343-57604343 on Build GRCh38
rs1017497232 Chr.15: 57604292-57604292 on Build GRCh38
rs778191892 Chr.15: 57604361-57604361 on Build GRCh38
rs906202874 Chr.15: 57604112-57604112 on Build GRCh38
rs574457473 Chr.15: 57604287-57604287 on Build GRCh38
rs367740908 Chr.15: 57604540-57604540 on Build GRCh38
rs763178575 Chr.15: 57604389-57604389 on Build GRCh38
rs773347544 Chr.15: 57604326-57604326 on Build GRCh38
rs1029774477 Chr.15: 57604420-57604420 on Build GRCh38
rs927760192 Chr.15: 57604517-57604517 on Build GRCh38
rs560019733 Chr.15: 57604143-57604143 on Build GRCh38
rs779051692 Chr.15: 57604237-57604237 on Build GRCh38
rs897279676 Chr.15: 57604179-57604179 on Build GRCh38
rs555455993 Chr.15: 57604439-57604439 on Build GRCh38
rs994714148 Chr.15: 57604409-57604409 on Build GRCh38
rs529197267 Chr.15: 57604538-57604538 on Build GRCh38
rs2172611 Chr.15: 57604168-57604168 on Build GRCh38
rs140163703 Chr.15: 57604294-57604294 on Build GRCh38

View Full Product Details