Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 76216146-76216719 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ETFA
Entrez Gene ID
2108
Gene Name
electron transfer flavoprotein alpha subunit
Gene Aliases
EMA, GA2, MADD
UniGene
Hs.39925
Gene Chromosome Location
Chr.15: 76216228-76311469 on Build GRCh38
Transcripts
NM_000126.3 NM_001127716.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
965096 c.986T>C p.I329T Chr.15: 76216575-76216575 on Build GRCh38
122902 c.993G>C p.K331N Chr.15: 76216568-76216568 on Build GRCh38
6015873 c.973G>A p.E325K Chr.15: 76216588-76216588 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs894182234 Chr.15: 76216305-76216305 on Build GRCh38
rs757885420 Chr.15: 76216598-76216598 on Build GRCh38
rs1011239116 Chr.15: 76216314-76216314 on Build GRCh38
rs746324276 Chr.15: 76216532-76216532 on Build GRCh38
rs770017562 Chr.15: 76216534-76216534 on Build GRCh38
rs183981964 Chr.15: 76216513-76216513 on Build GRCh38
rs888810875 Chr.15: 76216488-76216488 on Build GRCh38
rs752246889 Chr.15: 76216580-76216580 on Build GRCh38
rs757193547 Chr.15: 76216613-76216613 on Build GRCh38
rs558438624 Chr.15: 76216691-76216691 on Build GRCh38
rs751413443 Chr.15: 76216612-76216612 on Build GRCh38
rs539654182 Chr.15: 76216271-76216271 on Build GRCh38
rs902903489 Chr.15: 76216362-76216362 on Build GRCh38
rs774771335 Chr.15: 76216554-76216554 on Build GRCh38
rs763013799 Chr.15: 76216541-76216541 on Build GRCh38
rs770760376 Chr.15: 76216528-76216528 on Build GRCh38
rs1046532469 Chr.15: 76216212-76216212 on Build GRCh38
rs746786188 Chr.15: 76216527-76216527 on Build GRCh38
rs771862007 Chr.15: 76216333-76216333 on Build GRCh38
rs187702861 Chr.15: 76216337-76216337 on Build GRCh38
rs1803551 Chr.15: 76216387-76216387 on Build GRCh38
rs938153736 Chr.15: 76216676-76216676 on Build GRCh38
rs537265828 Chr.15: 76216632-76216632 on Build GRCh38
rs1010835441 Chr.15: 76216467-76216467 on Build GRCh38
rs928703840 Chr.15: 76216230-76216230 on Build GRCh38
rs765039243 Chr.15: 76216576-76216576 on Build GRCh38
rs758341953 Chr.15: 76216521-76216521 on Build GRCh38
rs908636035 Chr.15: 76216581-76216581 on Build GRCh38
rs750818335 Chr.15: 76216564-76216564 on Build GRCh38
rs199789825 Chr.15: 76216548-76216548 on Build GRCh38
rs570818940 Chr.15: 76216617-76216617 on Build GRCh38
rs1007177094 Chr.15: 76216492-76216492 on Build GRCh38
rs112013323 Chr.15: 76216343-76216343 on Build GRCh38
rs188037746 Chr.15: 76216532-76216532 on Build GRCh38
rs1803554 Chr.15: 76216420-76216420 on Build GRCh38
rs770003380 Chr.15: 76216638-76216638 on Build GRCh38
rs1044020946 Chr.15: 76216475-76216475 on Build GRCh38
rs143535013 Chr.15: 76216225-76216228 on Build GRCh38
rs758815411 Chr.15: 76216612-76216614 on Build GRCh38
rs984102947 Chr.15: 76216561-76216562 on Build GRCh38
rs1803550 Chr.15: 76216343-76216343 on Build GRCh38
rs940193710 Chr.15: 76216190-76216190 on Build GRCh38
rs200260934 Chr.15: 76216537-76216537 on Build GRCh38
rs754706670 Chr.15: 76216512-76216512 on Build GRCh38
rs201424632 Chr.15: 76216560-76216560 on Build GRCh38
rs369208131 Chr.15: 76216675-76216675 on Build GRCh38
rs80292319 Chr.15: 76216291-76216291 on Build GRCh38
rs886051487 Chr.15: 76216504-76216504 on Build GRCh38
rs368489298 Chr.15: 76216228-76216228 on Build GRCh38
rs999112929 Chr.15: 76216453-76216453 on Build GRCh38
rs200391818 Chr.15: 76216555-76216555 on Build GRCh38
rs752726045 Chr.15: 76216221-76216224 on Build GRCh38
rs3830441 Chr.15: 76216236-76216239 on Build GRCh38
rs934748062 Chr.15: 76216695-76216695 on Build GRCh38
rs140121559 Chr.15: 76216460-76216460 on Build GRCh38
rs1018683716 Chr.15: 76216497-76216497 on Build GRCh38
rs778811845 Chr.15: 76216195-76216195 on Build GRCh38
rs779678354 Chr.15: 76216615-76216618 on Build GRCh38
rs777638283 Chr.15: 76216525-76216525 on Build GRCh38
rs865799474 Chr.15: 76216323-76216323 on Build GRCh38
rs747621281 Chr.15: 76216516-76216516 on Build GRCh38
rs192522256 Chr.15: 76216466-76216466 on Build GRCh38
rs981435358 Chr.15: 76216662-76216662 on Build GRCh38
rs982587908 Chr.15: 76216672-76216672 on Build GRCh38
rs777742977 Chr.15: 76216601-76216601 on Build GRCh38
rs1055372802 Chr.15: 76216292-76216292 on Build GRCh38
rs759203081 Chr.15: 76216567-76216567 on Build GRCh38
rs762435540 Chr.15: 76216558-76216558 on Build GRCh38
rs3198975 Chr.15: 76216514-76216514 on Build GRCh38
rs531912622 Chr.15: 76216435-76216435 on Build GRCh38

View Full Product Details