Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 78275369-78275858 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJA4
Entrez Gene ID
55466
Gene Name
DnaJ heat shock protein family (Hsp40) member A4
Gene Aliases
MST104, MSTP104, PRO1472
UniGene
Hs.513053
Gene Chromosome Location
Chr.15: 78264145-78282196 on Build GRCh38
Transcripts
NM_001130182.1 NM_001130183.1 NM_018602.3 XM_005254535.3 XM_006720601.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
965343 c.859G>A p.V287I Chr.15: 78275710-78275710 on Build GRCh38
3816822 c.877+2T>C p.? Chr.15: 78275730-78275730 on Build GRCh38
965341 c.851G>A p.R284Q Chr.15: 78275702-78275702 on Build GRCh38
1588109 c.778G>A p.V260I Chr.15: 78275710-78275710 on Build GRCh38
7317944 c.639C>G p.V213V Chr.15: 78275571-78275571 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs569472097 Chr.15: 78275815-78275815 on Build GRCh38
rs776120715 Chr.15: 78275751-78275751 on Build GRCh38
rs775828560 Chr.15: 78275641-78275641 on Build GRCh38
rs1044485931 Chr.15: 78275718-78275718 on Build GRCh38
rs747913548 Chr.15: 78275418-78275418 on Build GRCh38
rs761731720 Chr.15: 78275455-78275468 on Build GRCh38
rs757504374 Chr.15: 78275710-78275710 on Build GRCh38
rs781515229 Chr.15: 78275744-78275745 on Build GRCh38
rs369601244 Chr.15: 78275713-78275713 on Build GRCh38
rs751859584 Chr.15: 78275702-78275702 on Build GRCh38
rs752694531 Chr.15: 78275761-78275761 on Build GRCh38
rs763264121 Chr.15: 78275734-78275735 on Build GRCh38
rs765508494 Chr.15: 78275626-78275627 on Build GRCh38
rs942115381 Chr.15: 78275708-78275708 on Build GRCh38
rs777655450 Chr.15: 78275701-78275701 on Build GRCh38
rs753342413 Chr.15: 78275669-78275669 on Build GRCh38
rs772012724 Chr.15: 78275472-78275472 on Build GRCh38
rs377270562 Chr.15: 78275745-78275745 on Build GRCh38
rs17852881 Chr.15: 78275528-78275528 on Build GRCh38
rs374556413 Chr.15: 78275549-78275549 on Build GRCh38
rs929462398 Chr.15: 78275409-78275409 on Build GRCh38
rs769010222 Chr.15: 78275739-78275739 on Build GRCh38
rs749983602 Chr.15: 78275453-78275453 on Build GRCh38
rs747222954 Chr.15: 78275578-78275578 on Build GRCh38
rs771150154 Chr.15: 78275597-78275597 on Build GRCh38
rs767180015 Chr.15: 78275815-78275817 on Build GRCh38
rs780664664 Chr.15: 78275726-78275726 on Build GRCh38
rs988620503 Chr.15: 78275590-78275590 on Build GRCh38
rs770623271 Chr.15: 78275746-78275746 on Build GRCh38
rs781248827 Chr.15: 78275712-78275712 on Build GRCh38
rs749233343 Chr.15: 78275465-78275465 on Build GRCh38
rs758326761 Chr.15: 78275699-78275699 on Build GRCh38
rs985208054 Chr.15: 78275827-78275827 on Build GRCh38
rs902040867 Chr.15: 78275406-78275406 on Build GRCh38
rs772292438 Chr.15: 78275489-78275489 on Build GRCh38
rs760251066 Chr.15: 78275658-78275658 on Build GRCh38
rs762824361 Chr.15: 78275439-78275441 on Build GRCh38
rs753727387 Chr.15: 78275545-78275545 on Build GRCh38
rs377119803 Chr.15: 78275714-78275714 on Build GRCh38
rs781289588 Chr.15: 78275607-78275607 on Build GRCh38
rs777336776 Chr.15: 78275516-78275516 on Build GRCh38
rs574804218 Chr.15: 78275455-78275455 on Build GRCh38
rs750670908 Chr.15: 78275657-78275658 on Build GRCh38
rs751951146 Chr.15: 78275739-78275740 on Build GRCh38
rs550894511 Chr.15: 78275542-78275542 on Build GRCh38
rs749734650 Chr.15: 78275733-78275733 on Build GRCh38
rs78756785 Chr.15: 78275707-78275707 on Build GRCh38
rs905876057 Chr.15: 78275740-78275740 on Build GRCh38
rs1017953458 Chr.15: 78275831-78275831 on Build GRCh38
rs372092147 Chr.15: 78275693-78275693 on Build GRCh38
rs755569100 Chr.15: 78275743-78275743 on Build GRCh38
rs967821 Chr.15: 78275511-78275511 on Build GRCh38
rs563571693 Chr.15: 78275753-78275758 on Build GRCh38
rs758236723 Chr.15: 78275560-78275560 on Build GRCh38
rs1021454573 Chr.15: 78275813-78275813 on Build GRCh38
rs776756103 Chr.15: 78275648-78275648 on Build GRCh38
rs148370822 Chr.15: 78275682-78275682 on Build GRCh38
rs1048296090 Chr.15: 78275414-78275414 on Build GRCh38
rs374360974 Chr.15: 78275531-78275531 on Build GRCh38
rs760918969 Chr.15: 78275570-78275570 on Build GRCh38
rs1010560185 Chr.15: 78275799-78275799 on Build GRCh38
rs765263538 Chr.15: 78275681-78275681 on Build GRCh38
rs368143683 Chr.15: 78275650-78275650 on Build GRCh38
rs1021798739 Chr.15: 78275602-78275602 on Build GRCh38
rs199939846 Chr.15: 78275526-78275526 on Build GRCh38
rs956082664 Chr.15: 78275589-78275589 on Build GRCh38
rs765907325 Chr.15: 78275661-78275661 on Build GRCh38
rs202118844 Chr.15: 78275614-78275614 on Build GRCh38
rs753068873 Chr.15: 78275774-78275776 on Build GRCh38
rs765549959 Chr.15: 78275523-78275523 on Build GRCh38
rs963715518 Chr.15: 78275798-78275798 on Build GRCh38
rs202103276 Chr.15: 78275615-78275615 on Build GRCh38
rs376559741 Chr.15: 78275484-78275484 on Build GRCh38
rs368335700 Chr.15: 78275537-78275537 on Build GRCh38
rs778208624 Chr.15: 78275564-78275564 on Build GRCh38
rs764641416 Chr.15: 78275755-78275755 on Build GRCh38
rs760020416 Chr.15: 78275520-78275520 on Build GRCh38
rs142259465 Chr.15: 78275741-78275741 on Build GRCh38
rs778599846 Chr.15: 78275553-78275553 on Build GRCh38
rs764000890 Chr.15: 78275774-78275774 on Build GRCh38
rs377692773 Chr.15: 78275457-78275457 on Build GRCh38
rs370699762 Chr.15: 78275494-78275494 on Build GRCh38
rs373981227 Chr.15: 78275742-78275742 on Build GRCh38
rs371732302 Chr.15: 78275753-78275753 on Build GRCh38
rs370302985 Chr.15: 78275477-78275477 on Build GRCh38
rs778706654 Chr.15: 78275483-78275483 on Build GRCh38
rs752590260 Chr.15: 78275558-78275558 on Build GRCh38
rs193218284 Chr.15: 78275674-78275674 on Build GRCh38
rs766891817 Chr.15: 78275735-78275736 on Build GRCh38
rs747126967 Chr.15: 78275496-78275496 on Build GRCh38
rs887941803 Chr.15: 78275705-78275705 on Build GRCh38
rs762919789 Chr.15: 78275773-78275773 on Build GRCh38

View Full Product Details