Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 82989801-82990239 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C15orf40
Entrez Gene ID
123207
Gene Name
chromosome 15 open reading frame 40
Gene Aliases
-
UniGene
Hs.352541
Gene Chromosome Location
Chr.15: 82988297-83012298 on Build GRCh38
Transcripts
NM_001160116.1 XM_011521213.2 NM_031452.3

Gene Symbol
FAM103A1
Entrez Gene ID
83640
Gene Name
family with sequence similarity 103 member A1
Gene Aliases
C15orf18, HsT19360, RAM
UniGene
Hs.80624
Gene Chromosome Location
Chr.15: 82986203-82991057 on Build GRCh38
Transcripts
NM_001160116.1 XM_011521213.2 NM_031452.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5028012 c.209G>C p.R70T Chr.15: 82989919-82989919 on Build GRCh38
1129083 c.203A>G p.D68G Chr.15: 82989913-82989913 on Build GRCh38
5649234 c.194G>C p.R65T Chr.15: 82989904-82989904 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs773262295 Chr.15: 82989958-82989958 on Build GRCh38
rs774236972 Chr.15: 82990068-82990068 on Build GRCh38
rs775320229 Chr.15: 82989972-82989972 on Build GRCh38
rs754187287 Chr.15: 82990011-82990011 on Build GRCh38
rs200149891 Chr.15: 82990056-82990056 on Build GRCh38
rs372110976 Chr.15: 82989977-82989977 on Build GRCh38
rs750009440 Chr.15: 82990074-82990074 on Build GRCh38
rs371817583 Chr.15: 82989987-82989987 on Build GRCh38
rs759202087 Chr.15: 82989877-82989877 on Build GRCh38
rs750012838 Chr.15: 82989993-82989993 on Build GRCh38
rs749792257 Chr.15: 82989940-82989940 on Build GRCh38
rs200941963 Chr.15: 82989949-82989949 on Build GRCh38
rs745561686 Chr.15: 82989926-82989926 on Build GRCh38
rs201626883 Chr.15: 82990044-82990044 on Build GRCh38
rs773496944 Chr.15: 82989871-82989871 on Build GRCh38
rs199995569 Chr.15: 82990080-82990080 on Build GRCh38
rs948781474 Chr.15: 82990042-82990042 on Build GRCh38
rs780271255 Chr.15: 82990010-82990010 on Build GRCh38
rs761069366 Chr.15: 82990027-82990029 on Build GRCh38
rs779511862 Chr.15: 82990023-82990023 on Build GRCh38
rs1007278149 Chr.15: 82990200-82990201 on Build GRCh38
rs767086479 Chr.15: 82989992-82989992 on Build GRCh38
rs368166772 Chr.15: 82990055-82990055 on Build GRCh38
rs758376783 Chr.15: 82989898-82989898 on Build GRCh38
rs115038311 Chr.15: 82989836-82989836 on Build GRCh38
rs769346792 Chr.15: 82989942-82989942 on Build GRCh38
rs751514592 Chr.15: 82989985-82989985 on Build GRCh38
rs891989727 Chr.15: 82990101-82990101 on Build GRCh38
rs753565744 Chr.15: 82990098-82990098 on Build GRCh38
rs373665648 Chr.15: 82990021-82990021 on Build GRCh38
rs763578602 Chr.15: 82989913-82989913 on Build GRCh38
rs774624347 Chr.15: 82989856-82989856 on Build GRCh38
rs766915523 Chr.15: 82990072-82990072 on Build GRCh38
rs938952496 Chr.15: 82990075-82990075 on Build GRCh38
rs775771129 Chr.15: 82989831-82989831 on Build GRCh38
rs761184040 Chr.15: 82989986-82989986 on Build GRCh38
rs753230663 Chr.15: 82990112-82990112 on Build GRCh38
rs903609204 Chr.15: 82990146-82990146 on Build GRCh38
rs752542510 Chr.15: 82989895-82989895 on Build GRCh38
rs768827464 Chr.15: 82989838-82989838 on Build GRCh38
rs756394469 Chr.15: 82989927-82989927 on Build GRCh38
rs777954563 Chr.15: 82990029-82990029 on Build GRCh38
rs368003453 Chr.15: 82989930-82989930 on Build GRCh38
rs772512462 Chr.15: 82989830-82989831 on Build GRCh38
rs200751289 Chr.15: 82989921-82989921 on Build GRCh38
rs182796186 Chr.15: 82989925-82989925 on Build GRCh38
rs772214697 Chr.15: 82989952-82989952 on Build GRCh38
rs763956639 Chr.15: 82989983-82989983 on Build GRCh38
rs780939173 Chr.15: 82989992-82989993 on Build GRCh38
rs765107913 Chr.15: 82989879-82989879 on Build GRCh38
rs764576250 Chr.15: 82990071-82990072 on Build GRCh38
rs1022080752 Chr.15: 82990137-82990137 on Build GRCh38
rs367747091 Chr.15: 82989967-82989967 on Build GRCh38
rs371706950 Chr.15: 82989943-82989943 on Build GRCh38
rs1003044557 Chr.15: 82989826-82989827 on Build GRCh38
rs147533881 Chr.15: 82989963-82989963 on Build GRCh38
rs761850135 Chr.15: 82990069-82990069 on Build GRCh38
rs760242337 Chr.15: 82990078-82990078 on Build GRCh38
rs755841042 Chr.15: 82990009-82990009 on Build GRCh38
rs768465912 Chr.15: 82990063-82990063 on Build GRCh38
rs955418748 Chr.15: 82990019-82990019 on Build GRCh38
rs1029682304 Chr.15: 82989988-82989988 on Build GRCh38
rs780508972 Chr.15: 82989934-82989934 on Build GRCh38
rs1044382171 Chr.15: 82990065-82990065 on Build GRCh38
rs35679302 Chr.15: 82989968-82989969 on Build GRCh38
rs748696584 Chr.15: 82990026-82990026 on Build GRCh38
rs574710727 Chr.15: 82990217-82990217 on Build GRCh38
rs201913813 Chr.15: 82990111-82990111 on Build GRCh38
rs755353627 Chr.15: 82990103-82990103 on Build GRCh38
rs771249659 Chr.15: 82990045-82990045 on Build GRCh38
rs762120699 Chr.15: 82989860-82989860 on Build GRCh38
rs1017300056 Chr.15: 82990201-82990205 on Build GRCh38
rs772009818 Chr.15: 82990027-82990027 on Build GRCh38

View Full Product Details