Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 66722751-66723341 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DYNC1LI2
Entrez Gene ID
1783
Gene Name
dynein cytoplasmic 1 light intermediate chain 2
Gene Aliases
DNCLI2, LIC2
UniGene
Hs.369068
Gene Chromosome Location
Chr.16: 66720893-66751828 on Build GRCh38
Transcripts
NM_001286157.1 NM_001323955.1 NM_006141.2 NR_136658.1

SNP Detail

SNP ID Chromosome Location
rs1040946853 Chr.16: 66722833-66722833 on Build GRCh38
rs942314141 Chr.16: 66722787-66722787 on Build GRCh38
rs1052517931 Chr.16: 66723172-66723172 on Build GRCh38
rs560335414 Chr.16: 66722851-66722851 on Build GRCh38
rs1003960026 Chr.16: 66723284-66723284 on Build GRCh38
rs893715486 Chr.16: 66722910-66722910 on Build GRCh38
rs565603735 Chr.16: 66722801-66722801 on Build GRCh38
rs748896149 Chr.16: 66722820-66722820 on Build GRCh38
rs906625944 Chr.16: 66722860-66722860 on Build GRCh38
rs1026740377 Chr.16: 66723264-66723264 on Build GRCh38
rs533397006 Chr.16: 66722989-66722989 on Build GRCh38
rs951072666 Chr.16: 66722773-66722773 on Build GRCh38
rs763904241 Chr.16: 66722932-66722932 on Build GRCh38
rs1039607083 Chr.16: 66723277-66723280 on Build GRCh38
rs909584901 Chr.16: 66723045-66723045 on Build GRCh38
rs140699046 Chr.16: 66723182-66723182 on Build GRCh38
rs753557019 Chr.16: 66723280-66723280 on Build GRCh38
rs572758397 Chr.16: 66722908-66722908 on Build GRCh38
rs963833704 Chr.16: 66722982-66722982 on Build GRCh38
rs932192902 Chr.16: 66723106-66723106 on Build GRCh38
rs760993001 Chr.16: 66722987-66722987 on Build GRCh38
rs999511021 Chr.16: 66722893-66722893 on Build GRCh38
rs1026707860 Chr.16: 66722994-66722994 on Build GRCh38
rs546784016 Chr.16: 66723033-66723033 on Build GRCh38
rs140091728 Chr.16: 66722951-66722951 on Build GRCh38
rs546069573 Chr.16: 66723094-66723094 on Build GRCh38
rs573644297 Chr.16: 66722909-66722909 on Build GRCh38
rs371571850 Chr.16: 66722809-66722812 on Build GRCh38
rs916836594 Chr.16: 66723072-66723072 on Build GRCh38
rs143957422 Chr.16: 66722965-66722965 on Build GRCh38
rs774036381 Chr.16: 66722920-66722920 on Build GRCh38
rs935362376 Chr.16: 66723144-66723144 on Build GRCh38
rs948220720 Chr.16: 66722809-66722809 on Build GRCh38
rs146426107 Chr.16: 66723035-66723035 on Build GRCh38
rs558037340 Chr.16: 66723231-66723231 on Build GRCh38
rs899768488 Chr.16: 66723283-66723283 on Build GRCh38
rs965415999 Chr.16: 66723045-66723045 on Build GRCh38
rs896522626 Chr.16: 66723177-66723177 on Build GRCh38
rs976397727 Chr.16: 66723079-66723079 on Build GRCh38
rs557815931 Chr.16: 66723221-66723221 on Build GRCh38
rs180675837 Chr.16: 66723192-66723192 on Build GRCh38
rs982387820 Chr.16: 66722796-66722796 on Build GRCh38
rs935444456 Chr.16: 66723180-66723180 on Build GRCh38
rs935339182 Chr.16: 66722937-66722937 on Build GRCh38
rs1023965250 Chr.16: 66722967-66722967 on Build GRCh38
rs529344996 Chr.16: 66723179-66723179 on Build GRCh38
rs922164701 Chr.16: 66722848-66722848 on Build GRCh38
rs1052400052 Chr.16: 66722950-66722950 on Build GRCh38
rs1019317500 Chr.16: 66723297-66723297 on Build GRCh38
rs866697793 Chr.16: 66723174-66723174 on Build GRCh38
rs1031445015 Chr.16: 66722907-66722907 on Build GRCh38
rs979985910 Chr.16: 66723114-66723114 on Build GRCh38
rs951228797 Chr.16: 66723025-66723025 on Build GRCh38
rs925373453 Chr.16: 66723128-66723128 on Build GRCh38
rs973583721 Chr.16: 66723057-66723057 on Build GRCh38
rs903922155 Chr.16: 66722931-66722931 on Build GRCh38
rs777182006 Chr.16: 66723196-66723196 on Build GRCh38
rs951105324 Chr.16: 66722998-66722998 on Build GRCh38
rs531252347 Chr.16: 66722957-66722957 on Build GRCh38
rs375180487 Chr.16: 66723216-66723216 on Build GRCh38
rs188457622 Chr.16: 66722916-66722916 on Build GRCh38
rs771847007 Chr.16: 66722986-66722986 on Build GRCh38
rs948219484 Chr.16: 66723086-66723086 on Build GRCh38
rs886864013 Chr.16: 66723279-66723279 on Build GRCh38
rs1013542892 Chr.16: 66722961-66722961 on Build GRCh38
rs908145453 Chr.16: 66722819-66722819 on Build GRCh38
rs564346267 Chr.16: 66722972-66722972 on Build GRCh38
rs895355628 Chr.16: 66722797-66722797 on Build GRCh38

View Full Product Details