Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 58115889-58116284 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFAP20
Entrez Gene ID
29105
Gene Name
cilia and flagella associated protein 20
Gene Aliases
C16orf80, EVORF, GTL3, fSAP23
UniGene
Hs.532755
Gene Chromosome Location
Chr.16: 58113588-58129425 on Build GRCh38
Transcripts
NM_013242.2 NR_136632.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
971955 c.267C>T p.F89F Chr.16: 58116050-58116050 on Build GRCh38
971956 c.174T>C p.Y58Y Chr.16: 58116143-58116143 on Build GRCh38
352606 c.173A>G p.Y58C Chr.16: 58116144-58116144 on Build GRCh38
4186564 c.257A>G p.Y86C Chr.16: 58116060-58116060 on Build GRCh38
1721563 c.220C>T p.P74S Chr.16: 58116097-58116097 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750431990 Chr.16: 58116179-58116179 on Build GRCh38
rs903401622 Chr.16: 58116241-58116241 on Build GRCh38
rs954612366 Chr.16: 58116120-58116120 on Build GRCh38
rs1046402380 Chr.16: 58116209-58116209 on Build GRCh38
rs778506878 Chr.16: 58116164-58116164 on Build GRCh38
rs368794662 Chr.16: 58116038-58116038 on Build GRCh38
rs748876456 Chr.16: 58116157-58116157 on Build GRCh38
rs537375750 Chr.16: 58116227-58116227 on Build GRCh38
rs370126486 Chr.16: 58116155-58116155 on Build GRCh38
rs755771991 Chr.16: 58116145-58116145 on Build GRCh38
rs772917562 Chr.16: 58116176-58116176 on Build GRCh38
rs535828181 Chr.16: 58116180-58116180 on Build GRCh38
rs1043319140 Chr.16: 58116231-58116231 on Build GRCh38
rs774424328 Chr.16: 58116020-58116020 on Build GRCh38
rs374579477 Chr.16: 58116050-58116050 on Build GRCh38
rs765237816 Chr.16: 58116113-58116113 on Build GRCh38
rs778033905 Chr.16: 58116132-58116132 on Build GRCh38
rs780309823 Chr.16: 58116246-58116246 on Build GRCh38
rs768774349 Chr.16: 58116001-58116001 on Build GRCh38
rs771226850 Chr.16: 58116031-58116032 on Build GRCh38
rs374513704 Chr.16: 58116022-58116022 on Build GRCh38
rs554558802 Chr.16: 58116202-58116202 on Build GRCh38
rs376526057 Chr.16: 58116183-58116183 on Build GRCh38
rs369645269 Chr.16: 58116159-58116159 on Build GRCh38
rs964334310 Chr.16: 58115937-58115937 on Build GRCh38
rs146197714 Chr.16: 58116023-58116023 on Build GRCh38
rs140567256 Chr.16: 58116143-58116143 on Build GRCh38
rs916090791 Chr.16: 58116216-58116216 on Build GRCh38
rs373566442 Chr.16: 58116181-58116181 on Build GRCh38
rs532781083 Chr.16: 58115959-58115959 on Build GRCh38
rs778262462 Chr.16: 58116004-58116005 on Build GRCh38
rs772004271 Chr.16: 58116170-58116170 on Build GRCh38
rs774727875 Chr.16: 58116115-58116117 on Build GRCh38
rs941895713 Chr.16: 58116171-58116171 on Build GRCh38
rs112723051 Chr.16: 58116114-58116114 on Build GRCh38
rs779471629 Chr.16: 58115998-58115998 on Build GRCh38
rs113401330 Chr.16: 58116226-58116226 on Build GRCh38
rs1011068727 Chr.16: 58115938-58115938 on Build GRCh38
rs759596720 Chr.16: 58116053-58116053 on Build GRCh38
rs748118318 Chr.16: 58116165-58116165 on Build GRCh38
rs922908521 Chr.16: 58116046-58116046 on Build GRCh38
rs377474377 Chr.16: 58116156-58116156 on Build GRCh38
rs766588053 Chr.16: 58116043-58116043 on Build GRCh38
rs769451889 Chr.16: 58115991-58115991 on Build GRCh38
rs749733917 Chr.16: 58116028-58116029 on Build GRCh38
rs773010246 Chr.16: 58116025-58116025 on Build GRCh38
rs765325805 Chr.16: 58116194-58116194 on Build GRCh38

View Full Product Details