Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 55869521-55869892 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CES5A
Entrez Gene ID
221223
Gene Name
carboxylesterase 5A
Gene Aliases
CAUXIN, CES4C1, CES5, CES7, HEL126
UniGene
Hs.350800
Gene Chromosome Location
Chr.16: 55846154-55956031 on Build GRCh38
Transcripts
NM_001143685.1 NM_001190158.1 NM_145024.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5517015 c.505-1G>A p.? Chr.16: 55869745-55869745 on Build GRCh38
6144786 c.473A>C p.D158A Chr.16: 55869689-55869689 on Build GRCh38
703401 c.445T>A p.F149I Chr.16: 55869717-55869717 on Build GRCh38
5420692 c.468C>T p.I156I Chr.16: 55869694-55869694 on Build GRCh38
6144790 c.484C>A p.L162M Chr.16: 55869678-55869678 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs147903375 Chr.16: 55869611-55869611 on Build GRCh38
rs537674498 Chr.16: 55869769-55869769 on Build GRCh38
rs548244981 Chr.16: 55869783-55869801 on Build GRCh38
rs140373908 Chr.16: 55869668-55869668 on Build GRCh38
rs764669055 Chr.16: 55869657-55869657 on Build GRCh38
rs13338753 Chr.16: 55869636-55869636 on Build GRCh38
rs386791040 Chr.16: 55869783-55869804 on Build GRCh38
rs777727565 Chr.16: 55869635-55869635 on Build GRCh38
rs534853602 Chr.16: 55869661-55869661 on Build GRCh38
rs868721312 Chr.16: 55869755-55869755 on Build GRCh38
rs541452405 Chr.16: 55869702-55869702 on Build GRCh38
rs552972383 Chr.16: 55869681-55869681 on Build GRCh38
rs764759062 Chr.16: 55869583-55869583 on Build GRCh38
rs545885054 Chr.16: 55869736-55869736 on Build GRCh38
rs550867880 Chr.16: 55869804-55869804 on Build GRCh38
rs566984526 Chr.16: 55869863-55869863 on Build GRCh38
rs756976918 Chr.16: 55869720-55869720 on Build GRCh38
rs183860763 Chr.16: 55869580-55869580 on Build GRCh38
rs13338754 Chr.16: 55869642-55869642 on Build GRCh38
rs772022505 Chr.16: 55869620-55869620 on Build GRCh38
rs753366410 Chr.16: 55869678-55869678 on Build GRCh38
rs370935298 Chr.16: 55869783-55869783 on Build GRCh38
rs529553324 Chr.16: 55869796-55869796 on Build GRCh38
rs757935443 Chr.16: 55869683-55869683 on Build GRCh38
rs762995423 Chr.16: 55869655-55869655 on Build GRCh38
rs776865733 Chr.16: 55869560-55869560 on Build GRCh38
rs752219905 Chr.16: 55869682-55869682 on Build GRCh38
rs568884687 Chr.16: 55869839-55869839 on Build GRCh38
rs373929877 Chr.16: 55869605-55869605 on Build GRCh38
rs774943049 Chr.16: 55869752-55869752 on Build GRCh38
rs775618040 Chr.16: 55869733-55869733 on Build GRCh38
rs533173579 Chr.16: 55869850-55869850 on Build GRCh38
rs768208581 Chr.16: 55869758-55869758 on Build GRCh38
rs751953582 Chr.16: 55869586-55869586 on Build GRCh38
rs13338809 Chr.16: 55869754-55869754 on Build GRCh38
rs149581721 Chr.16: 55869862-55869862 on Build GRCh38
rs765369709 Chr.16: 55869570-55869570 on Build GRCh38
rs751950787 Chr.16: 55869738-55869738 on Build GRCh38
rs759481969 Chr.16: 55869561-55869561 on Build GRCh38
rs768880588 Chr.16: 55869750-55869750 on Build GRCh38
rs550582032 Chr.16: 55869701-55869701 on Build GRCh38
rs142782650 Chr.16: 55869637-55869637 on Build GRCh38
rs757740160 Chr.16: 55869604-55869604 on Build GRCh38
rs144674577 Chr.16: 55869610-55869610 on Build GRCh38
rs200272022 Chr.16: 55869794-55869794 on Build GRCh38
rs368829056 Chr.16: 55869763-55869763 on Build GRCh38
rs368830616 Chr.16: 55869727-55869727 on Build GRCh38
rs777831855 Chr.16: 55869784-55869784 on Build GRCh38
rs145577128 Chr.16: 55869728-55869728 on Build GRCh38
rs757841774 Chr.16: 55869774-55869774 on Build GRCh38
rs574722799 Chr.16: 55869697-55869697 on Build GRCh38
rs138599318 Chr.16: 55869762-55869762 on Build GRCh38
rs775629408 Chr.16: 55869646-55869646 on Build GRCh38
rs752315467 Chr.16: 55869773-55869773 on Build GRCh38
rs563233890 Chr.16: 55869722-55869722 on Build GRCh38
rs373764241 Chr.16: 55869645-55869645 on Build GRCh38
rs760325275 Chr.16: 55869678-55869681 on Build GRCh38

View Full Product Details