Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 10704939-10705447 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADPRM
Entrez Gene ID
56985
Gene Name
ADP-ribose/CDP-alcohol diphosphatase, manganese dependent
Gene Aliases
C17orf48, MDS006, NBLA03831
UniGene
Hs.47668
Gene Chromosome Location
Chr.17: 10697610-10711558 on Build GRCh38
Transcripts
NM_020233.4 XM_005256738.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
287765 c.218G>C p.G73A Chr.17: 10705144-10705144 on Build GRCh38
2925617 c.47G>A p.R16H Chr.17: 10704973-10704973 on Build GRCh38
2925616 c.46C>T p.R16C Chr.17: 10704972-10704972 on Build GRCh38
2925620 c.169G>A p.E57K Chr.17: 10705095-10705095 on Build GRCh38
6595364 c.229G>A p.D77N Chr.17: 10705155-10705155 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780260481 Chr.17: 10705054-10705054 on Build GRCh38
rs762398462 Chr.17: 10705108-10705108 on Build GRCh38
rs1044866597 Chr.17: 10705361-10705361 on Build GRCh38
rs772727299 Chr.17: 10705107-10705107 on Build GRCh38
rs375110496 Chr.17: 10705200-10705200 on Build GRCh38
rs780987054 Chr.17: 10705073-10705073 on Build GRCh38
rs533592080 Chr.17: 10705234-10705234 on Build GRCh38
rs145477522 Chr.17: 10705212-10705212 on Build GRCh38
rs762714267 Chr.17: 10705000-10705000 on Build GRCh38
rs746749892 Chr.17: 10705061-10705061 on Build GRCh38
rs779101506 Chr.17: 10705041-10705041 on Build GRCh38
rs953306247 Chr.17: 10704986-10704986 on Build GRCh38
rs186083978 Chr.17: 10705269-10705269 on Build GRCh38
rs866517700 Chr.17: 10705105-10705105 on Build GRCh38
rs566687256 Chr.17: 10705348-10705348 on Build GRCh38
rs762091779 Chr.17: 10705330-10705330 on Build GRCh38
rs751758193 Chr.17: 10705376-10705376 on Build GRCh38
rs551272880 Chr.17: 10705160-10705160 on Build GRCh38
rs112596400 Chr.17: 10705176-10705176 on Build GRCh38
rs774137621 Chr.17: 10705280-10705280 on Build GRCh38
rs777016595 Chr.17: 10705294-10705294 on Build GRCh38
rs761104737 Chr.17: 10705149-10705149 on Build GRCh38
rs1035529608 Chr.17: 10705420-10705420 on Build GRCh38
rs759805636 Chr.17: 10705008-10705008 on Build GRCh38
rs376596572 Chr.17: 10705341-10705341 on Build GRCh38
rs376218266 Chr.17: 10704989-10704989 on Build GRCh38
rs766486800 Chr.17: 10705374-10705374 on Build GRCh38
rs79976834 Chr.17: 10704990-10704990 on Build GRCh38
rs771649830 Chr.17: 10705292-10705292 on Build GRCh38
rs769413233 Chr.17: 10704996-10704996 on Build GRCh38
rs746814549 Chr.17: 10705296-10705296 on Build GRCh38
rs765697723 Chr.17: 10705182-10705182 on Build GRCh38
rs766150924 Chr.17: 10705002-10705002 on Build GRCh38
rs1052600 Chr.17: 10705092-10705092 on Build GRCh38
rs537951182 Chr.17: 10705124-10705124 on Build GRCh38
rs746840041 Chr.17: 10704973-10704973 on Build GRCh38
rs539316390 Chr.17: 10705211-10705211 on Build GRCh38
rs767289812 Chr.17: 10705079-10705080 on Build GRCh38
rs377007137 Chr.17: 10704971-10704971 on Build GRCh38
rs961826422 Chr.17: 10705028-10705028 on Build GRCh38
rs749300500 Chr.17: 10705421-10705421 on Build GRCh38
rs770620729 Chr.17: 10705274-10705274 on Build GRCh38
rs151290748 Chr.17: 10705050-10705050 on Build GRCh38
rs745552088 Chr.17: 10705285-10705285 on Build GRCh38
rs755144056 Chr.17: 10705385-10705385 on Build GRCh38
rs936339998 Chr.17: 10705162-10705162 on Build GRCh38
rs769164270 Chr.17: 10705102-10705102 on Build GRCh38
rs755872218 Chr.17: 10705403-10705403 on Build GRCh38
rs926304936 Chr.17: 10705154-10705154 on Build GRCh38
rs74947284 Chr.17: 10705155-10705155 on Build GRCh38
rs201484409 Chr.17: 10705219-10705219 on Build GRCh38
rs764615973 Chr.17: 10705150-10705150 on Build GRCh38
rs142369493 Chr.17: 10705370-10705370 on Build GRCh38
rs754423462 Chr.17: 10705027-10705027 on Build GRCh38
rs762336014 Chr.17: 10705165-10705165 on Build GRCh38
rs756036026 Chr.17: 10705247-10705247 on Build GRCh38
rs370126868 Chr.17: 10704972-10704972 on Build GRCh38
rs202121379 Chr.17: 10705388-10705388 on Build GRCh38
rs375371081 Chr.17: 10705186-10705186 on Build GRCh38
rs142410541 Chr.17: 10704995-10704995 on Build GRCh38
rs369477322 Chr.17: 10705357-10705357 on Build GRCh38
rs761123314 Chr.17: 10705016-10705016 on Build GRCh38
rs34940296 Chr.17: 10705201-10705201 on Build GRCh38
rs752469408 Chr.17: 10705402-10705402 on Build GRCh38
rs757256713 Chr.17: 10705031-10705031 on Build GRCh38
rs546267018 Chr.17: 10705113-10705113 on Build GRCh38
rs758488017 Chr.17: 10705189-10705189 on Build GRCh38
rs777574932 Chr.17: 10705267-10705267 on Build GRCh38
rs772709476 Chr.17: 10704998-10704998 on Build GRCh38
rs777751083 Chr.17: 10705406-10705406 on Build GRCh38
rs1052601 Chr.17: 10705401-10705401 on Build GRCh38
rs775334747 Chr.17: 10705148-10705149 on Build GRCh38
rs773081127 Chr.17: 10705246-10705246 on Build GRCh38
rs768498506 Chr.17: 10705066-10705066 on Build GRCh38
rs368444216 Chr.17: 10705093-10705093 on Build GRCh38
rs371191811 Chr.17: 10705114-10705114 on Build GRCh38

View Full Product Details