Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 16342437-16343003 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CENPV
Entrez Gene ID
201161
Gene Name
centromere protein V
Gene Aliases
3110013H01Rik, CENP-V, PRR6, p30
UniGene
Hs.433422
Gene Chromosome Location
Chr.17: 16342534-16353498 on Build GRCh38
Transcripts
NM_181716.2 XM_017025349.1 XM_017025350.1 XM_017025352.1 XM_017025353.1 XM_017025354.1 XM_017025355.1 XM_017025356.1

Gene Symbol
PIGL
Entrez Gene ID
9487
Gene Name
phosphatidylinositol glycan anchor biosynthesis class L
Gene Aliases
CHIME
UniGene
Hs.499793
Gene Chromosome Location
Chr.17: 16217195-16351800 on Build GRCh38
Transcripts
NM_181716.2 XM_017025349.1 XM_017025350.1 XM_017025352.1 XM_017025353.1 XM_017025354.1 XM_017025355.1 XM_017025356.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5750375 c.704C>T p.P235L Chr.17: 16342932-16342932 on Build GRCh38
4843185 c.798C>T p.I266I Chr.17: 16342838-16342838 on Build GRCh38
5513267 c.695-4G>A p.? Chr.17: 16342945-16342945 on Build GRCh38
6880549 c.737T>C p.M246T Chr.17: 16342899-16342899 on Build GRCh38
4671551 c.706_707insC p.H236fs*5 Chr.17: 16342929-16342930 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs878942516 Chr.17: 16342880-16342880 on Build GRCh38
rs3112524 Chr.17: 16342464-16342464 on Build GRCh38
rs200571765 Chr.17: 16342842-16342842 on Build GRCh38
rs575054843 Chr.17: 16342704-16342704 on Build GRCh38
rs191566838 Chr.17: 16342596-16342596 on Build GRCh38
rs925932814 Chr.17: 16342762-16342762 on Build GRCh38
rs1039215319 Chr.17: 16342888-16342888 on Build GRCh38
rs757070811 Chr.17: 16342619-16342619 on Build GRCh38
rs150259876 Chr.17: 16342480-16342480 on Build GRCh38
rs1036486485 Chr.17: 16342517-16342517 on Build GRCh38
rs900025668 Chr.17: 16342660-16342660 on Build GRCh38
rs770946294 Chr.17: 16342906-16342906 on Build GRCh38
rs141730289 Chr.17: 16342806-16342806 on Build GRCh38
rs755968089 Chr.17: 16342746-16342746 on Build GRCh38
rs1050365460 Chr.17: 16342501-16342501 on Build GRCh38
rs934159044 Chr.17: 16342933-16342933 on Build GRCh38
rs751719331 Chr.17: 16342788-16342788 on Build GRCh38
rs546227206 Chr.17: 16342770-16342770 on Build GRCh38
rs751134025 Chr.17: 16342584-16342584 on Build GRCh38
rs763576945 Chr.17: 16342541-16342541 on Build GRCh38
rs897539289 Chr.17: 16342840-16342840 on Build GRCh38
rs550008287 Chr.17: 16342815-16342815 on Build GRCh38
rs904865995 Chr.17: 16342873-16342873 on Build GRCh38
rs753833978 Chr.17: 16342607-16342607 on Build GRCh38
rs1036215609 Chr.17: 16342831-16342831 on Build GRCh38
rs1004170937 Chr.17: 16342740-16342740 on Build GRCh38
rs751951346 Chr.17: 16342789-16342789 on Build GRCh38
rs1027290552 Chr.17: 16342663-16342663 on Build GRCh38
rs767263125 Chr.17: 16342939-16342939 on Build GRCh38
rs773521150 Chr.17: 16342883-16342883 on Build GRCh38
rs981268422 Chr.17: 16342758-16342758 on Build GRCh38
rs755573201 Chr.17: 16342830-16342831 on Build GRCh38
rs116070540 Chr.17: 16342744-16342744 on Build GRCh38
rs563803424 Chr.17: 16342748-16342748 on Build GRCh38
rs778537468 Chr.17: 16342946-16342946 on Build GRCh38
rs766588766 Chr.17: 16342782-16342782 on Build GRCh38
rs777205703 Chr.17: 16342812-16342812 on Build GRCh38
rs570998535 Chr.17: 16342556-16342557 on Build GRCh38
rs182541337 Chr.17: 16342940-16342940 on Build GRCh38
rs931902252 Chr.17: 16342496-16342496 on Build GRCh38
rs376909753 Chr.17: 16342513-16342513 on Build GRCh38
rs949900891 Chr.17: 16342544-16342544 on Build GRCh38
rs748554307 Chr.17: 16342967-16342967 on Build GRCh38
rs890280694 Chr.17: 16342507-16342507 on Build GRCh38
rs750132900 Chr.17: 16342707-16342707 on Build GRCh38
rs781744849 Chr.17: 16342928-16342929 on Build GRCh38
rs756078695 Chr.17: 16342965-16342966 on Build GRCh38
rs558516718 Chr.17: 16342465-16342465 on Build GRCh38
rs767301376 Chr.17: 16342797-16342797 on Build GRCh38
rs777997033 Chr.17: 16342910-16342910 on Build GRCh38
rs114309459 Chr.17: 16342752-16342752 on Build GRCh38
rs547635883 Chr.17: 16342838-16342838 on Build GRCh38
rs915763061 Chr.17: 16342481-16342481 on Build GRCh38
rs778731942 Chr.17: 16342825-16342825 on Build GRCh38
rs1008763589 Chr.17: 16342515-16342515 on Build GRCh38
rs371456499 Chr.17: 16342874-16342874 on Build GRCh38
rs778363238 Chr.17: 16342972-16342972 on Build GRCh38
rs781480377 Chr.17: 16342964-16342964 on Build GRCh38
rs765285345 Chr.17: 16342774-16342774 on Build GRCh38
rs202110735 Chr.17: 16342953-16342953 on Build GRCh38
rs1001724331 Chr.17: 16342678-16342678 on Build GRCh38
rs4791667 Chr.17: 16342905-16342905 on Build GRCh38
rs934324126 Chr.17: 16342604-16342604 on Build GRCh38
rs930381512 Chr.17: 16342841-16342841 on Build GRCh38
rs943034127 Chr.17: 16342845-16342845 on Build GRCh38
rs913351045 Chr.17: 16342463-16342463 on Build GRCh38
rs924498232 Chr.17: 16342581-16342581 on Build GRCh38
rs892804583 Chr.17: 16342617-16342617 on Build GRCh38
rs960477418 Chr.17: 16342705-16342705 on Build GRCh38
rs374037613 Chr.17: 16342954-16342954 on Build GRCh38
rs7477 Chr.17: 16342702-16342702 on Build GRCh38
rs969055197 Chr.17: 16342669-16342669 on Build GRCh38
rs373072160 Chr.17: 16342796-16342796 on Build GRCh38
rs994886463 Chr.17: 16342625-16342625 on Build GRCh38
rs756771426 Chr.17: 16342816-16342816 on Build GRCh38
rs202152998 Chr.17: 16342957-16342960 on Build GRCh38
rs1038771541 Chr.17: 16342639-16342639 on Build GRCh38
rs370608826 Chr.17: 16342945-16342945 on Build GRCh38
rs986035381 Chr.17: 16342805-16342805 on Build GRCh38
rs774584321 Chr.17: 16342907-16342907 on Build GRCh38
rs773331721 Chr.17: 16342775-16342775 on Build GRCh38
rs536038408 Chr.17: 16342745-16342745 on Build GRCh38
rs769991641 Chr.17: 16342970-16342970 on Build GRCh38
rs760595963 Chr.17: 16342943-16342943 on Build GRCh38
rs763232508 Chr.17: 16342781-16342781 on Build GRCh38
rs377031551 Chr.17: 16342867-16342867 on Build GRCh38
rs924526302 Chr.17: 16342761-16342761 on Build GRCh38

View Full Product Details