Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 3977778-3978330 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EEF2
Entrez Gene ID
1938
Gene Name
eukaryotic translation elongation factor 2
Gene Aliases
EEF-2, EF-2, EF2, SCA26
UniGene
Hs.515070
Gene Chromosome Location
Chr.19: 3976056-3985463 on Build GRCh38
Transcripts
NM_001961.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
122778 c.1898G>A p.R633H Chr.19: 3977988-3977988 on Build GRCh38
6651763 c.1896G>A p.A632A Chr.19: 3977990-3977990 on Build GRCh38
3960047 c.1940G>C p.R647P Chr.19: 3977946-3977946 on Build GRCh38
6651761 c.2044G>A p.G682S Chr.19: 3977842-3977842 on Build GRCh38
3823003 c.2016C>G p.L672L Chr.19: 3977870-3977870 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770914707 Chr.19: 3977805-3977805 on Build GRCh38
rs781370166 Chr.19: 3978223-3978224 on Build GRCh38
rs746161915 Chr.19: 3977975-3977975 on Build GRCh38
rs761397803 Chr.19: 3977803-3977803 on Build GRCh38
rs898745314 Chr.19: 3978232-3978232 on Build GRCh38
rs112383785 Chr.19: 3978175-3978175 on Build GRCh38
rs997082004 Chr.19: 3978233-3978233 on Build GRCh38
rs767608925 Chr.19: 3978226-3978226 on Build GRCh38
rs776225912 Chr.19: 3977802-3977802 on Build GRCh38
rs959565247 Chr.19: 3978306-3978306 on Build GRCh38
rs587777052 Chr.19: 3978099-3978099 on Build GRCh38
rs149651258 Chr.19: 3977867-3977867 on Build GRCh38
rs767911696 Chr.19: 3977948-3977948 on Build GRCh38
rs370836485 Chr.19: 3978125-3978125 on Build GRCh38
rs113239377 Chr.19: 3978255-3978255 on Build GRCh38
rs148878859 Chr.19: 3978140-3978140 on Build GRCh38
rs368819417 Chr.19: 3977876-3977876 on Build GRCh38
rs774639089 Chr.19: 3977941-3977941 on Build GRCh38
rs759879199 Chr.19: 3977945-3977945 on Build GRCh38
rs754364920 Chr.19: 3978013-3978013 on Build GRCh38
rs779931357 Chr.19: 3977882-3977882 on Build GRCh38
rs945307250 Chr.19: 3977907-3977907 on Build GRCh38
rs975604065 Chr.19: 3977825-3977825 on Build GRCh38
rs997431543 Chr.19: 3978036-3978036 on Build GRCh38
rs769162318 Chr.19: 3977918-3977918 on Build GRCh38
rs536867142 Chr.19: 3978288-3978288 on Build GRCh38
rs35987350 Chr.19: 3978121-3978121 on Build GRCh38
rs770242626 Chr.19: 3978044-3978044 on Build GRCh38
rs145540039 Chr.19: 3978116-3978116 on Build GRCh38
rs139615561 Chr.19: 3978056-3978056 on Build GRCh38
rs148771691 Chr.19: 3977915-3977915 on Build GRCh38
rs781672509 Chr.19: 3977832-3977832 on Build GRCh38
rs751931356 Chr.19: 3978272-3978272 on Build GRCh38
rs1054859937 Chr.19: 3977799-3977799 on Build GRCh38
rs773875633 Chr.19: 3978051-3978051 on Build GRCh38
rs993788425 Chr.19: 3978309-3978309 on Build GRCh38
rs766965840 Chr.19: 3978164-3978164 on Build GRCh38
rs201976284 Chr.19: 3977994-3977994 on Build GRCh38
rs143308239 Chr.19: 3978196-3978196 on Build GRCh38
rs750648849 Chr.19: 3978018-3978018 on Build GRCh38
rs111645561 Chr.19: 3977817-3977817 on Build GRCh38
rs979060977 Chr.19: 3978095-3978095 on Build GRCh38
rs148318922 Chr.19: 3978290-3978290 on Build GRCh38
rs1016486852 Chr.19: 3978148-3978148 on Build GRCh38
rs149189348 Chr.19: 3977981-3977981 on Build GRCh38
rs143659055 Chr.19: 3978158-3978158 on Build GRCh38
rs763197514 Chr.19: 3978122-3978122 on Build GRCh38
rs112868528 Chr.19: 3977924-3977924 on Build GRCh38
rs923316325 Chr.19: 3978245-3978245 on Build GRCh38
rs772796691 Chr.19: 3977920-3977920 on Build GRCh38
rs1053673842 Chr.19: 3978049-3978049 on Build GRCh38
rs1031985273 Chr.19: 3978297-3978297 on Build GRCh38
rs754786270 Chr.19: 3977977-3977977 on Build GRCh38
rs1000492123 Chr.19: 3978292-3978292 on Build GRCh38
rs746684937 Chr.19: 3978215-3978215 on Build GRCh38
rs979345487 Chr.19: 3977911-3977911 on Build GRCh38
rs1010199601 Chr.19: 3977814-3977814 on Build GRCh38
rs755411634 Chr.19: 3977828-3977828 on Build GRCh38
rs142731069 Chr.19: 3977916-3977916 on Build GRCh38
rs971710297 Chr.19: 3978152-3978152 on Build GRCh38
rs200515602 Chr.19: 3977969-3977969 on Build GRCh38
rs1007902036 Chr.19: 3978055-3978055 on Build GRCh38
rs777751138 Chr.19: 3978035-3978035 on Build GRCh38
rs913793636 Chr.19: 3978227-3978227 on Build GRCh38
rs755946485 Chr.19: 3978034-3978034 on Build GRCh38
rs768970441 Chr.19: 3978076-3978076 on Build GRCh38
rs200284152 Chr.19: 3978258-3978259 on Build GRCh38
rs567219818 Chr.19: 3978209-3978209 on Build GRCh38
rs1029194030 Chr.19: 3977951-3977951 on Build GRCh38
rs755931480 Chr.19: 3978129-3978129 on Build GRCh38
rs144336814 Chr.19: 3977868-3977868 on Build GRCh38
rs777373355 Chr.19: 3978301-3978301 on Build GRCh38
rs753183188 Chr.19: 3977959-3977959 on Build GRCh38
rs753259149 Chr.19: 3977843-3977843 on Build GRCh38
rs779267368 Chr.19: 3977973-3977973 on Build GRCh38
rs369669207 Chr.19: 3977801-3977801 on Build GRCh38
rs373868450 Chr.19: 3978149-3978149 on Build GRCh38
rs966389526 Chr.19: 3978079-3978079 on Build GRCh38
rs771632838 Chr.19: 3978205-3978205 on Build GRCh38
rs778327793 Chr.19: 3978185-3978185 on Build GRCh38
rs745451916 Chr.19: 3977897-3977897 on Build GRCh38
rs762472006 Chr.19: 3977921-3977921 on Build GRCh38
rs772529820 Chr.19: 3977983-3977983 on Build GRCh38
rs926371830 Chr.19: 3978206-3978206 on Build GRCh38
rs773988050 Chr.19: 3977991-3977991 on Build GRCh38
rs147383237 Chr.19: 3977960-3977960 on Build GRCh38
rs139427550 Chr.19: 3977978-3977978 on Build GRCh38
rs566897549 Chr.19: 3978161-3978161 on Build GRCh38
rs781090546 Chr.19: 3978026-3978026 on Build GRCh38
rs377744684 Chr.19: 3978043-3978043 on Build GRCh38
rs138298305 Chr.19: 3977809-3977809 on Build GRCh38
rs777167717 Chr.19: 3978101-3978101 on Build GRCh38
rs201326053 Chr.19: 3978050-3978050 on Build GRCh38
rs757356247 Chr.19: 3978016-3978016 on Build GRCh38
rs759664411 Chr.19: 3978203-3978203 on Build GRCh38
rs752880795 Chr.19: 3978228-3978228 on Build GRCh38
rs145009005 Chr.19: 3978005-3978005 on Build GRCh38
rs368234090 Chr.19: 3978068-3978068 on Build GRCh38
rs1008330982 Chr.19: 3978269-3978269 on Build GRCh38
rs750010552 Chr.19: 3977807-3977807 on Build GRCh38
rs779756129 Chr.19: 3978208-3978208 on Build GRCh38
rs764042018 Chr.19: 3977968-3977968 on Build GRCh38
rs374223472 Chr.19: 3978262-3978262 on Build GRCh38
rs980995029 Chr.19: 3978010-3978010 on Build GRCh38
rs149757304 Chr.19: 3978066-3978066 on Build GRCh38
rs933393660 Chr.19: 3978256-3978256 on Build GRCh38
rs757520735 Chr.19: 3978205-3978206 on Build GRCh38
rs200589086 Chr.19: 3977955-3977955 on Build GRCh38
rs867219501 Chr.19: 3977946-3977946 on Build GRCh38
rs878966894 Chr.19: 3977816-3977816 on Build GRCh38
rs373062093 Chr.19: 3977990-3977990 on Build GRCh38
rs776708734 Chr.19: 3977903-3977903 on Build GRCh38
rs945326951 Chr.19: 3978230-3978230 on Build GRCh38
rs775864126 Chr.19: 3978002-3978002 on Build GRCh38
rs1038339661 Chr.19: 3978212-3978212 on Build GRCh38
rs147570651 Chr.19: 3978017-3978017 on Build GRCh38
rs766599785 Chr.19: 3977810-3977810 on Build GRCh38
rs375212891 Chr.19: 3978053-3978053 on Build GRCh38
rs374243613 Chr.19: 3978271-3978271 on Build GRCh38
rs758714357 Chr.19: 3978020-3978020 on Build GRCh38
rs751952139 Chr.19: 3977827-3977827 on Build GRCh38
rs200869338 Chr.19: 3978107-3978107 on Build GRCh38
rs759237233 Chr.19: 3977993-3977993 on Build GRCh38
rs986429881 Chr.19: 3977879-3977879 on Build GRCh38
rs775329938 Chr.19: 3977972-3977972 on Build GRCh38
rs1020646538 Chr.19: 3978059-3978059 on Build GRCh38

View Full Product Details