Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 29817012-29817577 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNE1
Entrez Gene ID
898
Gene Name
cyclin E1
Gene Aliases
CCNE, pCCNE1
UniGene
Hs.244723
Gene Chromosome Location
Chr.19: 29811994-29824317 on Build GRCh38
Transcripts
NM_001238.3 NM_001322259.1 NM_001322261.1 NM_001322262.1 XM_011527440.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5567092 c.284G>A p.R95Q Chr.19: 29817240-29817240 on Build GRCh38
994395 c.191A>T p.N64I Chr.19: 29817147-29817147 on Build GRCh38
6965476 c.419T>G p.L140R Chr.19: 29817498-29817498 on Build GRCh38
5608026 c.434G>T p.R145L Chr.19: 29817513-29817513 on Build GRCh38
6919352 c.353A>C p.K118T Chr.19: 29817432-29817432 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs918480017 Chr.19: 29817503-29817503 on Build GRCh38
rs775011048 Chr.19: 29817251-29817251 on Build GRCh38
rs370834318 Chr.19: 29817314-29817314 on Build GRCh38
rs904766205 Chr.19: 29817411-29817411 on Build GRCh38
rs199930427 Chr.19: 29817291-29817291 on Build GRCh38
rs749237881 Chr.19: 29817119-29817119 on Build GRCh38
rs746361520 Chr.19: 29817166-29817166 on Build GRCh38
rs762615251 Chr.19: 29817174-29817174 on Build GRCh38
rs530187511 Chr.19: 29817312-29817312 on Build GRCh38
rs751795932 Chr.19: 29817305-29817305 on Build GRCh38
rs1013008407 Chr.19: 29817043-29817043 on Build GRCh38
rs776946024 Chr.19: 29817159-29817159 on Build GRCh38
rs985275587 Chr.19: 29817202-29817202 on Build GRCh38
rs141877912 Chr.19: 29817146-29817146 on Build GRCh38
rs763924283 Chr.19: 29817106-29817106 on Build GRCh38
rs777759310 Chr.19: 29817116-29817116 on Build GRCh38
rs948660664 Chr.19: 29817059-29817059 on Build GRCh38
rs566527651 Chr.19: 29817110-29817110 on Build GRCh38
rs773174972 Chr.19: 29817390-29817390 on Build GRCh38
rs113246237 Chr.19: 29817518-29817518 on Build GRCh38
rs771807268 Chr.19: 29817269-29817269 on Build GRCh38
rs747414395 Chr.19: 29817227-29817227 on Build GRCh38
rs998966219 Chr.19: 29817162-29817162 on Build GRCh38
rs774197405 Chr.19: 29817177-29817177 on Build GRCh38
rs368948662 Chr.19: 29817264-29817264 on Build GRCh38
rs367736690 Chr.19: 29817331-29817331 on Build GRCh38
rs1023190641 Chr.19: 29817061-29817061 on Build GRCh38
rs771474736 Chr.19: 29817152-29817152 on Build GRCh38
rs3218045 Chr.19: 29817478-29817478 on Build GRCh38
rs200655498 Chr.19: 29817268-29817268 on Build GRCh38
rs770236821 Chr.19: 29817240-29817240 on Build GRCh38
rs761971284 Chr.19: 29817427-29817427 on Build GRCh38
rs767327617 Chr.19: 29817192-29817192 on Build GRCh38
rs201895026 Chr.19: 29817421-29817421 on Build GRCh38
rs781431455 Chr.19: 29817309-29817309 on Build GRCh38
rs377242899 Chr.19: 29817101-29817101 on Build GRCh38
rs867465231 Chr.19: 29817249-29817249 on Build GRCh38
rs754338237 Chr.19: 29817493-29817493 on Build GRCh38
rs563038680 Chr.19: 29817303-29817303 on Build GRCh38
rs200391933 Chr.19: 29817118-29817118 on Build GRCh38
rs762884762 Chr.19: 29817292-29817292 on Build GRCh38
rs200523129 Chr.19: 29817468-29817468 on Build GRCh38
rs778224499 Chr.19: 29817147-29817147 on Build GRCh38
rs1043317884 Chr.19: 29817402-29817402 on Build GRCh38
rs747657093 Chr.19: 29817537-29817537 on Build GRCh38
rs756615915 Chr.19: 29817318-29817318 on Build GRCh38
rs112478436 Chr.19: 29817087-29817087 on Build GRCh38
rs920500412 Chr.19: 29817271-29817271 on Build GRCh38
rs750448424 Chr.19: 29817193-29817193 on Build GRCh38
rs1047210476 Chr.19: 29817097-29817097 on Build GRCh38
rs574899763 Chr.19: 29817239-29817239 on Build GRCh38
rs779134402 Chr.19: 29817143-29817143 on Build GRCh38
rs373301343 Chr.19: 29817289-29817289 on Build GRCh38
rs558562399 Chr.19: 29817120-29817120 on Build GRCh38
rs759514188 Chr.19: 29817400-29817400 on Build GRCh38
rs778451343 Chr.19: 29817333-29817333 on Build GRCh38
rs749779505 Chr.19: 29817322-29817322 on Build GRCh38
rs755163346 Chr.19: 29817134-29817134 on Build GRCh38
rs764883335 Chr.19: 29817096-29817096 on Build GRCh38
rs148162215 Chr.19: 29817229-29817229 on Build GRCh38
rs79630552 Chr.19: 29817297-29817297 on Build GRCh38
rs1029777496 Chr.19: 29817401-29817401 on Build GRCh38
rs760714380 Chr.19: 29817285-29817285 on Build GRCh38
rs778311875 Chr.19: 29817536-29817536 on Build GRCh38
rs780479023 Chr.19: 29817320-29817320 on Build GRCh38
rs748726668 Chr.19: 29817253-29817253 on Build GRCh38
rs1031890538 Chr.19: 29817346-29817346 on Build GRCh38
rs371619938 Chr.19: 29817086-29817086 on Build GRCh38
rs756093210 Chr.19: 29817199-29817199 on Build GRCh38
rs373059711 Chr.19: 29817355-29817355 on Build GRCh38
rs546304591 Chr.19: 29817068-29817068 on Build GRCh38
rs973067352 Chr.19: 29817495-29817495 on Build GRCh38
rs560232932 Chr.19: 29817551-29817551 on Build GRCh38
rs769333246 Chr.19: 29817167-29817167 on Build GRCh38
rs770510994 Chr.19: 29817392-29817393 on Build GRCh38
rs147120845 Chr.19: 29817203-29817203 on Build GRCh38
rs781616452 Chr.19: 29817238-29817238 on Build GRCh38
rs576282928 Chr.19: 29817391-29817391 on Build GRCh38
rs758728524 Chr.19: 29817209-29817209 on Build GRCh38
rs577353301 Chr.19: 29817280-29817280 on Build GRCh38
rs776768280 Chr.19: 29817393-29817393 on Build GRCh38
rs763612235 Chr.19: 29817175-29817175 on Build GRCh38
rs201174024 Chr.19: 29817150-29817150 on Build GRCh38
rs767658103 Chr.19: 29817431-29817431 on Build GRCh38
rs1041100306 Chr.19: 29817334-29817334 on Build GRCh38
rs771629660 Chr.19: 29817544-29817544 on Build GRCh38
rs201199358 Chr.19: 29817426-29817426 on Build GRCh38
rs374431756 Chr.19: 29817145-29817145 on Build GRCh38
rs752779016 Chr.19: 29817205-29817205 on Build GRCh38
rs761454024 Chr.19: 29817178-29817178 on Build GRCh38
rs748567177 Chr.19: 29817323-29817323 on Build GRCh38
rs757478852 Chr.19: 29817308-29817308 on Build GRCh38
rs777793550 Chr.19: 29817210-29817210 on Build GRCh38
rs869223733 Chr.19: 29817505-29817506 on Build GRCh38
rs376093279 Chr.19: 29817356-29817356 on Build GRCh38
rs920427774 Chr.19: 29817069-29817069 on Build GRCh38
rs373713583 Chr.19: 29817169-29817169 on Build GRCh38
rs750796728 Chr.19: 29817445-29817445 on Build GRCh38
rs141027304 Chr.19: 29817259-29817259 on Build GRCh38

View Full Product Details