Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 50014492-50015045 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLEU2
Entrez Gene ID
8847
Gene Name
deleted in lymphocytic leukemia 2 (non-protein coding)
Gene Aliases
1B4, BCMSUN, DLB2, LEU2, LINC00022, MIR15AHG, NCRNA00022, RFP2OS, TRIM13OS
UniGene
Hs.547964
Gene Chromosome Location
Chr.13: 49982552-50125541 on Build GRCh38
Transcripts
NR_002612.1 NM_001007278.2 NM_005798.4 NM_052811.3 NM_213590.2

Gene Symbol
TRIM13
Entrez Gene ID
10206
Gene Name
tripartite motif containing 13
Gene Aliases
CAR, DLEU5, LEU5, RFP2, RNF77
UniGene
Hs.436922
Gene Chromosome Location
Chr.13: 49997007-50018467 on Build GRCh38
Transcripts
NR_002612.1 NM_001007278.2 NM_005798.4 NM_052811.3 NM_213590.2

SNP Detail

SNP ID Chromosome Location
rs547402113 Chr.13: 50014912-50014912 on Build GRCh38
rs527617884 Chr.13: 50014575-50014575 on Build GRCh38
rs930037815 Chr.13: 50014560-50014560 on Build GRCh38
rs1022131887 Chr.13: 50014527-50014530 on Build GRCh38
rs985619114 Chr.13: 50014792-50014792 on Build GRCh38
rs918625463 Chr.13: 50015008-50015008 on Build GRCh38
rs148246424 Chr.13: 50014688-50014689 on Build GRCh38
rs892225351 Chr.13: 50014688-50014688 on Build GRCh38
rs561158365 Chr.13: 50014761-50014761 on Build GRCh38
rs1035466518 Chr.13: 50014744-50014745 on Build GRCh38
rs553883836 Chr.13: 50014630-50014630 on Build GRCh38
rs888304393 Chr.13: 50014578-50014578 on Build GRCh38
rs976745870 Chr.13: 50014526-50014526 on Build GRCh38
rs1028686859 Chr.13: 50014889-50014889 on Build GRCh38
rs944070956 Chr.13: 50014525-50014525 on Build GRCh38
rs906417787 Chr.13: 50014635-50014635 on Build GRCh38
rs1030644458 Chr.13: 50014649-50014649 on Build GRCh38
rs895667868 Chr.13: 50014747-50014747 on Build GRCh38
rs549892244 Chr.13: 50014836-50014836 on Build GRCh38
rs750363148 Chr.13: 50014821-50014821 on Build GRCh38
rs918668140 Chr.13: 50014551-50014551 on Build GRCh38
rs1010687001 Chr.13: 50014702-50014702 on Build GRCh38
rs947888094 Chr.13: 50014582-50014582 on Build GRCh38
rs953229525 Chr.13: 50014898-50014898 on Build GRCh38
rs1035518870 Chr.13: 50014573-50014574 on Build GRCh38
rs973135510 Chr.13: 50014632-50014633 on Build GRCh38
rs929962372 Chr.13: 50015018-50015018 on Build GRCh38
rs548871638 Chr.13: 50014636-50014636 on Build GRCh38
rs200009739 Chr.13: 50014806-50014808 on Build GRCh38
rs555220364 Chr.13: 50014858-50014858 on Build GRCh38
rs970207315 Chr.13: 50014535-50014538 on Build GRCh38
rs1003625226 Chr.13: 50014644-50014644 on Build GRCh38
rs752112091 Chr.13: 50014720-50014720 on Build GRCh38
rs1001652242 Chr.13: 50014725-50014725 on Build GRCh38
rs192136233 Chr.13: 50014892-50014892 on Build GRCh38
rs912758504 Chr.13: 50014660-50014661 on Build GRCh38
rs1043096154 Chr.13: 50014695-50014695 on Build GRCh38
rs921671895 Chr.13: 50014633-50014634 on Build GRCh38
rs1001767483 Chr.13: 50014539-50014543 on Build GRCh38
rs977176158 Chr.13: 50014936-50014936 on Build GRCh38
rs1053378224 Chr.13: 50014684-50014685 on Build GRCh38
rs531394373 Chr.13: 50014832-50014832 on Build GRCh38
rs1018950948 Chr.13: 50014867-50014867 on Build GRCh38
rs764147325 Chr.13: 50014692-50014692 on Build GRCh38
rs931781934 Chr.13: 50014639-50014640 on Build GRCh38
rs911473329 Chr.13: 50014522-50014522 on Build GRCh38
rs1022034047 Chr.13: 50014749-50014749 on Build GRCh38
rs907521892 Chr.13: 50014584-50014584 on Build GRCh38
rs905930803 Chr.13: 50014706-50014706 on Build GRCh38
rs891756016 Chr.13: 50014685-50014688 on Build GRCh38
rs963364007 Chr.13: 50014826-50014826 on Build GRCh38
rs944247130 Chr.13: 50014667-50014667 on Build GRCh38
rs571413782 Chr.13: 50014854-50014854 on Build GRCh38
rs963379991 Chr.13: 50014603-50014603 on Build GRCh38
rs1044996082 Chr.13: 50014587-50014587 on Build GRCh38

View Full Product Details