Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 93450346-93450892 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM120AOS
Entrez Gene ID
158293
Gene Name
family with sequence similarity 120A opposite strand
Gene Aliases
C9orf10OS
UniGene
Hs.350364
Gene Chromosome Location
Chr.9: 93446500-93453592 on Build GRCh38
Transcripts
NM_001322224.1 NM_198841.2 NR_136229.1 NR_136230.1 NR_136231.1 NR_136232.1 NR_136233.1 NR_136234.1 NR_136235.1 NR_136236.1 NR_136237.1 NR_136238.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2732461 c.683A>G p.K228R Chr.9: 93450480-93450480 on Build GRCh38
6253992 c.620C>A p.P207H Chr.9: 93450543-93450543 on Build GRCh38
3849100 c.602T>G p.V201G Chr.9: 93450561-93450561 on Build GRCh38
6658214 c.591C>T p.N197N Chr.9: 93450572-93450572 on Build GRCh38
6567691 c.575G>A p.R192K Chr.9: 93450588-93450588 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780265385 Chr.9: 93450720-93450720 on Build GRCh38
rs754894917 Chr.9: 93450495-93450495 on Build GRCh38
rs781358857 Chr.9: 93450727-93450727 on Build GRCh38
rs985142741 Chr.9: 93450743-93450743 on Build GRCh38
rs765798712 Chr.9: 93450702-93450702 on Build GRCh38
rs748845603 Chr.9: 93450805-93450805 on Build GRCh38
rs371621274 Chr.9: 93450398-93450398 on Build GRCh38
rs1031290193 Chr.9: 93450659-93450659 on Build GRCh38
rs41274386 Chr.9: 93450531-93450531 on Build GRCh38
rs932389735 Chr.9: 93450416-93450416 on Build GRCh38
rs372881078 Chr.9: 93450620-93450620 on Build GRCh38
rs780681354 Chr.9: 93450431-93450431 on Build GRCh38
rs776645181 Chr.9: 93450618-93450618 on Build GRCh38
rs3815563 Chr.9: 93450864-93450864 on Build GRCh38
rs560747344 Chr.9: 93450715-93450715 on Build GRCh38
rs376076312 Chr.9: 93450545-93450545 on Build GRCh38
rs755085145 Chr.9: 93450724-93450724 on Build GRCh38
rs746102004 Chr.9: 93450491-93450491 on Build GRCh38
rs774323185 Chr.9: 93450804-93450804 on Build GRCh38
rs572908467 Chr.9: 93450425-93450425 on Build GRCh38
rs751413654 Chr.9: 93450414-93450414 on Build GRCh38
rs976249894 Chr.9: 93450701-93450701 on Build GRCh38
rs769836381 Chr.9: 93450740-93450740 on Build GRCh38
rs533083153 Chr.9: 93450627-93450627 on Build GRCh38
rs966677101 Chr.9: 93450663-93450663 on Build GRCh38
rs968040242 Chr.9: 93450624-93450624 on Build GRCh38
rs199802049 Chr.9: 93450450-93450450 on Build GRCh38
rs768600004 Chr.9: 93450519-93450519 on Build GRCh38
rs756598879 Chr.9: 93450446-93450446 on Build GRCh38
rs754562714 Chr.9: 93450429-93450429 on Build GRCh38
rs755688003 Chr.9: 93450552-93450552 on Build GRCh38
rs748584897 Chr.9: 93450701-93450702 on Build GRCh38
rs965486809 Chr.9: 93450844-93450844 on Build GRCh38
rs1003403843 Chr.9: 93450526-93450526 on Build GRCh38
rs780374376 Chr.9: 93450583-93450583 on Build GRCh38
rs371620525 Chr.9: 93450635-93450635 on Build GRCh38
rs77711372 Chr.9: 93450561-93450561 on Build GRCh38
rs976218656 Chr.9: 93450410-93450410 on Build GRCh38
rs774165687 Chr.9: 93450625-93450625 on Build GRCh38
rs756285651 Chr.9: 93450442-93450442 on Build GRCh38
rs941284129 Chr.9: 93450496-93450496 on Build GRCh38
rs1037087902 Chr.9: 93450814-93450814 on Build GRCh38
rs144779571 Chr.9: 93450500-93450500 on Build GRCh38
rs768851148 Chr.9: 93450611-93450611 on Build GRCh38
rs752266830 Chr.9: 93450551-93450551 on Build GRCh38
rs1004360160 Chr.9: 93450840-93450840 on Build GRCh38
rs949427489 Chr.9: 93450771-93450771 on Build GRCh38
rs369518147 Chr.9: 93450521-93450521 on Build GRCh38
rs748189092 Chr.9: 93450736-93450736 on Build GRCh38
rs1056333231 Chr.9: 93450851-93450851 on Build GRCh38
rs921219018 Chr.9: 93450558-93450558 on Build GRCh38
rs41274388 Chr.9: 93450682-93450682 on Build GRCh38
rs948124156 Chr.9: 93450856-93450856 on Build GRCh38
rs1027204233 Chr.9: 93450829-93450829 on Build GRCh38
rs757780907 Chr.9: 93450574-93450574 on Build GRCh38
rs909615803 Chr.9: 93450795-93450795 on Build GRCh38
rs138719486 Chr.9: 93450518-93450518 on Build GRCh38
rs561960307 Chr.9: 93450487-93450487 on Build GRCh38
rs771489242 Chr.9: 93450485-93450485 on Build GRCh38
rs373071458 Chr.9: 93450522-93450522 on Build GRCh38
rs749725690 Chr.9: 93450478-93450478 on Build GRCh38
rs1023909815 Chr.9: 93450404-93450404 on Build GRCh38
rs758994985 Chr.9: 93450615-93450615 on Build GRCh38
rs547510368 Chr.9: 93450427-93450427 on Build GRCh38
rs887275639 Chr.9: 93450838-93450838 on Build GRCh38
rs201449587 Chr.9: 93450581-93450581 on Build GRCh38
rs527836678 Chr.9: 93450716-93450716 on Build GRCh38
rs747318347 Chr.9: 93450723-93450723 on Build GRCh38
rs368163670 Chr.9: 93450626-93450626 on Build GRCh38
rs894939416 Chr.9: 93450855-93450855 on Build GRCh38
rs779425724 Chr.9: 93450578-93450578 on Build GRCh38
rs370424829 Chr.9: 93450630-93450630 on Build GRCh38
rs746196889 Chr.9: 93450579-93450579 on Build GRCh38
rs752207973 Chr.9: 93450432-93450432 on Build GRCh38
rs754357428 Chr.9: 93450570-93450570 on Build GRCh38
rs939223587 Chr.9: 93450845-93450845 on Build GRCh38
rs1036986085 Chr.9: 93450503-93450503 on Build GRCh38
rs766224553 Chr.9: 93450524-93450524 on Build GRCh38
rs953799499 Chr.9: 93450737-93450737 on Build GRCh38
rs549699478 Chr.9: 93450822-93450822 on Build GRCh38
rs747171350 Chr.9: 93450591-93450591 on Build GRCh38
rs995711388 Chr.9: 93450438-93450438 on Build GRCh38
rs868647846 Chr.9: 93450490-93450490 on Build GRCh38
rs760240488 Chr.9: 93450632-93450632 on Build GRCh38
rs142905434 Chr.9: 93450603-93450604 on Build GRCh38
rs557946465 Chr.9: 93450417-93450417 on Build GRCh38
rs905482222 Chr.9: 93450811-93450811 on Build GRCh38
rs182605932 Chr.9: 93450383-93450383 on Build GRCh38
rs778391977 Chr.9: 93450455-93450455 on Build GRCh38
rs1019387356 Chr.9: 93450841-93450841 on Build GRCh38
rs780472991 Chr.9: 93450487-93450487 on Build GRCh38
rs775745010 Chr.9: 93450517-93450517 on Build GRCh38
rs146281455 Chr.9: 93450570-93450570 on Build GRCh38
rs779280624 Chr.9: 93450696-93450698 on Build GRCh38
rs1013967626 Chr.9: 93450384-93450384 on Build GRCh38
rs763507041 Chr.9: 93450557-93450557 on Build GRCh38
rs753229357 Chr.9: 93450642-93450642 on Build GRCh38
rs757860614 Chr.9: 93450650-93450650 on Build GRCh38

View Full Product Details